Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43318333-43318841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
API5
Entrez Gene ID
8539
Gene Name
apoptosis inhibitor 5
Gene Aliases
AAC-11, AAC11
UniGene
Hs.435771
Gene Chromosome Location
Chr.11: 43311955-43344530 on Build GRCh38
Transcripts
NM_001142930.1 NM_001142931.1 NM_001243747.1 NM_006595.3 NR_024625.1 XM_006718359.3 XM_017018464.1 XM_017018465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4601418 c.141A>T p.Q47H Chr.11: 43318711-43318711 on Build GRCh38
6602151 c.197C>T p.A66V Chr.11: 43318767-43318767 on Build GRCh38
1978928 c.139C>T p.Q47* Chr.11: 43318709-43318709 on Build GRCh38
1978929 c.139C>T p.Q47* Chr.11: 43318709-43318709 on Build GRCh38
6336964 c.94T>C p.L32L Chr.11: 43318664-43318664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751077949 Chr.11: 43318483-43318486 on Build GRCh38
rs1033403752 Chr.11: 43318359-43318359 on Build GRCh38
rs927800744 Chr.11: 43318575-43318575 on Build GRCh38
rs751614535 Chr.11: 43318609-43318609 on Build GRCh38
rs748944289 Chr.11: 43318722-43318722 on Build GRCh38
rs5743215 Chr.11: 43318526-43318526 on Build GRCh38
rs374835296 Chr.11: 43318711-43318711 on Build GRCh38
rs768611140 Chr.11: 43318499-43318499 on Build GRCh38
rs200771952 Chr.11: 43318650-43318650 on Build GRCh38
rs775981581 Chr.11: 43318460-43318460 on Build GRCh38
rs772587038 Chr.11: 43318610-43318610 on Build GRCh38
rs772933662 Chr.11: 43318720-43318720 on Build GRCh38
rs976621103 Chr.11: 43318483-43318483 on Build GRCh38
rs909792066 Chr.11: 43318742-43318742 on Build GRCh38
rs1049820644 Chr.11: 43318568-43318568 on Build GRCh38
rs769011217 Chr.11: 43318687-43318687 on Build GRCh38
rs750788233 Chr.11: 43318670-43318670 on Build GRCh38
rs533276159 Chr.11: 43318562-43318562 on Build GRCh38
rs989126269 Chr.11: 43318447-43318447 on Build GRCh38
rs913550238 Chr.11: 43318451-43318451 on Build GRCh38
rs747563359 Chr.11: 43318686-43318686 on Build GRCh38
rs35999189 Chr.11: 43318683-43318683 on Build GRCh38
rs1037071192 Chr.11: 43318678-43318678 on Build GRCh38
rs373732162 Chr.11: 43318592-43318592 on Build GRCh38
rs370431495 Chr.11: 43318613-43318613 on Build GRCh38
rs770911252 Chr.11: 43318738-43318738 on Build GRCh38
rs759561318 Chr.11: 43318774-43318774 on Build GRCh38
rs757532643 Chr.11: 43318647-43318647 on Build GRCh38
rs759381941 Chr.11: 43318612-43318612 on Build GRCh38
rs559055377 Chr.11: 43318729-43318729 on Build GRCh38
rs780378979 Chr.11: 43318685-43318685 on Build GRCh38
rs767320246 Chr.11: 43318796-43318796 on Build GRCh38
rs780783169 Chr.11: 43318697-43318697 on Build GRCh38
rs778225365 Chr.11: 43318798-43318798 on Build GRCh38
rs773164718 Chr.11: 43318491-43318491 on Build GRCh38
rs762780384 Chr.11: 43318515-43318515 on Build GRCh38
rs779047063 Chr.11: 43318657-43318657 on Build GRCh38
rs764800884 Chr.11: 43318590-43318590 on Build GRCh38
rs774269010 Chr.11: 43318745-43318745 on Build GRCh38
rs957944231 Chr.11: 43318393-43318393 on Build GRCh38
rs545438952 Chr.11: 43318584-43318584 on Build GRCh38
rs569287991 Chr.11: 43318470-43318470 on Build GRCh38
rs769501251 Chr.11: 43318715-43318715 on Build GRCh38
rs758505097 Chr.11: 43318608-43318609 on Build GRCh38
rs761715433 Chr.11: 43318556-43318556 on Build GRCh38
rs879180404 Chr.11: 43318375-43318375 on Build GRCh38
rs764303029 Chr.11: 43318626-43318627 on Build GRCh38
rs536271922 Chr.11: 43318471-43318471 on Build GRCh38
rs758458370 Chr.11: 43318382-43318382 on Build GRCh38
rs1010987285 Chr.11: 43318469-43318469 on Build GRCh38
rs888499945 Chr.11: 43318576-43318576 on Build GRCh38
rs758826178 Chr.11: 43318675-43318675 on Build GRCh38
rs753834258 Chr.11: 43318619-43318619 on Build GRCh38
rs370323707 Chr.11: 43318571-43318571 on Build GRCh38

View Full Product Details