Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61755310-61755803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434K028
Entrez Gene ID
26070
Gene Name
uncharacterized LOC26070
Gene Aliases
-
UniGene
Hs.564254
Gene Chromosome Location
Chr.11: 61754029-61757664 on Build GRCh38
Transcripts
NR_026882.1 NM_001127392.2 NM_013279.3 XM_005274222.1 XM_005274223.1 XM_005274224.1 XM_005274225.1 XM_005274226.1 XM_005274227.1 XM_005274228.1 XM_011545234.2

Gene Symbol
MYRF
Entrez Gene ID
745
Gene Name
myelin regulatory factor
Gene Aliases
C11orf9, MRF, Ndt80, pqn-47
UniGene
Hs.473109
Gene Chromosome Location
Chr.11: 61752617-61788518 on Build GRCh38
Transcripts
NR_026882.1 NM_001127392.2 NM_013279.3 XM_005274222.1 XM_005274223.1 XM_005274224.1 XM_005274225.1 XM_005274226.1 XM_005274227.1 XM_005274228.1 XM_011545234.2

SNP Detail

SNP ID Chromosome Location
rs776034949 Chr.11: 61755539-61755539 on Build GRCh38
rs540460365 Chr.11: 61755586-61755586 on Build GRCh38
rs541270014 Chr.11: 61755536-61755536 on Build GRCh38
rs982043551 Chr.11: 61755328-61755328 on Build GRCh38
rs574226184 Chr.11: 61755686-61755686 on Build GRCh38
rs780125154 Chr.11: 61755425-61755425 on Build GRCh38
rs760768348 Chr.11: 61755449-61755449 on Build GRCh38
rs768873948 Chr.11: 61755429-61755429 on Build GRCh38
rs938054884 Chr.11: 61755345-61755345 on Build GRCh38
rs973280164 Chr.11: 61755713-61755713 on Build GRCh38
rs557997427 Chr.11: 61755482-61755482 on Build GRCh38
rs918758104 Chr.11: 61755720-61755720 on Build GRCh38
rs749571634 Chr.11: 61755427-61755427 on Build GRCh38
rs770211856 Chr.11: 61755649-61755649 on Build GRCh38
rs373749424 Chr.11: 61755448-61755448 on Build GRCh38
rs942992325 Chr.11: 61755751-61755751 on Build GRCh38
rs895025152 Chr.11: 61755507-61755507 on Build GRCh38
rs1046222813 Chr.11: 61755563-61755563 on Build GRCh38
rs927971716 Chr.11: 61755342-61755342 on Build GRCh38
rs934178333 Chr.11: 61755728-61755728 on Build GRCh38
rs1038730971 Chr.11: 61755768-61755768 on Build GRCh38
rs564235176 Chr.11: 61755634-61755634 on Build GRCh38
rs1017346383 Chr.11: 61755489-61755489 on Build GRCh38
rs958048177 Chr.11: 61755627-61755627 on Build GRCh38
rs1032308580 Chr.11: 61755355-61755355 on Build GRCh38
rs770526531 Chr.11: 61755382-61755382 on Build GRCh38
rs1003413141 Chr.11: 61755632-61755632 on Build GRCh38
rs767437262 Chr.11: 61755488-61755488 on Build GRCh38
rs781186733 Chr.11: 61755416-61755416 on Build GRCh38
rs2286008 Chr.11: 61755487-61755487 on Build GRCh38
rs931006764 Chr.11: 61755712-61755712 on Build GRCh38
rs766408807 Chr.11: 61755668-61755668 on Build GRCh38
rs769407025 Chr.11: 61755459-61755459 on Build GRCh38
rs1013438086 Chr.11: 61755548-61755548 on Build GRCh38
rs757205763 Chr.11: 61755412-61755412 on Build GRCh38
rs919613020 Chr.11: 61755680-61755680 on Build GRCh38
rs990856224 Chr.11: 61755346-61755346 on Build GRCh38
rs911498683 Chr.11: 61755736-61755736 on Build GRCh38
rs961144911 Chr.11: 61755663-61755663 on Build GRCh38
rs753967868 Chr.11: 61755731-61755731 on Build GRCh38
rs751455273 Chr.11: 61755406-61755406 on Build GRCh38
rs537758871 Chr.11: 61755474-61755474 on Build GRCh38
rs962843187 Chr.11: 61755692-61755692 on Build GRCh38
rs12286908 Chr.11: 61755520-61755520 on Build GRCh38
rs933492369 Chr.11: 61755426-61755426 on Build GRCh38
rs941509720 Chr.11: 61755366-61755366 on Build GRCh38
rs972454144 Chr.11: 61755672-61755672 on Build GRCh38
rs376462837 Chr.11: 61755385-61755385 on Build GRCh38
rs772724412 Chr.11: 61755443-61755443 on Build GRCh38
rs1020793417 Chr.11: 61755585-61755585 on Build GRCh38
rs893574153 Chr.11: 61755495-61755495 on Build GRCh38
rs560374226 Chr.11: 61755603-61755603 on Build GRCh38
rs762695558 Chr.11: 61755469-61755469 on Build GRCh38
rs1055037704 Chr.11: 61755496-61755496 on Build GRCh38
rs577873423 Chr.11: 61755651-61755651 on Build GRCh38
rs752465205 Chr.11: 61755387-61755387 on Build GRCh38
rs762451771 Chr.11: 61755376-61755376 on Build GRCh38
rs1011128034 Chr.11: 61755574-61755574 on Build GRCh38
rs776217831 Chr.11: 61755447-61755447 on Build GRCh38
rs756283532 Chr.11: 61755417-61755417 on Build GRCh38
rs762645062 Chr.11: 61755390-61755390 on Build GRCh38
rs376887727 Chr.11: 61755471-61755471 on Build GRCh38
rs893511818 Chr.11: 61755780-61755780 on Build GRCh38
rs763913960 Chr.11: 61755391-61755391 on Build GRCh38
rs543059287 Chr.11: 61755695-61755695 on Build GRCh38
rs978087635 Chr.11: 61755354-61755354 on Build GRCh38
rs756124535 Chr.11: 61755689-61755689 on Build GRCh38
rs1032380808 Chr.11: 61755612-61755612 on Build GRCh38
rs773369030 Chr.11: 61755379-61755382 on Build GRCh38
rs908016821 Chr.11: 61755656-61755656 on Build GRCh38
rs909996783 Chr.11: 61755347-61755347 on Build GRCh38
rs374281370 Chr.11: 61755383-61755383 on Build GRCh38
rs758968257 Chr.11: 61755449-61755449 on Build GRCh38
rs375249070 Chr.11: 61755465-61755465 on Build GRCh38
rs748617159 Chr.11: 61755431-61755431 on Build GRCh38
rs373384599 Chr.11: 61755386-61755386 on Build GRCh38

View Full Product Details