Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63827636-63828073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf84
Entrez Gene ID
144097
Gene Name
chromosome 11 open reading frame 84
Gene Aliases
-
UniGene
Hs.502793
Gene Chromosome Location
Chr.11: 63813374-63827718 on Build GRCh38
Transcripts
NM_138471.2 XM_006718437.1 XM_011544770.1 XM_011544771.1 XM_011544772.1

SNP Detail

SNP ID Chromosome Location
rs532797287 Chr.11: 63827931-63827931 on Build GRCh38
rs773562029 Chr.11: 63827790-63827790 on Build GRCh38
rs925395609 Chr.11: 63828024-63828024 on Build GRCh38
rs1033430136 Chr.11: 63827982-63827982 on Build GRCh38
rs1015448900 Chr.11: 63827662-63827662 on Build GRCh38
rs537209850 Chr.11: 63827723-63827723 on Build GRCh38
rs572154758 Chr.11: 63827864-63827864 on Build GRCh38
rs949549014 Chr.11: 63827850-63827850 on Build GRCh38
rs78517959 Chr.11: 63827902-63827902 on Build GRCh38
rs1026747845 Chr.11: 63827895-63827895 on Build GRCh38
rs556900514 Chr.11: 63827737-63827737 on Build GRCh38
rs896751058 Chr.11: 63827836-63827836 on Build GRCh38
rs558902454 Chr.11: 63827858-63827858 on Build GRCh38
rs935366598 Chr.11: 63827823-63827823 on Build GRCh38
rs545788481 Chr.11: 63827827-63827827 on Build GRCh38
rs925248176 Chr.11: 63827758-63827758 on Build GRCh38
rs924900795 Chr.11: 63828041-63828041 on Build GRCh38
rs936169978 Chr.11: 63828043-63828043 on Build GRCh38
rs977630855 Chr.11: 63828033-63828033 on Build GRCh38
rs747872837 Chr.11: 63827928-63827928 on Build GRCh38
rs917475899 Chr.11: 63827741-63827741 on Build GRCh38
rs552632977 Chr.11: 63828037-63828037 on Build GRCh38
rs543965729 Chr.11: 63827921-63827921 on Build GRCh38
rs563731218 Chr.11: 63827922-63827922 on Build GRCh38
rs965166949 Chr.11: 63827710-63827710 on Build GRCh38
rs968596695 Chr.11: 63827998-63827998 on Build GRCh38
rs118013809 Chr.11: 63827899-63827899 on Build GRCh38
rs918167083 Chr.11: 63828039-63828039 on Build GRCh38
rs1041847300 Chr.11: 63827826-63827826 on Build GRCh38
rs775118587 Chr.11: 63827673-63827674 on Build GRCh38
rs1025082012 Chr.11: 63828011-63828011 on Build GRCh38
rs949661744 Chr.11: 63828044-63828044 on Build GRCh38
rs2285030 Chr.11: 63827745-63827745 on Build GRCh38
rs996819530 Chr.11: 63827893-63827893 on Build GRCh38
rs935300986 Chr.11: 63828025-63828025 on Build GRCh38
rs995692740 Chr.11: 63827891-63827891 on Build GRCh38
rs1045197391 Chr.11: 63828048-63828048 on Build GRCh38
rs922001099 Chr.11: 63827724-63827724 on Build GRCh38
rs924766453 Chr.11: 63827831-63827831 on Build GRCh38
rs973798732 Chr.11: 63827702-63827702 on Build GRCh38
rs962429091 Chr.11: 63827702-63827703 on Build GRCh38
rs979904206 Chr.11: 63828005-63828005 on Build GRCh38
rs904922798 Chr.11: 63827888-63827888 on Build GRCh38
rs995372140 Chr.11: 63827942-63827942 on Build GRCh38
rs1028595894 Chr.11: 63827988-63827988 on Build GRCh38
rs541104019 Chr.11: 63827883-63827883 on Build GRCh38

View Full Product Details