Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 39304629-39305086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF21A
Entrez Gene ID
55605
Gene Name
kinesin family member 21A
Gene Aliases
CFEOM1, FEOM1, FEOM3A
UniGene
Hs.374201
Gene Chromosome Location
Chr.12: 39293228-39443414 on Build GRCh38
Transcripts
NM_001173463.1 NM_001173464.1 NM_001173465.1 NM_017641.3 XM_005269007.2 XM_005269008.2 XM_005269009.2 XM_005269010.2 XM_005269011.2 XM_005269012.2 XM_005269013.2 XM_005269014.2 XM_006719493.2 XM_006719494.2 XM_011538556.2 XM_017019607.1 XM_017019608.1 XM_017019609.1 XM_017019610.1 XM_017019611.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4427090 c.4473A>G p.G1491G Chr.12: 39304911-39304911 on Build GRCh38
391100 c.4404delG p.R1468fs*8 Chr.12: 39304938-39304938 on Build GRCh38
4427089 c.4431A>G p.G1477G Chr.12: 39304911-39304911 on Build GRCh38
2065029 c.4404-1G>T p.? Chr.12: 39304939-39304939 on Build GRCh38
1255817 c.4421A>G p.K1474R Chr.12: 39304921-39304921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551194192 Chr.12: 39305044-39305044 on Build GRCh38
rs201827269 Chr.12: 39304778-39304778 on Build GRCh38
rs889069695 Chr.12: 39304981-39304981 on Build GRCh38
rs931221280 Chr.12: 39304874-39304874 on Build GRCh38
rs1018940915 Chr.12: 39305012-39305012 on Build GRCh38
rs998904148 Chr.12: 39305026-39305026 on Build GRCh38
rs775229724 Chr.12: 39304964-39304964 on Build GRCh38
rs188390387 Chr.12: 39304759-39304759 on Build GRCh38
rs772876668 Chr.12: 39304748-39304748 on Build GRCh38
rs762922377 Chr.12: 39304975-39304975 on Build GRCh38
rs375548931 Chr.12: 39304859-39304859 on Build GRCh38
rs759445300 Chr.12: 39305024-39305024 on Build GRCh38
rs368581056 Chr.12: 39304814-39304814 on Build GRCh38
rs144103791 Chr.12: 39304845-39304845 on Build GRCh38
rs62000373 Chr.12: 39304866-39304866 on Build GRCh38
rs769601884 Chr.12: 39304963-39304963 on Build GRCh38
rs751006090 Chr.12: 39304882-39304882 on Build GRCh38
rs1031676880 Chr.12: 39305044-39305045 on Build GRCh38
rs113883947 Chr.12: 39305011-39305011 on Build GRCh38
rs1021777615 Chr.12: 39304711-39304711 on Build GRCh38
rs747186773 Chr.12: 39304953-39304953 on Build GRCh38
rs756983619 Chr.12: 39304887-39304887 on Build GRCh38
rs996951649 Chr.12: 39304688-39304688 on Build GRCh38
rs767064636 Chr.12: 39304889-39304889 on Build GRCh38
rs748150530 Chr.12: 39304910-39304910 on Build GRCh38
rs774522417 Chr.12: 39304806-39304806 on Build GRCh38
rs772392400 Chr.12: 39304911-39304911 on Build GRCh38
rs779089214 Chr.12: 39304907-39304907 on Build GRCh38
rs768868658 Chr.12: 39304791-39304791 on Build GRCh38
rs780506197 Chr.12: 39304902-39304902 on Build GRCh38
rs11171689 Chr.12: 39304839-39304839 on Build GRCh38
rs750868436 Chr.12: 39304895-39304895 on Build GRCh38
rs559104078 Chr.12: 39304873-39304873 on Build GRCh38
rs780704293 Chr.12: 39304773-39304773 on Build GRCh38
rs773298629 Chr.12: 39304942-39304942 on Build GRCh38
rs1032051926 Chr.12: 39304774-39304774 on Build GRCh38
rs772148885 Chr.12: 39304827-39304827 on Build GRCh38
rs764001672 Chr.12: 39304983-39304983 on Build GRCh38
rs957789955 Chr.12: 39305045-39305045 on Build GRCh38
rs751274359 Chr.12: 39304959-39304960 on Build GRCh38
rs1023072936 Chr.12: 39304834-39304834 on Build GRCh38
rs111388318 Chr.12: 39304919-39304919 on Build GRCh38
rs775422512 Chr.12: 39304864-39304864 on Build GRCh38
rs756554787 Chr.12: 39304901-39304901 on Build GRCh38
rs184215859 Chr.12: 39304745-39304745 on Build GRCh38
rs755501569 Chr.12: 39304904-39304904 on Build GRCh38
rs886049340 Chr.12: 39304853-39304853 on Build GRCh38
rs747771381 Chr.12: 39304669-39304669 on Build GRCh38
rs760472442 Chr.12: 39305018-39305018 on Build GRCh38
rs756744884 Chr.12: 39304771-39304771 on Build GRCh38

View Full Product Details