Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74722787-74723279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1A1
Entrez Gene ID
1543
Gene Name
cytochrome P450 family 1 subfamily A member 1
Gene Aliases
AHH, AHRR, CP11, CYP1, CYPIA1, P1-450, P450-C, P450DX
UniGene
Hs.72912
Gene Chromosome Location
Chr.15: 74719542-74725610 on Build GRCh38
Transcripts
NM_000499.4 NM_001319216.1 NM_001319217.1 XM_017021953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4463380 c.128C>T p.P43L Chr.15: 74722970-74722970 on Build GRCh38
964886 c.112A>G p.K38E Chr.15: 74722986-74722986 on Build GRCh38
1284384 c.37_38CT>AC p.L13T Chr.15: 74723060-74723061 on Build GRCh38
5731594 c.260G>A p.S87N Chr.15: 74722838-74722838 on Build GRCh38
964887 c.52A>G p.I18V Chr.15: 74723046-74723046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754114915 Chr.15: 74722957-74722958 on Build GRCh38
rs755133321 Chr.15: 74723072-74723072 on Build GRCh38
rs28399425 Chr.15: 74723130-74723130 on Build GRCh38
rs773817962 Chr.15: 74722917-74722917 on Build GRCh38
rs764836276 Chr.15: 74723086-74723086 on Build GRCh38
rs768956417 Chr.15: 74722851-74722851 on Build GRCh38
rs45528935 Chr.15: 74722820-74722820 on Build GRCh38
rs148505285 Chr.15: 74723075-74723075 on Build GRCh38
rs753812224 Chr.15: 74723069-74723069 on Build GRCh38
rs769983960 Chr.15: 74722829-74722829 on Build GRCh38
rs888842899 Chr.15: 74723210-74723210 on Build GRCh38
rs747317438 Chr.15: 74723001-74723001 on Build GRCh38
rs994989002 Chr.15: 74723177-74723177 on Build GRCh38
rs112517897 Chr.15: 74722889-74722889 on Build GRCh38
rs35035798 Chr.15: 74722902-74722902 on Build GRCh38
rs56105568 Chr.15: 74723161-74723161 on Build GRCh38
rs897480465 Chr.15: 74723172-74723172 on Build GRCh38
rs893613070 Chr.15: 74722835-74722835 on Build GRCh38
rs780078658 Chr.15: 74722960-74722960 on Build GRCh38
rs370225248 Chr.15: 74722885-74722885 on Build GRCh38
rs757365807 Chr.15: 74723145-74723146 on Build GRCh38
rs749328902 Chr.15: 74722897-74722897 on Build GRCh38
rs888665442 Chr.15: 74723216-74723216 on Build GRCh38
rs755005663 Chr.15: 74722965-74722965 on Build GRCh38
rs754416936 Chr.15: 74722806-74722806 on Build GRCh38
rs770299285 Chr.15: 74723122-74723122 on Build GRCh38
rs762383674 Chr.15: 74722927-74722927 on Build GRCh38
rs780925747 Chr.15: 74722826-74722826 on Build GRCh38
rs184128730 Chr.15: 74723068-74723068 on Build GRCh38
rs779251470 Chr.15: 74722969-74722969 on Build GRCh38
rs761407107 Chr.15: 74723037-74723037 on Build GRCh38
rs777905948 Chr.15: 74723079-74723079 on Build GRCh38
rs746029569 Chr.15: 74722827-74722827 on Build GRCh38
rs761140013 Chr.15: 74722919-74722919 on Build GRCh38
rs751160077 Chr.15: 74722948-74722948 on Build GRCh38
rs750100688 Chr.15: 74723251-74723251 on Build GRCh38
rs913685889 Chr.15: 74722928-74722928 on Build GRCh38
rs749494772 Chr.15: 74722837-74722837 on Build GRCh38
rs371190271 Chr.15: 74722869-74722869 on Build GRCh38
rs1007321699 Chr.15: 74723244-74723244 on Build GRCh38
rs4646422 Chr.15: 74722964-74722964 on Build GRCh38
rs774097027 Chr.15: 74723028-74723028 on Build GRCh38
rs946563307 Chr.15: 74723105-74723105 on Build GRCh38
rs756138124 Chr.15: 74722958-74722958 on Build GRCh38
rs926026682 Chr.15: 74722999-74722999 on Build GRCh38
rs578251068 Chr.15: 74722921-74722921 on Build GRCh38
rs761719283 Chr.15: 74722949-74722949 on Build GRCh38
rs138474634 Chr.15: 74722856-74722856 on Build GRCh38
rs17861094 Chr.15: 74722865-74722865 on Build GRCh38
rs778933231 Chr.15: 74722900-74722900 on Build GRCh38
rs554072118 Chr.15: 74722874-74722874 on Build GRCh38
rs1040038759 Chr.15: 74723167-74723167 on Build GRCh38
rs764934348 Chr.15: 74722813-74722813 on Build GRCh38
rs77425771 Chr.15: 74722836-74722836 on Build GRCh38
rs994461848 Chr.15: 74723191-74723191 on Build GRCh38
rs149846182 Chr.15: 74722922-74722922 on Build GRCh38
rs2229150 Chr.15: 74722821-74722821 on Build GRCh38
rs576805455 Chr.15: 74723074-74723074 on Build GRCh38
rs952982161 Chr.15: 74722887-74722887 on Build GRCh38
rs1047768353 Chr.15: 74723204-74723204 on Build GRCh38
rs758562697 Chr.15: 74723078-74723078 on Build GRCh38
rs767189328 Chr.15: 74722950-74722950 on Build GRCh38
rs143070677 Chr.15: 74722914-74722914 on Build GRCh38
rs149262994 Chr.15: 74723162-74723162 on Build GRCh38
rs748295448 Chr.15: 74722972-74722972 on Build GRCh38
rs763875538 Chr.15: 74722935-74722935 on Build GRCh38
rs769009780 Chr.15: 74722898-74722898 on Build GRCh38
rs757606619 Chr.15: 74723085-74723085 on Build GRCh38
rs771432878 Chr.15: 74723013-74723013 on Build GRCh38
rs767145702 Chr.15: 74723040-74723040 on Build GRCh38
rs994552466 Chr.15: 74722833-74722833 on Build GRCh38
rs895831176 Chr.15: 74723098-74723098 on Build GRCh38
rs375315082 Chr.15: 74723027-74723027 on Build GRCh38
rs758238061 Chr.15: 74722818-74722818 on Build GRCh38
rs537005466 Chr.15: 74722854-74722854 on Build GRCh38
rs777119337 Chr.15: 74723018-74723018 on Build GRCh38
rs373246855 Chr.15: 74722859-74722859 on Build GRCh38
rs548705429 Chr.15: 74723023-74723023 on Build GRCh38
rs376399672 Chr.15: 74722884-74722884 on Build GRCh38
rs145924908 Chr.15: 74723026-74723026 on Build GRCh38
rs565370983 Chr.15: 74722952-74722952 on Build GRCh38
rs560039501 Chr.15: 74723084-74723084 on Build GRCh38
rs746333343 Chr.15: 74723113-74723113 on Build GRCh38
rs969437424 Chr.15: 74722930-74722930 on Build GRCh38
rs112522537 Chr.15: 74722852-74722852 on Build GRCh38
rs373568981 Chr.15: 74722881-74722881 on Build GRCh38
rs778141626 Chr.15: 74722996-74722996 on Build GRCh38
rs1005741715 Chr.15: 74723236-74723236 on Build GRCh38
rs543414137 Chr.15: 74723047-74723047 on Build GRCh38
rs572138243 Chr.15: 74722893-74722893 on Build GRCh38
rs750252519 Chr.15: 74723041-74723041 on Build GRCh38
rs55864856 Chr.15: 74722858-74722858 on Build GRCh38
rs868017066 Chr.15: 74722810-74722810 on Build GRCh38
rs139919943 Chr.15: 74722882-74722882 on Build GRCh38
rs982120784 Chr.15: 74722938-74722938 on Build GRCh38
rs373137650 Chr.15: 74722814-74722814 on Build GRCh38
rs772526583 Chr.15: 74722987-74722987 on Build GRCh38

View Full Product Details