Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75339882-75340382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD4
Entrez Gene ID
54939
Gene Name
COMM domain containing 4
Gene Aliases
-
UniGene
Hs.351327
Gene Chromosome Location
Chr.15: 75335996-75341927 on Build GRCh38
Transcripts
NM_001284377.1 NM_001284378.1 NM_001284379.1 NM_001321844.1 NM_001321845.1 NM_001321846.1 NM_001321847.1 NM_001321848.1 NM_017828.4 NR_104312.1 NR_104313.1 XM_005254509.3 XM_005254510.3 XM_011521739.1 XM_011521740.1 XM_011521741.1 XM_017022385.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5164132 c.559+7C>T p.? Chr.15: 75339885-75339885 on Build GRCh38
3504067 c.579C>T p.T193T Chr.15: 75339984-75339984 on Build GRCh38
4408134 c.594G>A p.L198L Chr.15: 75339999-75339999 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377076813 Chr.15: 75339983-75339983 on Build GRCh38
rs765826666 Chr.15: 75339994-75339994 on Build GRCh38
rs760258635 Chr.15: 75340052-75340052 on Build GRCh38
rs752474029 Chr.15: 75340004-75340004 on Build GRCh38
rs1055696205 Chr.15: 75340313-75340313 on Build GRCh38
rs935834857 Chr.15: 75339977-75339977 on Build GRCh38
rs528036439 Chr.15: 75340346-75340346 on Build GRCh38
rs774207362 Chr.15: 75340226-75340226 on Build GRCh38
rs898641112 Chr.15: 75340077-75340077 on Build GRCh38
rs932720173 Chr.15: 75340224-75340224 on Build GRCh38
rs758611975 Chr.15: 75339912-75339912 on Build GRCh38
rs746027459 Chr.15: 75340027-75340027 on Build GRCh38
rs553260938 Chr.15: 75340084-75340084 on Build GRCh38
rs1033883305 Chr.15: 75340094-75340094 on Build GRCh38
rs955356638 Chr.15: 75340136-75340136 on Build GRCh38
rs898727392 Chr.15: 75340271-75340271 on Build GRCh38
rs747066136 Chr.15: 75339925-75339925 on Build GRCh38
rs202141846 Chr.15: 75339995-75339995 on Build GRCh38
rs79543560 Chr.15: 75339902-75339902 on Build GRCh38
rs369615128 Chr.15: 75339932-75339932 on Build GRCh38
rs866512905 Chr.15: 75340088-75340088 on Build GRCh38
rs775713962 Chr.15: 75339944-75339944 on Build GRCh38
rs940041733 Chr.15: 75340237-75340237 on Build GRCh38
rs575226079 Chr.15: 75340286-75340286 on Build GRCh38
rs147946131 Chr.15: 75340046-75340046 on Build GRCh38
rs781690563 Chr.15: 75339937-75339937 on Build GRCh38
rs770903019 Chr.15: 75340117-75340117 on Build GRCh38
rs758236034 Chr.15: 75340007-75340007 on Build GRCh38
rs879362418 Chr.15: 75340048-75340048 on Build GRCh38
rs990376291 Chr.15: 75340192-75340192 on Build GRCh38
rs188362483 Chr.15: 75340091-75340091 on Build GRCh38
rs770407524 Chr.15: 75340053-75340053 on Build GRCh38
rs775996154 Chr.15: 75340057-75340057 on Build GRCh38
rs3803465 Chr.15: 75340019-75340019 on Build GRCh38
rs1023967434 Chr.15: 75340116-75340116 on Build GRCh38
rs149829938 Chr.15: 75339988-75339988 on Build GRCh38
rs117112913 Chr.15: 75339939-75339939 on Build GRCh38
rs1031610480 Chr.15: 75340149-75340149 on Build GRCh38
rs928160178 Chr.15: 75340285-75340285 on Build GRCh38
rs991552116 Chr.15: 75339917-75339917 on Build GRCh38
rs12898435 Chr.15: 75339952-75339952 on Build GRCh38
rs977317629 Chr.15: 75340203-75340203 on Build GRCh38
rs781140222 Chr.15: 75340020-75340020 on Build GRCh38
rs887389811 Chr.15: 75340287-75340287 on Build GRCh38
rs770950711 Chr.15: 75339935-75339935 on Build GRCh38
rs748540807 Chr.15: 75339901-75339902 on Build GRCh38
rs915543717 Chr.15: 75340259-75340259 on Build GRCh38
rs200240397 Chr.15: 75340045-75340045 on Build GRCh38
rs921130738 Chr.15: 75340210-75340210 on Build GRCh38
rs751805173 Chr.15: 75340018-75340018 on Build GRCh38
rs895353063 Chr.15: 75340332-75340332 on Build GRCh38
rs558182510 Chr.15: 75340037-75340037 on Build GRCh38
rs573164519 Chr.15: 75340110-75340110 on Build GRCh38
rs977296345 Chr.15: 75340125-75340125 on Build GRCh38
rs941582613 Chr.15: 75340320-75340320 on Build GRCh38
rs199928624 Chr.15: 75339901-75339901 on Build GRCh38
rs1012704281 Chr.15: 75340114-75340114 on Build GRCh38
rs957197624 Chr.15: 75340150-75340150 on Build GRCh38
rs989803303 Chr.15: 75340253-75340253 on Build GRCh38
rs563728193 Chr.15: 75340345-75340345 on Build GRCh38
rs1037215690 Chr.15: 75340255-75340255 on Build GRCh38
rs546888741 Chr.15: 75340352-75340352 on Build GRCh38
rs371688081 Chr.15: 75339904-75339904 on Build GRCh38
rs765281857 Chr.15: 75339997-75339997 on Build GRCh38
rs948514599 Chr.15: 75340262-75340262 on Build GRCh38
rs1045512448 Chr.15: 75340353-75340353 on Build GRCh38
rs746378612 Chr.15: 75339938-75339938 on Build GRCh38
rs906669980 Chr.15: 75340042-75340042 on Build GRCh38
rs924404893 Chr.15: 75339959-75339959 on Build GRCh38
rs778023131 Chr.15: 75339924-75339924 on Build GRCh38
rs778585502 Chr.15: 75339903-75339903 on Build GRCh38
rs981568910 Chr.15: 75340283-75340283 on Build GRCh38
rs773326497 Chr.15: 75339942-75339942 on Build GRCh38
rs545828867 Chr.15: 75340297-75340297 on Build GRCh38
rs530783045 Chr.15: 75340196-75340196 on Build GRCh38
rs201193312 Chr.15: 75340036-75340036 on Build GRCh38
rs777934808 Chr.15: 75340013-75340013 on Build GRCh38
rs563433120 Chr.15: 75340231-75340231 on Build GRCh38

View Full Product Details