Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7351768-7352266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD11
Entrez Gene ID
147040
Gene Name
potassium channel tetramerization domain containing 11
Gene Aliases
C17orf36, KCASH1, REN, REN/KCTD11
UniGene
Hs.592112
Gene Chromosome Location
Chr.17: 7351889-7354944 on Build GRCh38
Transcripts
NM_001002914.2

SNP Detail

SNP ID Chromosome Location
rs947132041 Chr.17: 7352084-7352084 on Build GRCh38
rs1018578886 Chr.17: 7351803-7351803 on Build GRCh38
rs1026286350 Chr.17: 7352166-7352166 on Build GRCh38
rs923178437 Chr.17: 7352051-7352051 on Build GRCh38
rs984887287 Chr.17: 7352023-7352023 on Build GRCh38
rs1002349489 Chr.17: 7351918-7351918 on Build GRCh38
rs887795324 Chr.17: 7352180-7352180 on Build GRCh38
rs545206417 Chr.17: 7352058-7352058 on Build GRCh38
rs1044105736 Chr.17: 7352093-7352093 on Build GRCh38
rs1007044326 Chr.17: 7352208-7352208 on Build GRCh38
rs577851505 Chr.17: 7352031-7352031 on Build GRCh38
rs879123199 Chr.17: 7351989-7351989 on Build GRCh38
rs927042098 Chr.17: 7352232-7352232 on Build GRCh38
rs930873035 Chr.17: 7351995-7351995 on Build GRCh38
rs905649363 Chr.17: 7352105-7352105 on Build GRCh38
rs1043844064 Chr.17: 7351903-7351903 on Build GRCh38
rs974748830 Chr.17: 7352067-7352067 on Build GRCh38
rs984603542 Chr.17: 7351806-7351806 on Build GRCh38
rs923209720 Chr.17: 7351857-7351857 on Build GRCh38
rs990638700 Chr.17: 7352152-7352152 on Build GRCh38
rs999576831 Chr.17: 7351939-7351939 on Build GRCh38
rs920319997 Chr.17: 7351829-7351829 on Build GRCh38
rs964723114 Chr.17: 7351804-7351804 on Build GRCh38
rs891297837 Chr.17: 7351969-7351969 on Build GRCh38
rs934333190 Chr.17: 7351861-7351861 on Build GRCh38
rs963015641 Chr.17: 7352048-7352048 on Build GRCh38
rs927755648 Chr.17: 7352099-7352099 on Build GRCh38
rs973120721 Chr.17: 7351827-7351827 on Build GRCh38
rs952267345 Chr.17: 7351802-7351802 on Build GRCh38
rs935360360 Chr.17: 7352190-7352190 on Build GRCh38
rs902528287 Chr.17: 7351929-7351929 on Build GRCh38
rs1056598205 Chr.17: 7352121-7352121 on Build GRCh38
rs983122008 Chr.17: 7352082-7352082 on Build GRCh38
rs561080627 Chr.17: 7352203-7352203 on Build GRCh38
rs946627844 Chr.17: 7352162-7352162 on Build GRCh38
rs929046896 Chr.17: 7351832-7351832 on Build GRCh38
rs916420888 Chr.17: 7352158-7352158 on Build GRCh38
rs1018928398 Chr.17: 7352037-7352037 on Build GRCh38
rs1011747914 Chr.17: 7351899-7351899 on Build GRCh38
rs938436794 Chr.17: 7352238-7352238 on Build GRCh38
rs927765297 Chr.17: 7351872-7351872 on Build GRCh38
rs961282144 Chr.17: 7351927-7351927 on Build GRCh38
rs796274463 Chr.17: 7352065-7352065 on Build GRCh38
rs946691115 Chr.17: 7351905-7351905 on Build GRCh38
rs939188136 Chr.17: 7351874-7351874 on Build GRCh38
rs549274568 Chr.17: 7352201-7352201 on Build GRCh38
rs1043710153 Chr.17: 7351909-7351909 on Build GRCh38
rs896696639 Chr.17: 7352139-7352139 on Build GRCh38
rs573588624 Chr.17: 7351930-7351930 on Build GRCh38
rs544885660 Chr.17: 7351790-7351790 on Build GRCh38
rs185745499 Chr.17: 7351851-7351851 on Build GRCh38
rs773968548 Chr.17: 7352223-7352223 on Build GRCh38
rs1035221539 Chr.17: 7351921-7351921 on Build GRCh38
rs955533281 Chr.17: 7351797-7351797 on Build GRCh38
rs1018419581 Chr.17: 7352217-7352217 on Build GRCh38
rs979771113 Chr.17: 7352230-7352230 on Build GRCh38
rs527554461 Chr.17: 7352189-7352189 on Build GRCh38
rs905387623 Chr.17: 7351914-7351914 on Build GRCh38
rs1030585159 Chr.17: 7352207-7352207 on Build GRCh38
rs1029755921 Chr.17: 7351953-7351953 on Build GRCh38
rs1054962024 Chr.17: 7351882-7351882 on Build GRCh38
rs902473916 Chr.17: 7352176-7352176 on Build GRCh38
rs938174355 Chr.17: 7352110-7352110 on Build GRCh38
rs895094354 Chr.17: 7351883-7351883 on Build GRCh38
rs560415436 Chr.17: 7352075-7352075 on Build GRCh38
rs1006252048 Chr.17: 7352188-7352188 on Build GRCh38
rs1027291334 Chr.17: 7352071-7352071 on Build GRCh38
rs1044042649 Chr.17: 7352166-7352166 on Build GRCh38
rs543923105 Chr.17: 7351962-7351962 on Build GRCh38
rs939176499 Chr.17: 7352139-7352139 on Build GRCh38
rs985671652 Chr.17: 7351799-7351799 on Build GRCh38

View Full Product Details