Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36577892-36578396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGNBP2
Entrez Gene ID
79893
Gene Name
gametogenetin binding protein 2
Gene Aliases
DIF-3, DIF3, LCRG1, LZK1, ZFP403, ZNF403
UniGene
Hs.514116
Gene Chromosome Location
Chr.17: 36544888-36589848 on Build GRCh38
Transcripts
NM_024835.4 XM_005257689.2 XM_011525265.2 XM_011525266.1 XM_017025107.1

SNP Detail

SNP ID Chromosome Location
rs767476497 Chr.17: 36578093-36578093 on Build GRCh38
rs750076611 Chr.17: 36578111-36578111 on Build GRCh38
rs115395253 Chr.17: 36578261-36578261 on Build GRCh38
rs779251106 Chr.17: 36578024-36578024 on Build GRCh38
rs572599406 Chr.17: 36577923-36577923 on Build GRCh38
rs933417740 Chr.17: 36577919-36577919 on Build GRCh38
rs776618402 Chr.17: 36578023-36578023 on Build GRCh38
rs971250948 Chr.17: 36577942-36577942 on Build GRCh38
rs773416024 Chr.17: 36577991-36577991 on Build GRCh38
rs746771180 Chr.17: 36578235-36578236 on Build GRCh38
rs79146532 Chr.17: 36578246-36578246 on Build GRCh38
rs760866809 Chr.17: 36578003-36578003 on Build GRCh38
rs758635301 Chr.17: 36578226-36578226 on Build GRCh38
rs776723860 Chr.17: 36578223-36578223 on Build GRCh38
rs779393330 Chr.17: 36578176-36578176 on Build GRCh38
rs373802940 Chr.17: 36578187-36578187 on Build GRCh38
rs1003280826 Chr.17: 36578290-36578290 on Build GRCh38
rs948259432 Chr.17: 36578146-36578146 on Build GRCh38
rs753306931 Chr.17: 36578129-36578129 on Build GRCh38
rs757588893 Chr.17: 36577967-36577967 on Build GRCh38
rs779195030 Chr.17: 36577971-36577971 on Build GRCh38
rs529880542 Chr.17: 36578363-36578363 on Build GRCh38
rs1025230214 Chr.17: 36578302-36578302 on Build GRCh38
rs942796720 Chr.17: 36578297-36578297 on Build GRCh38
rs780061929 Chr.17: 36578231-36578231 on Build GRCh38
rs561212794 Chr.17: 36578330-36578330 on Build GRCh38
rs954766085 Chr.17: 36578357-36578357 on Build GRCh38
rs552546811 Chr.17: 36578358-36578358 on Build GRCh38
rs202093366 Chr.17: 36578210-36578210 on Build GRCh38
rs909955641 Chr.17: 36578284-36578284 on Build GRCh38
rs751342465 Chr.17: 36578047-36578047 on Build GRCh38
rs772415304 Chr.17: 36578178-36578178 on Build GRCh38
rs1054041962 Chr.17: 36578064-36578064 on Build GRCh38
rs750587575 Chr.17: 36578148-36578148 on Build GRCh38
rs772213307 Chr.17: 36577980-36577980 on Build GRCh38
rs984664401 Chr.17: 36578258-36578258 on Build GRCh38
rs975257892 Chr.17: 36578342-36578342 on Build GRCh38
rs753666774 Chr.17: 36578331-36578331 on Build GRCh38
rs201268453 Chr.17: 36578033-36578033 on Build GRCh38
rs987559848 Chr.17: 36578364-36578364 on Build GRCh38
rs1039725793 Chr.17: 36578324-36578324 on Build GRCh38
rs771024865 Chr.17: 36578015-36578015 on Build GRCh38
rs761707709 Chr.17: 36578086-36578086 on Build GRCh38
rs1005858837 Chr.17: 36578310-36578310 on Build GRCh38
rs755761821 Chr.17: 36578122-36578122 on Build GRCh38
rs201112415 Chr.17: 36578190-36578190 on Build GRCh38
rs1051765817 Chr.17: 36577965-36577965 on Build GRCh38
rs779575680 Chr.17: 36578127-36578127 on Build GRCh38
rs958475296 Chr.17: 36577918-36577918 on Build GRCh38
rs530836206 Chr.17: 36578217-36578217 on Build GRCh38
rs769074525 Chr.17: 36577955-36577955 on Build GRCh38
rs762908587 Chr.17: 36578028-36578028 on Build GRCh38
rs758547768 Chr.17: 36578185-36578185 on Build GRCh38
rs886778944 Chr.17: 36578303-36578303 on Build GRCh38
rs756045235 Chr.17: 36578305-36578305 on Build GRCh38
rs748522090 Chr.17: 36577972-36577972 on Build GRCh38
rs142083015 Chr.17: 36578327-36578327 on Build GRCh38
rs755452058 Chr.17: 36578161-36578161 on Build GRCh38
rs893941764 Chr.17: 36578069-36578069 on Build GRCh38
rs748718311 Chr.17: 36578177-36578177 on Build GRCh38
rs991106264 Chr.17: 36577922-36577922 on Build GRCh38
rs113988816 Chr.17: 36578050-36578050 on Build GRCh38
rs749770909 Chr.17: 36577927-36577927 on Build GRCh38
rs779869858 Chr.17: 36578300-36578300 on Build GRCh38
rs1029543514 Chr.17: 36578355-36578355 on Build GRCh38

View Full Product Details