Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67055965-67056473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG1
Entrez Gene ID
786
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 1
Gene Aliases
CACNLG
UniGene
Hs.147989
Gene Chromosome Location
Chr.17: 67044536-67056797 on Build GRCh38
Transcripts
NM_000727.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
231939 c.611T>C p.L204P Chr.17: 67056213-67056213 on Build GRCh38
6618683 c.465G>A p.V155V Chr.17: 67056067-67056067 on Build GRCh38
6618682 c.571C>G p.L191V Chr.17: 67056173-67056173 on Build GRCh38
1736751 c.592G>A p.A198T Chr.17: 67056194-67056194 on Build GRCh38
6618685 c.637G>C p.E213Q Chr.17: 67056239-67056239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146687061 Chr.17: 67056155-67056155 on Build GRCh38
rs114830174 Chr.17: 67056064-67056064 on Build GRCh38
rs755447133 Chr.17: 67056200-67056200 on Build GRCh38
rs551015807 Chr.17: 67056411-67056411 on Build GRCh38
rs756364163 Chr.17: 67056154-67056154 on Build GRCh38
rs374071089 Chr.17: 67056218-67056218 on Build GRCh38
rs762527259 Chr.17: 67056256-67056256 on Build GRCh38
rs112559599 Chr.17: 67056037-67056037 on Build GRCh38
rs763637036 Chr.17: 67056095-67056095 on Build GRCh38
rs989368247 Chr.17: 67056078-67056078 on Build GRCh38
rs141132419 Chr.17: 67056304-67056304 on Build GRCh38
rs371885900 Chr.17: 67056227-67056227 on Build GRCh38
rs1001250993 Chr.17: 67056350-67056350 on Build GRCh38
rs564381868 Chr.17: 67056007-67056007 on Build GRCh38
rs377293425 Chr.17: 67056186-67056186 on Build GRCh38
rs768735643 Chr.17: 67056228-67056228 on Build GRCh38
rs201378629 Chr.17: 67056085-67056085 on Build GRCh38
rs770852192 Chr.17: 67056017-67056017 on Build GRCh38
rs748305407 Chr.17: 67056141-67056141 on Build GRCh38
rs767154616 Chr.17: 67056028-67056028 on Build GRCh38
rs1024707786 Chr.17: 67056205-67056205 on Build GRCh38
rs750892136 Chr.17: 67056043-67056043 on Build GRCh38
rs780232550 Chr.17: 67056054-67056054 on Build GRCh38
rs747158989 Chr.17: 67056006-67056006 on Build GRCh38
rs1009930600 Chr.17: 67056366-67056366 on Build GRCh38
rs201720862 Chr.17: 67056290-67056290 on Build GRCh38
rs375091485 Chr.17: 67056317-67056317 on Build GRCh38
rs748932584 Chr.17: 67056293-67056293 on Build GRCh38
rs372395180 Chr.17: 67056179-67056179 on Build GRCh38
rs776597087 Chr.17: 67056184-67056184 on Build GRCh38
rs148854871 Chr.17: 67056163-67056163 on Build GRCh38
rs529288668 Chr.17: 67056400-67056400 on Build GRCh38
rs776361976 Chr.17: 67056102-67056102 on Build GRCh38
rs770409772 Chr.17: 67056260-67056260 on Build GRCh38
rs778578397 Chr.17: 67056041-67056041 on Build GRCh38
rs1021247429 Chr.17: 67056272-67056272 on Build GRCh38
rs759433254 Chr.17: 67056088-67056088 on Build GRCh38
rs781551661 Chr.17: 67056131-67056131 on Build GRCh38
rs761921218 Chr.17: 67056013-67056013 on Build GRCh38
rs761154799 Chr.17: 67056290-67056291 on Build GRCh38
rs1045923566 Chr.17: 67056369-67056369 on Build GRCh38
rs773893674 Chr.17: 67056305-67056305 on Build GRCh38
rs774331354 Chr.17: 67056019-67056019 on Build GRCh38
rs368979237 Chr.17: 67056000-67056000 on Build GRCh38
rs202044777 Chr.17: 67056316-67056316 on Build GRCh38
rs115573053 Chr.17: 67056289-67056289 on Build GRCh38
rs758974258 Chr.17: 67056045-67056045 on Build GRCh38
rs773768347 Chr.17: 67056262-67056262 on Build GRCh38
rs781345975 Chr.17: 67056286-67056286 on Build GRCh38
rs561976556 Chr.17: 67056077-67056077 on Build GRCh38
rs143055819 Chr.17: 67056092-67056092 on Build GRCh38
rs140761602 Chr.17: 67056093-67056093 on Build GRCh38
rs368387791 Chr.17: 67056002-67056002 on Build GRCh38
rs765030923 Chr.17: 67056187-67056187 on Build GRCh38
rs566817643 Chr.17: 67056224-67056224 on Build GRCh38
rs754022875 Chr.17: 67056278-67056278 on Build GRCh38
rs966960852 Chr.17: 67056385-67056385 on Build GRCh38
rs528442512 Chr.17: 67056034-67056034 on Build GRCh38
rs768730910 Chr.17: 67056061-67056061 on Build GRCh38
rs909857893 Chr.17: 67056312-67056312 on Build GRCh38
rs752571949 Chr.17: 67056032-67056032 on Build GRCh38
rs550820114 Chr.17: 67056125-67056125 on Build GRCh38
rs1799938 Chr.17: 67056188-67056188 on Build GRCh38
rs540562451 Chr.17: 67056346-67056346 on Build GRCh38
rs778861540 Chr.17: 67056222-67056222 on Build GRCh38
rs368486677 Chr.17: 67056116-67056116 on Build GRCh38
rs758382037 Chr.17: 67056281-67056281 on Build GRCh38
rs977418601 Chr.17: 67056437-67056437 on Build GRCh38
rs759091884 Chr.17: 67056315-67056315 on Build GRCh38
rs754934609 Chr.17: 67056283-67056283 on Build GRCh38
rs772505677 Chr.17: 67055999-67055999 on Build GRCh38
rs148226819 Chr.17: 67056219-67056219 on Build GRCh38
rs758809345 Chr.17: 67056124-67056124 on Build GRCh38
rs372713806 Chr.17: 67056164-67056164 on Build GRCh38
rs777436465 Chr.17: 67056292-67056292 on Build GRCh38
rs144606776 Chr.17: 67056129-67056129 on Build GRCh38
rs552916194 Chr.17: 67056255-67056255 on Build GRCh38
rs756311199 Chr.17: 67056215-67056215 on Build GRCh38
rs755429816 Chr.17: 67056130-67056130 on Build GRCh38
rs112255499 Chr.17: 67056117-67056117 on Build GRCh38
rs749974379 Chr.17: 67056119-67056119 on Build GRCh38
rs766783706 Chr.17: 67056194-67056194 on Build GRCh38
rs866479653 Chr.17: 67056063-67056063 on Build GRCh38
rs764452143 Chr.17: 67056277-67056277 on Build GRCh38
rs747450818 Chr.17: 67056059-67056059 on Build GRCh38
rs950027589 Chr.17: 67056328-67056328 on Build GRCh38
rs369927080 Chr.17: 67056039-67056052 on Build GRCh38
rs752048505 Chr.17: 67056196-67056196 on Build GRCh38
rs753087164 Chr.17: 67056211-67056211 on Build GRCh38
rs1045538497 Chr.17: 67056334-67056334 on Build GRCh38
rs562086747 Chr.17: 67056351-67056351 on Build GRCh38
rs146071095 Chr.17: 67056240-67056240 on Build GRCh38
rs557177020 Chr.17: 67056414-67056414 on Build GRCh38
rs9912133 Chr.17: 67056280-67056280 on Build GRCh38
rs760218031 Chr.17: 67056033-67056033 on Build GRCh38
rs776551009 Chr.17: 67056008-67056008 on Build GRCh38
rs746930490 Chr.17: 67056178-67056178 on Build GRCh38
rs138929263 Chr.17: 67056263-67056263 on Build GRCh38
rs950158319 Chr.17: 67056355-67056355 on Build GRCh38
rs761507529 Chr.17: 67056185-67056185 on Build GRCh38
rs897347763 Chr.17: 67056348-67056348 on Build GRCh38
rs775552129 Chr.17: 67056173-67056173 on Build GRCh38
rs747995880 Chr.17: 67056249-67056249 on Build GRCh38
rs753050742 Chr.17: 67056273-67056273 on Build GRCh38
rs762591486 Chr.17: 67056193-67056193 on Build GRCh38
rs761082724 Chr.17: 67056276-67056276 on Build GRCh38
rs148046092 Chr.17: 67056203-67056203 on Build GRCh38
rs753391778 Chr.17: 67056035-67056035 on Build GRCh38
rs754021424 Chr.17: 67056115-67056115 on Build GRCh38
rs759486518 Chr.17: 67056025-67056025 on Build GRCh38

View Full Product Details