Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54362499-54363045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
UniGene
Hs.208701
Gene Chromosome Location
Chr.18: 54357914-54382035 on Build GRCh38
Transcripts
NM_001288980.1 NM_001288981.1 NM_001288982.1 NM_173529.5 NR_110234.1 XM_005258201.1 XM_005258203.1 XM_017025567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3821580 c.719G>T p.W240L Chr.18: 54362900-54362900 on Build GRCh38
6564604 c.615G>A p.L205L Chr.18: 54362796-54362796 on Build GRCh38
6430863 c.689C>A p.S230* Chr.18: 54362870-54362870 on Build GRCh38
988988 c.626G>T p.R209I Chr.18: 54362807-54362807 on Build GRCh38
5682437 c.736A>C p.T246P Chr.18: 54362917-54362917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143596631 Chr.18: 54362801-54362801 on Build GRCh38
rs747542067 Chr.18: 54362946-54362947 on Build GRCh38
rs181453274 Chr.18: 54363003-54363003 on Build GRCh38
rs748312524 Chr.18: 54362812-54362812 on Build GRCh38
rs780006933 Chr.18: 54362811-54362811 on Build GRCh38
rs1027707143 Chr.18: 54362936-54362936 on Build GRCh38
rs775839798 Chr.18: 54362850-54362850 on Build GRCh38
rs370834017 Chr.18: 54362553-54362553 on Build GRCh38
rs758757569 Chr.18: 54362814-54362814 on Build GRCh38
rs1041074152 Chr.18: 54362677-54362677 on Build GRCh38
rs975886972 Chr.18: 54363008-54363008 on Build GRCh38
rs146259965 Chr.18: 54362525-54362525 on Build GRCh38
rs565818363 Chr.18: 54362603-54362603 on Build GRCh38
rs953886060 Chr.18: 54363011-54363011 on Build GRCh38
rs6508290 Chr.18: 54362686-54362686 on Build GRCh38
rs557140324 Chr.18: 54362989-54362989 on Build GRCh38
rs567692537 Chr.18: 54362824-54362824 on Build GRCh38
rs537547737 Chr.18: 54362687-54362687 on Build GRCh38
rs904551781 Chr.18: 54362810-54362810 on Build GRCh38
rs553514585 Chr.18: 54362822-54362822 on Build GRCh38
rs746388601 Chr.18: 54362845-54362845 on Build GRCh38
rs906021320 Chr.18: 54362689-54362689 on Build GRCh38
rs538910823 Chr.18: 54362950-54362950 on Build GRCh38
rs1024060533 Chr.18: 54362577-54362577 on Build GRCh38
rs1015003708 Chr.18: 54362591-54362591 on Build GRCh38
rs947558233 Chr.18: 54362685-54362685 on Build GRCh38
rs1000420860 Chr.18: 54362844-54362844 on Build GRCh38
rs73476821 Chr.18: 54362593-54362593 on Build GRCh38
rs895833568 Chr.18: 54362869-54362869 on Build GRCh38
rs765556175 Chr.18: 54362745-54362745 on Build GRCh38
rs770141925 Chr.18: 54362848-54362848 on Build GRCh38
rs778182138 Chr.18: 54362922-54362922 on Build GRCh38
rs751442409 Chr.18: 54362532-54362532 on Build GRCh38
rs954133351 Chr.18: 54362612-54362613 on Build GRCh38
rs371979718 Chr.18: 54362999-54362999 on Build GRCh38
rs868213847 Chr.18: 54362920-54362921 on Build GRCh38
rs74862391 Chr.18: 54362887-54362887 on Build GRCh38
rs760532831 Chr.18: 54362898-54362898 on Build GRCh38
rs750355302 Chr.18: 54362795-54362795 on Build GRCh38
rs1004992981 Chr.18: 54362575-54362575 on Build GRCh38
rs907690576 Chr.18: 54362628-54362628 on Build GRCh38
rs6508289 Chr.18: 54362637-54362637 on Build GRCh38
rs766397263 Chr.18: 54362529-54362529 on Build GRCh38
rs561347486 Chr.18: 54362537-54362537 on Build GRCh38
rs140618781 Chr.18: 54362892-54362892 on Build GRCh38
rs771312594 Chr.18: 54362839-54362839 on Build GRCh38
rs1036132388 Chr.18: 54362543-54362546 on Build GRCh38
rs757117440 Chr.18: 54362788-54362788 on Build GRCh38
rs952043390 Chr.18: 54362623-54362623 on Build GRCh38
rs781148432 Chr.18: 54362956-54362956 on Build GRCh38
rs374155660 Chr.18: 54362937-54362937 on Build GRCh38
rs184805075 Chr.18: 54362633-54362633 on Build GRCh38
rs766270802 Chr.18: 54362905-54362905 on Build GRCh38
rs983347550 Chr.18: 54362627-54362627 on Build GRCh38
rs766954356 Chr.18: 54362873-54362873 on Build GRCh38
rs775078661 Chr.18: 54362746-54362746 on Build GRCh38
rs763530149 Chr.18: 54362766-54362766 on Build GRCh38
rs770355436 Chr.18: 54362973-54362973 on Build GRCh38
rs369589214 Chr.18: 54362860-54362860 on Build GRCh38
rs759985779 Chr.18: 54362726-54362726 on Build GRCh38
rs921811403 Chr.18: 54363014-54363014 on Build GRCh38
rs781695334 Chr.18: 54362840-54362840 on Build GRCh38
rs771616366 Chr.18: 54362952-54362952 on Build GRCh38
rs762592758 Chr.18: 54362858-54362858 on Build GRCh38
rs767526321 Chr.18: 54362791-54362791 on Build GRCh38
rs1015033517 Chr.18: 54362908-54362908 on Build GRCh38
rs746322279 Chr.18: 54362970-54362970 on Build GRCh38
rs528877227 Chr.18: 54362581-54362581 on Build GRCh38
rs895811551 Chr.18: 54362564-54362564 on Build GRCh38
rs778306204 Chr.18: 54362922-54362925 on Build GRCh38
rs985392386 Chr.18: 54362615-54362615 on Build GRCh38
rs773816620 Chr.18: 54362867-54362867 on Build GRCh38
rs1657908 Chr.18: 54362609-54362609 on Build GRCh38
rs144173544 Chr.18: 54362918-54362918 on Build GRCh38
rs932790139 Chr.18: 54362728-54362728 on Build GRCh38
rs762611126 Chr.18: 54362751-54362751 on Build GRCh38
rs555895916 Chr.18: 54362692-54362692 on Build GRCh38
rs189734541 Chr.18: 54363000-54363000 on Build GRCh38
rs748333505 Chr.18: 54362783-54362783 on Build GRCh38
rs1050173289 Chr.18: 54362767-54362767 on Build GRCh38
rs201042542 Chr.18: 54362917-54362917 on Build GRCh38

View Full Product Details