Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54378903-54379418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
UniGene
Hs.208701
Gene Chromosome Location
Chr.18: 54357914-54382035 on Build GRCh38
Transcripts
NM_001288980.1 NM_001288981.1 NM_001288982.1 NM_173529.5 NR_110234.1 XM_005258201.1 XM_005258203.1 XM_017025567.1

SNP Detail

SNP ID Chromosome Location
rs1023087860 Chr.18: 54378986-54378986 on Build GRCh38
rs768463732 Chr.18: 54379352-54379352 on Build GRCh38
rs1013010375 Chr.18: 54378950-54378950 on Build GRCh38
rs1015157233 Chr.18: 54379150-54379151 on Build GRCh38
rs905181651 Chr.18: 54379104-54379104 on Build GRCh38
rs541285013 Chr.18: 54379299-54379299 on Build GRCh38
rs912072282 Chr.18: 54379000-54379000 on Build GRCh38
rs915032480 Chr.18: 54379142-54379142 on Build GRCh38
rs1003777094 Chr.18: 54378933-54378933 on Build GRCh38
rs537867451 Chr.18: 54378939-54378939 on Build GRCh38
rs946989752 Chr.18: 54379345-54379345 on Build GRCh38
rs1045060239 Chr.18: 54379091-54379091 on Build GRCh38
rs1044478895 Chr.18: 54379334-54379334 on Build GRCh38
rs560710837 Chr.18: 54379158-54379158 on Build GRCh38
rs368729856 Chr.18: 54379304-54379304 on Build GRCh38
rs894082016 Chr.18: 54379068-54379068 on Build GRCh38
rs931737324 Chr.18: 54378965-54378965 on Build GRCh38
rs373684724 Chr.18: 54379335-54379335 on Build GRCh38
rs1035099330 Chr.18: 54379127-54379127 on Build GRCh38
rs1026332080 Chr.18: 54379363-54379363 on Build GRCh38
rs552242531 Chr.18: 54379054-54379054 on Build GRCh38
rs572070602 Chr.18: 54379251-54379251 on Build GRCh38
rs935548255 Chr.18: 54379025-54379025 on Build GRCh38
rs935071828 Chr.18: 54379289-54379289 on Build GRCh38
rs987121479 Chr.18: 54379285-54379285 on Build GRCh38
rs78552974 Chr.18: 54379143-54379143 on Build GRCh38
rs912066204 Chr.18: 54379060-54379060 on Build GRCh38
rs899275511 Chr.18: 54379350-54379350 on Build GRCh38
rs951009077 Chr.18: 54379014-54379014 on Build GRCh38
rs542540468 Chr.18: 54379149-54379149 on Build GRCh38
rs556480657 Chr.18: 54379034-54379034 on Build GRCh38
rs74880026 Chr.18: 54379144-54379144 on Build GRCh38
rs1026552689 Chr.18: 54378995-54378995 on Build GRCh38
rs202041250 Chr.18: 54379076-54379077 on Build GRCh38
rs535417748 Chr.18: 54379168-54379168 on Build GRCh38
rs116624237 Chr.18: 54379199-54379199 on Build GRCh38
rs1012124322 Chr.18: 54379310-54379310 on Build GRCh38
rs895867720 Chr.18: 54378940-54378940 on Build GRCh38
rs994861031 Chr.18: 54378994-54378994 on Build GRCh38
rs558088927 Chr.18: 54378999-54378999 on Build GRCh38
rs973689573 Chr.18: 54379195-54379195 on Build GRCh38
rs963335966 Chr.18: 54379184-54379184 on Build GRCh38
rs1011563135 Chr.18: 54379069-54379069 on Build GRCh38
rs911667084 Chr.18: 54379288-54379290 on Build GRCh38
rs1053032170 Chr.18: 54379026-54379026 on Build GRCh38
rs540348844 Chr.18: 54379248-54379248 on Build GRCh38
rs964781131 Chr.18: 54379102-54379102 on Build GRCh38
rs974458292 Chr.18: 54379138-54379138 on Build GRCh38
rs923827323 Chr.18: 54379193-54379193 on Build GRCh38
rs1003588696 Chr.18: 54379119-54379119 on Build GRCh38
rs963368235 Chr.18: 54378988-54378988 on Build GRCh38
rs1056662746 Chr.18: 54379245-54379245 on Build GRCh38
rs921568356 Chr.18: 54378946-54378946 on Build GRCh38

View Full Product Details