Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7696933-7697427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER2
Entrez Gene ID
2208
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
UniGene
Hs.465778
Gene Chromosome Location
Chr.19: 7688757-7702755 on Build GRCh38
Transcripts
NM_001207019.2 NM_001220500.1 NM_002002.4 XM_005272462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960701 c.355G>T p.D119Y Chr.19: 7697037-7697037 on Build GRCh38
4887193 c.286C>G p.R96G Chr.19: 7697266-7697266 on Build GRCh38
713890 c.258G>T p.T86T Chr.19: 7697294-7697294 on Build GRCh38
7167990 c.297G>A p.Q99Q Chr.19: 7697255-7697255 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767687089 Chr.19: 7697265-7697265 on Build GRCh38
rs926107334 Chr.19: 7697360-7697360 on Build GRCh38
rs756249131 Chr.19: 7697269-7697269 on Build GRCh38
rs771565535 Chr.19: 7697205-7697205 on Build GRCh38
rs567385173 Chr.19: 7697288-7697288 on Build GRCh38
rs780566367 Chr.19: 7697323-7697323 on Build GRCh38
rs549995522 Chr.19: 7696965-7696965 on Build GRCh38
rs747034906 Chr.19: 7696986-7696986 on Build GRCh38
rs771296088 Chr.19: 7697034-7697034 on Build GRCh38
rs777793578 Chr.19: 7697281-7697281 on Build GRCh38
rs931587885 Chr.19: 7697086-7697086 on Build GRCh38
rs746460429 Chr.19: 7697302-7697302 on Build GRCh38
rs765140667 Chr.19: 7697229-7697229 on Build GRCh38
rs141812261 Chr.19: 7696962-7696962 on Build GRCh38
rs752349516 Chr.19: 7696960-7696960 on Build GRCh38
rs761194466 Chr.19: 7697016-7697016 on Build GRCh38
rs555886538 Chr.19: 7697394-7697394 on Build GRCh38
rs74980343 Chr.19: 7697133-7697133 on Build GRCh38
rs748353538 Chr.19: 7697108-7697108 on Build GRCh38
rs780155670 Chr.19: 7696985-7696985 on Build GRCh38
rs1057220341 Chr.19: 7697154-7697154 on Build GRCh38
rs887293379 Chr.19: 7697134-7697134 on Build GRCh38
rs763027862 Chr.19: 7697261-7697261 on Build GRCh38
rs757094618 Chr.19: 7697082-7697082 on Build GRCh38
rs755136991 Chr.19: 7697006-7697006 on Build GRCh38
rs1003427637 Chr.19: 7697148-7697148 on Build GRCh38
rs373387153 Chr.19: 7697056-7697056 on Build GRCh38
rs774956412 Chr.19: 7697038-7697038 on Build GRCh38
rs750401656 Chr.19: 7697092-7697092 on Build GRCh38
rs752775281 Chr.19: 7697266-7697266 on Build GRCh38
rs759071522 Chr.19: 7696958-7696958 on Build GRCh38
rs377594003 Chr.19: 7696969-7696969 on Build GRCh38
rs776374918 Chr.19: 7697220-7697220 on Build GRCh38
rs751548674 Chr.19: 7697329-7697329 on Build GRCh38
rs779315614 Chr.19: 7697290-7697290 on Build GRCh38
rs746343109 Chr.19: 7697294-7697294 on Build GRCh38
rs750026899 Chr.19: 7697112-7697112 on Build GRCh38
rs765042293 Chr.19: 7697089-7697089 on Build GRCh38
rs371099977 Chr.19: 7697123-7697123 on Build GRCh38
rs767026479 Chr.19: 7697049-7697049 on Build GRCh38
rs140629973 Chr.19: 7697050-7697050 on Build GRCh38
rs528355833 Chr.19: 7697173-7697173 on Build GRCh38
rs773492796 Chr.19: 7697029-7697029 on Build GRCh38
rs767118350 Chr.19: 7696959-7696959 on Build GRCh38
rs761596541 Chr.19: 7697313-7697313 on Build GRCh38
rs987252022 Chr.19: 7697005-7697005 on Build GRCh38
rs894515778 Chr.19: 7697172-7697172 on Build GRCh38
rs778079414 Chr.19: 7697125-7697125 on Build GRCh38
rs183284748 Chr.19: 7696984-7696984 on Build GRCh38
rs773195059 Chr.19: 7697316-7697316 on Build GRCh38
rs760616294 Chr.19: 7697335-7697335 on Build GRCh38
rs200169018 Chr.19: 7697240-7697240 on Build GRCh38
rs377637515 Chr.19: 7697078-7697078 on Build GRCh38
rs749780974 Chr.19: 7697033-7697033 on Build GRCh38
rs747631191 Chr.19: 7697383-7697383 on Build GRCh38
rs1028077878 Chr.19: 7697159-7697159 on Build GRCh38
rs772612971 Chr.19: 7697295-7697295 on Build GRCh38
rs1031564800 Chr.19: 7697393-7697393 on Build GRCh38
rs936188137 Chr.19: 7697370-7697370 on Build GRCh38
rs186188865 Chr.19: 7697404-7697404 on Build GRCh38
rs769684627 Chr.19: 7697315-7697315 on Build GRCh38
rs774780248 Chr.19: 7697127-7697127 on Build GRCh38
rs780558442 Chr.19: 7697300-7697300 on Build GRCh38
rs768213654 Chr.19: 7697308-7697308 on Build GRCh38
rs572304844 Chr.19: 7697397-7697397 on Build GRCh38
rs571262008 Chr.19: 7697216-7697216 on Build GRCh38
rs754001038 Chr.19: 7697344-7697344 on Build GRCh38
rs370372077 Chr.19: 7697041-7697041 on Build GRCh38
rs753622173 Chr.19: 7697077-7697077 on Build GRCh38
rs961384969 Chr.19: 7697247-7697247 on Build GRCh38
rs535989649 Chr.19: 7697311-7697311 on Build GRCh38
rs1005420586 Chr.19: 7697210-7697210 on Build GRCh38
rs767264355 Chr.19: 7697083-7697084 on Build GRCh38
rs763914443 Chr.19: 7697074-7697074 on Build GRCh38
rs542037732 Chr.19: 7697257-7697257 on Build GRCh38
rs565751215 Chr.19: 7697126-7697126 on Build GRCh38
rs117434896 Chr.19: 7697219-7697219 on Build GRCh38
rs753809296 Chr.19: 7696974-7696974 on Build GRCh38
rs1053404666 Chr.19: 7697007-7697007 on Build GRCh38
rs952547310 Chr.19: 7697203-7697203 on Build GRCh38
rs764291342 Chr.19: 7697340-7697340 on Build GRCh38
rs113240986 Chr.19: 7697407-7697407 on Build GRCh38
rs867370250 Chr.19: 7697055-7697055 on Build GRCh38
rs866167964 Chr.19: 7697236-7697236 on Build GRCh38
rs781509818 Chr.19: 7697012-7697012 on Build GRCh38
rs551705532 Chr.19: 7697195-7697195 on Build GRCh38
rs754102526 Chr.19: 7697282-7697282 on Build GRCh38
rs766379194 Chr.19: 7697325-7697325 on Build GRCh38
rs369524074 Chr.19: 7697018-7697018 on Build GRCh38
rs998801664 Chr.19: 7697336-7697336 on Build GRCh38
rs376024411 Chr.19: 7697226-7697226 on Build GRCh38
rs900894537 Chr.19: 7697065-7697065 on Build GRCh38
rs781493170 Chr.19: 7697093-7697093 on Build GRCh38

View Full Product Details