Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35498794-35499296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMKN
Entrez Gene ID
93099
Gene Name
dermokine
Gene Aliases
UNQ729, ZD52F10
UniGene
Hs.417795
Gene Chromosome Location
Chr.19: 35497217-35513678 on Build GRCh38
Transcripts
NM_001035516.3 NM_001126056.2 NM_001126059.2 NM_001190347.1 NM_001308380.1 NM_001308383.1 NM_033317.4 NR_033746.1 XM_006723477.1 XM_006723484.1 XM_006723489.1 XM_006723493.2 XM_006723494.2 XM_006723503.2 XM_011527494.2 XM_011527495.2 XM_011527496.2 XM_011527497.2 XM_011527498.2 XM_011527499.2 XM_011527500.2 XM_011527501.2 XM_011527502.2 XM_011527503.2 XM_011527504.2 XM_011527505.2 XM_011527506.2 XM_011527507.2 XM_011527508.2 XM_011527509.2 XM_011527510.2 XM_011527513.1 XM_011527514.1 XM_017027475.1 XM_017027476.1 XM_017027477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3197845 c.206delG p.G69fs*19 Chr.19: 35498893-35498893 on Build GRCh38
4336511 c.1283delG p.G428fs*19 Chr.19: 35498893-35498893 on Build GRCh38
7240827 c.1360-10C>A p.? Chr.19: 35498907-35498907 on Build GRCh38
7240832 c.213-10C>A p.? Chr.19: 35498907-35498907 on Build GRCh38
4842725 c.1370C>T p.S457L Chr.19: 35498887-35498887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778215902 Chr.19: 35498854-35498854 on Build GRCh38
rs748931721 Chr.19: 35498840-35498840 on Build GRCh38
rs368794417 Chr.19: 35498932-35498932 on Build GRCh38
rs12975867 Chr.19: 35499264-35499264 on Build GRCh38
rs900394351 Chr.19: 35499020-35499020 on Build GRCh38
rs149935932 Chr.19: 35499179-35499179 on Build GRCh38
rs764267042 Chr.19: 35498896-35498896 on Build GRCh38
rs749735815 Chr.19: 35498861-35498861 on Build GRCh38
rs140257981 Chr.19: 35499042-35499042 on Build GRCh38
rs568720181 Chr.19: 35498977-35498977 on Build GRCh38
rs184563318 Chr.19: 35498898-35498898 on Build GRCh38
rs1048167617 Chr.19: 35499013-35499013 on Build GRCh38
rs142000561 Chr.19: 35498897-35498897 on Build GRCh38
rs2293693 Chr.19: 35499070-35499070 on Build GRCh38
rs997424261 Chr.19: 35499023-35499023 on Build GRCh38
rs978520054 Chr.19: 35499234-35499234 on Build GRCh38
rs746266009 Chr.19: 35498815-35498815 on Build GRCh38
rs753544971 Chr.19: 35498935-35498935 on Build GRCh38
rs754635250 Chr.19: 35498937-35498937 on Build GRCh38
rs777029827 Chr.19: 35498947-35498947 on Build GRCh38
rs537352785 Chr.19: 35499133-35499133 on Build GRCh38
rs923483447 Chr.19: 35498979-35498979 on Build GRCh38
rs780222714 Chr.19: 35498816-35498816 on Build GRCh38
rs942119957 Chr.19: 35499053-35499053 on Build GRCh38
rs778506275 Chr.19: 35498944-35498944 on Build GRCh38
rs369467672 Chr.19: 35498875-35498875 on Build GRCh38
rs1029697201 Chr.19: 35499054-35499054 on Build GRCh38
rs202112325 Chr.19: 35498923-35498923 on Build GRCh38
rs774726109 Chr.19: 35499068-35499068 on Build GRCh38
rs552878413 Chr.19: 35499255-35499255 on Build GRCh38
rs968342043 Chr.19: 35499217-35499217 on Build GRCh38
rs756515969 Chr.19: 35498830-35498830 on Build GRCh38
rs895695041 Chr.19: 35499140-35499140 on Build GRCh38
rs779918983 Chr.19: 35498832-35498832 on Build GRCh38
rs1009923804 Chr.19: 35499161-35499161 on Build GRCh38
rs775702761 Chr.19: 35498876-35498876 on Build GRCh38
rs1006434408 Chr.19: 35498978-35498978 on Build GRCh38
rs887616566 Chr.19: 35498952-35498952 on Build GRCh38
rs772620481 Chr.19: 35499174-35499174 on Build GRCh38
rs916113019 Chr.19: 35498822-35498822 on Build GRCh38
rs111882428 Chr.19: 35499139-35499139 on Build GRCh38
rs768290760 Chr.19: 35498848-35498848 on Build GRCh38
rs545562159 Chr.19: 35498836-35498836 on Build GRCh38
rs771954574 Chr.19: 35498862-35498862 on Build GRCh38
rs760534903 Chr.19: 35498874-35498874 on Build GRCh38
rs1038965074 Chr.19: 35499001-35499001 on Build GRCh38
rs550847808 Chr.19: 35499138-35499138 on Build GRCh38
rs763096533 Chr.19: 35498883-35498883 on Build GRCh38
rs948425059 Chr.19: 35498866-35498866 on Build GRCh38
rs764025438 Chr.19: 35498893-35498893 on Build GRCh38
rs773160478 Chr.19: 35498878-35498880 on Build GRCh38
rs758253239 Chr.19: 35498892-35498893 on Build GRCh38
rs756604194 Chr.19: 35498927-35498927 on Build GRCh38
rs773173663 Chr.19: 35498870-35498870 on Build GRCh38
rs1047469017 Chr.19: 35498926-35498926 on Build GRCh38
rs990692273 Chr.19: 35499261-35499261 on Build GRCh38
rs1019932063 Chr.19: 35499195-35499195 on Build GRCh38
rs998978430 Chr.19: 35499125-35499125 on Build GRCh38
rs905736901 Chr.19: 35499069-35499069 on Build GRCh38
rs541540517 Chr.19: 35499048-35499048 on Build GRCh38

View Full Product Details