Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35550006-35550547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4A
Entrez Gene ID
495
Gene Name
ATPase H+/K+ transporting alpha subunit
Gene Aliases
ATP6A
UniGene
Hs.36992
Gene Chromosome Location
Chr.19: 35550193-35563658 on Build GRCh38
Transcripts
NM_000704.2

SNP Detail

SNP ID Chromosome Location
rs1042076701 Chr.19: 35550472-35550472 on Build GRCh38
rs1013814293 Chr.19: 35550285-35550285 on Build GRCh38
rs115176252 Chr.19: 35550044-35550044 on Build GRCh38
rs896876682 Chr.19: 35550064-35550064 on Build GRCh38
rs943566068 Chr.19: 35550428-35550428 on Build GRCh38
rs186189094 Chr.19: 35550046-35550046 on Build GRCh38
rs1000567200 Chr.19: 35550255-35550255 on Build GRCh38
rs917266025 Chr.19: 35550069-35550069 on Build GRCh38
rs556378541 Chr.19: 35550141-35550141 on Build GRCh38
rs959221957 Chr.19: 35550275-35550275 on Build GRCh38
rs145988745 Chr.19: 35550477-35550477 on Build GRCh38
rs373981267 Chr.19: 35550034-35550034 on Build GRCh38
rs902067287 Chr.19: 35550224-35550224 on Build GRCh38
rs972093288 Chr.19: 35550367-35550367 on Build GRCh38
rs752742347 Chr.19: 35550259-35550259 on Build GRCh38
rs984880389 Chr.19: 35550404-35550404 on Build GRCh38
rs889273962 Chr.19: 35550179-35550179 on Build GRCh38
rs546039558 Chr.19: 35550509-35550509 on Build GRCh38
rs952134288 Chr.19: 35550394-35550394 on Build GRCh38
rs529732027 Chr.19: 35550481-35550481 on Build GRCh38
rs2733744 Chr.19: 35550128-35550128 on Build GRCh38
rs896903856 Chr.19: 35550376-35550376 on Build GRCh38
rs188942403 Chr.19: 35550168-35550168 on Build GRCh38
rs952331622 Chr.19: 35550131-35550131 on Build GRCh38
rs972019698 Chr.19: 35550104-35550104 on Build GRCh38
rs571786704 Chr.19: 35550183-35550183 on Build GRCh38
rs1023185453 Chr.19: 35550451-35550451 on Build GRCh38
rs555207086 Chr.19: 35550174-35550174 on Build GRCh38
rs563754707 Chr.19: 35550300-35550300 on Build GRCh38
rs142870685 Chr.19: 35550245-35550245 on Build GRCh38
rs1040902887 Chr.19: 35550441-35550441 on Build GRCh38
rs943719976 Chr.19: 35550352-35550352 on Build GRCh38
rs546183128 Chr.19: 35550311-35550311 on Build GRCh38
rs570828745 Chr.19: 35550089-35550089 on Build GRCh38
rs1022943563 Chr.19: 35550177-35550177 on Build GRCh38
rs576206234 Chr.19: 35550148-35550148 on Build GRCh38
rs981438810 Chr.19: 35550207-35550207 on Build GRCh38
rs1009188970 Chr.19: 35550229-35550229 on Build GRCh38
rs992069791 Chr.19: 35550057-35550057 on Build GRCh38
rs113572161 Chr.19: 35550322-35550322 on Build GRCh38
rs1040641012 Chr.19: 35550184-35550184 on Build GRCh38
rs1020540278 Chr.19: 35550230-35550230 on Build GRCh38
rs531849322 Chr.19: 35550444-35550444 on Build GRCh38
rs563192164 Chr.19: 35550423-35550423 on Build GRCh38
rs868706006 Chr.19: 35550353-35550353 on Build GRCh38
rs919259055 Chr.19: 35550388-35550388 on Build GRCh38
rs751402450 Chr.19: 35550462-35550462 on Build GRCh38
rs577177498 Chr.19: 35550310-35550310 on Build GRCh38
rs995256573 Chr.19: 35550377-35550377 on Build GRCh38
rs961424082 Chr.19: 35550356-35550356 on Build GRCh38
rs1025221219 Chr.19: 35550515-35550515 on Build GRCh38
rs894848375 Chr.19: 35550484-35550484 on Build GRCh38
rs945621704 Chr.19: 35550468-35550468 on Build GRCh38
rs181550982 Chr.19: 35550478-35550478 on Build GRCh38

View Full Product Details