Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35550743-35551239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4A
Entrez Gene ID
495
Gene Name
ATPase H+/K+ transporting alpha subunit
Gene Aliases
ATP6A
UniGene
Hs.36992
Gene Chromosome Location
Chr.19: 35550193-35563658 on Build GRCh38
Transcripts
NM_000704.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4615877 c.2910_2911insATC p.I970_V971insI Chr.19: 35551086-35551087 on Build GRCh38
6441025 c.2972A>G p.N991S Chr.19: 35551025-35551025 on Build GRCh38
317032 c.2951C>A p.P984H Chr.19: 35551046-35551046 on Build GRCh38
1718120 c.3050G>A p.R1017Q Chr.19: 35550863-35550863 on Build GRCh38
3692655 c.2952C>T p.P984P Chr.19: 35551045-35551045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746718673 Chr.19: 35551007-35551007 on Build GRCh38
rs1042605251 Chr.19: 35550769-35550769 on Build GRCh38
rs776415670 Chr.19: 35551159-35551159 on Build GRCh38
rs756036260 Chr.19: 35551042-35551042 on Build GRCh38
rs146641851 Chr.19: 35550894-35550894 on Build GRCh38
rs763693283 Chr.19: 35550896-35550896 on Build GRCh38
rs760879648 Chr.19: 35550849-35550849 on Build GRCh38
rs964860973 Chr.19: 35550959-35550959 on Build GRCh38
rs775351403 Chr.19: 35550808-35550808 on Build GRCh38
rs141390472 Chr.19: 35550895-35550895 on Build GRCh38
rs148503854 Chr.19: 35550879-35550879 on Build GRCh38
rs912387737 Chr.19: 35551006-35551006 on Build GRCh38
rs555560469 Chr.19: 35550772-35550772 on Build GRCh38
rs182728369 Chr.19: 35550778-35550778 on Build GRCh38
rs768537649 Chr.19: 35550966-35550966 on Build GRCh38
rs769077364 Chr.19: 35551133-35551133 on Build GRCh38
rs754876106 Chr.19: 35551063-35551063 on Build GRCh38
rs990540383 Chr.19: 35551071-35551071 on Build GRCh38
rs185924462 Chr.19: 35550856-35550856 on Build GRCh38
rs775752483 Chr.19: 35551009-35551009 on Build GRCh38
rs761452162 Chr.19: 35551020-35551020 on Build GRCh38
rs771198959 Chr.19: 35550979-35550979 on Build GRCh38
rs1041135235 Chr.19: 35551114-35551134 on Build GRCh38
rs780666796 Chr.19: 35550920-35550920 on Build GRCh38
rs781158416 Chr.19: 35551089-35551089 on Build GRCh38
rs779735781 Chr.19: 35550952-35550952 on Build GRCh38
rs756021473 Chr.19: 35550937-35550937 on Build GRCh38
rs368013648 Chr.19: 35551118-35551118 on Build GRCh38
rs1030486067 Chr.19: 35550973-35550973 on Build GRCh38
rs372231915 Chr.19: 35550936-35550936 on Build GRCh38
rs1046288378 Chr.19: 35550864-35550864 on Build GRCh38
rs1026923996 Chr.19: 35550886-35550886 on Build GRCh38
rs527310579 Chr.19: 35551048-35551048 on Build GRCh38
rs199522591 Chr.19: 35551069-35551069 on Build GRCh38
rs749899247 Chr.19: 35551041-35551041 on Build GRCh38
rs896885665 Chr.19: 35551149-35551149 on Build GRCh38
rs767019999 Chr.19: 35551030-35551030 on Build GRCh38
rs777066391 Chr.19: 35550875-35550875 on Build GRCh38
rs867597711 Chr.19: 35551002-35551002 on Build GRCh38
rs773459189 Chr.19: 35550848-35550848 on Build GRCh38
rs770290962 Chr.19: 35550998-35550998 on Build GRCh38
rs769534360 Chr.19: 35550803-35550803 on Build GRCh38
rs28581880 Chr.19: 35551035-35551035 on Build GRCh38
rs772372492 Chr.19: 35550831-35550831 on Build GRCh38
rs770523770 Chr.19: 35551063-35551064 on Build GRCh38
rs201163746 Chr.19: 35550785-35550785 on Build GRCh38
rs1046194969 Chr.19: 35551136-35551136 on Build GRCh38
rs751179419 Chr.19: 35550904-35550904 on Build GRCh38
rs751271471 Chr.19: 35551014-35551014 on Build GRCh38
rs41513445 Chr.19: 35551174-35551174 on Build GRCh38
rs774631292 Chr.19: 35551147-35551147 on Build GRCh38
rs201486149 Chr.19: 35550791-35550791 on Build GRCh38
rs370121207 Chr.19: 35550944-35550944 on Build GRCh38
rs374851312 Chr.19: 35551044-35551044 on Build GRCh38
rs778844114 Chr.19: 35551068-35551068 on Build GRCh38
rs768722481 Chr.19: 35550810-35550810 on Build GRCh38
rs144537248 Chr.19: 35550883-35550883 on Build GRCh38
rs748332451 Chr.19: 35550814-35550814 on Build GRCh38
rs201059478 Chr.19: 35551155-35551155 on Build GRCh38
rs200601286 Chr.19: 35551010-35551010 on Build GRCh38
rs375269377 Chr.19: 35550993-35550993 on Build GRCh38
rs376927656 Chr.19: 35551189-35551189 on Build GRCh38
rs377741206 Chr.19: 35551120-35551120 on Build GRCh38
rs866954095 Chr.19: 35550971-35550971 on Build GRCh38
rs756807518 Chr.19: 35550908-35550908 on Build GRCh38
rs562280607 Chr.19: 35551191-35551191 on Build GRCh38
rs374197390 Chr.19: 35551093-35551093 on Build GRCh38
rs896980014 Chr.19: 35550880-35550880 on Build GRCh38
rs767158003 Chr.19: 35551160-35551160 on Build GRCh38
rs559698579 Chr.19: 35550906-35550906 on Build GRCh38
rs780077958 Chr.19: 35551045-35551045 on Build GRCh38
rs149744668 Chr.19: 35551057-35551057 on Build GRCh38
rs865870936 Chr.19: 35551043-35551043 on Build GRCh38
rs200663366 Chr.19: 35551086-35551086 on Build GRCh38
rs749069418 Chr.19: 35550964-35550964 on Build GRCh38
rs759978664 Chr.19: 35550995-35550995 on Build GRCh38
rs769733660 Chr.19: 35551099-35551099 on Build GRCh38
rs945655761 Chr.19: 35550767-35550767 on Build GRCh38
rs372203615 Chr.19: 35550972-35550972 on Build GRCh38
rs745794777 Chr.19: 35550792-35550792 on Build GRCh38
rs778188004 Chr.19: 35550968-35550968 on Build GRCh38
rs755477472 Chr.19: 35550779-35550779 on Build GRCh38
rs867856832 Chr.19: 35550782-35550782 on Build GRCh38
rs764421480 Chr.19: 35551011-35551011 on Build GRCh38

View Full Product Details