Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111144399-111144899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L11
Entrez Gene ID
10018
Gene Name
BCL2 like 11
Gene Aliases
BAM, BIM, BOD
UniGene
Hs.469658
Gene Chromosome Location
Chr.2: 111120914-111168445 on Build GRCh38
Transcripts
NM_001204106.1 NM_001204107.1 NM_001204108.1 NM_001204109.1 NM_001204110.1 NM_001204111.1 NM_001204112.1 NM_001204113.1 NM_006538.4 NM_138621.4 NM_138622.3 NM_138623.3 NM_138624.3 NM_138625.3 NM_138626.3 NM_138627.3 NM_207003.2 XM_005263550.3 XM_005263551.2 XM_005263552.3 XM_005263553.2 XM_005263554.2 XM_005263555.3 XM_005263556.3 XM_005263557.4 XM_005263559.2 XM_005263561.3 XM_017003101.1 XM_017003102.1

SNP Detail

SNP ID Chromosome Location
rs932619417 Chr.2: 111144794-111144794 on Build GRCh38
rs185182851 Chr.2: 111144764-111144764 on Build GRCh38
rs969687192 Chr.2: 111144588-111144589 on Build GRCh38
rs983413905 Chr.2: 111144680-111144680 on Build GRCh38
rs919714207 Chr.2: 111144426-111144426 on Build GRCh38
rs1021396519 Chr.2: 111144560-111144560 on Build GRCh38
rs145826988 Chr.2: 111144747-111144747 on Build GRCh38
rs571983259 Chr.2: 111144645-111144645 on Build GRCh38
rs983053566 Chr.2: 111144449-111144449 on Build GRCh38
rs979774619 Chr.2: 111144595-111144595 on Build GRCh38
rs963683206 Chr.2: 111144738-111144738 on Build GRCh38
rs890864120 Chr.2: 111144671-111144671 on Build GRCh38
rs770120722 Chr.2: 111144443-111144443 on Build GRCh38
rs577360498 Chr.2: 111144470-111144470 on Build GRCh38
rs563381218 Chr.2: 111144862-111144862 on Build GRCh38
rs916546229 Chr.2: 111144847-111144847 on Build GRCh38
rs776964241 Chr.2: 111144472-111144472 on Build GRCh38
rs553647984 Chr.2: 111144870-111144870 on Build GRCh38
rs939387181 Chr.2: 111144800-111144800 on Build GRCh38
rs891379160 Chr.2: 111144465-111144465 on Build GRCh38
rs992252193 Chr.2: 111144806-111144806 on Build GRCh38
rs772422702 Chr.2: 111144440-111144440 on Build GRCh38
rs73954956 Chr.2: 111144517-111144517 on Build GRCh38
rs960229033 Chr.2: 111144667-111144667 on Build GRCh38
rs536932265 Chr.2: 111144737-111144737 on Build GRCh38
rs56866966 Chr.2: 111144720-111144720 on Build GRCh38
rs117840429 Chr.2: 111144773-111144773 on Build GRCh38
rs1031618250 Chr.2: 111144660-111144660 on Build GRCh38
rs929821114 Chr.2: 111144447-111144447 on Build GRCh38
rs1032254165 Chr.2: 111144483-111144483 on Build GRCh38
rs114702002 Chr.2: 111144547-111144547 on Build GRCh38
rs760927858 Chr.2: 111144455-111144455 on Build GRCh38
rs116081708 Chr.2: 111144450-111144450 on Build GRCh38
rs982617935 Chr.2: 111144768-111144768 on Build GRCh38
rs891166091 Chr.2: 111144801-111144801 on Build GRCh38
rs973747343 Chr.2: 111144743-111144743 on Build GRCh38
rs567928248 Chr.2: 111144701-111144701 on Build GRCh38
rs115636317 Chr.2: 111144464-111144464 on Build GRCh38
rs764226375 Chr.2: 111144609-111144609 on Build GRCh38
rs920012794 Chr.2: 111144785-111144785 on Build GRCh38
rs904141777 Chr.2: 111144568-111144568 on Build GRCh38
rs536251895 Chr.2: 111144637-111144637 on Build GRCh38

View Full Product Details