Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 79904751-79905295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANTXR2
Entrez Gene ID
118429
Gene Name
anthrax toxin receptor 2
Gene Aliases
CMG-2, CMG2, HFS, ISH, JHF
UniGene
Hs.162963
Gene Chromosome Location
Chr.4: 79901617-80073472 on Build GRCh38
Transcripts
NM_001286780.1 NM_001286781.1 NM_058172.5

SNP Detail

SNP ID Chromosome Location
rs1044385855 Chr.4: 79904978-79904978 on Build GRCh38
rs1045206751 Chr.4: 79905113-79905113 on Build GRCh38
rs978394899 Chr.4: 79905145-79905145 on Build GRCh38
rs982091703 Chr.4: 79904804-79904804 on Build GRCh38
rs116794675 Chr.4: 79905184-79905184 on Build GRCh38
rs974417580 Chr.4: 79904883-79904884 on Build GRCh38
rs1018645431 Chr.4: 79905066-79905066 on Build GRCh38
rs895943930 Chr.4: 79904965-79904965 on Build GRCh38
rs962773328 Chr.4: 79904918-79904918 on Build GRCh38
rs951321206 Chr.4: 79905137-79905137 on Build GRCh38
rs115504770 Chr.4: 79905126-79905126 on Build GRCh38
rs1010662099 Chr.4: 79905039-79905039 on Build GRCh38
rs1035330882 Chr.4: 79904840-79904840 on Build GRCh38
rs933982024 Chr.4: 79904949-79904949 on Build GRCh38
rs982592332 Chr.4: 79904877-79904877 on Build GRCh38
rs377764782 Chr.4: 79905175-79905175 on Build GRCh38
rs975871505 Chr.4: 79904928-79904928 on Build GRCh38
rs1009103605 Chr.4: 79905064-79905064 on Build GRCh38
rs557461414 Chr.4: 79905185-79905185 on Build GRCh38
rs563152250 Chr.4: 79905221-79905221 on Build GRCh38
rs1044519123 Chr.4: 79905242-79905242 on Build GRCh38
rs967757310 Chr.4: 79905143-79905143 on Build GRCh38
rs1013008817 Chr.4: 79904968-79904968 on Build GRCh38
rs186733755 Chr.4: 79904983-79904983 on Build GRCh38
rs764291190 Chr.4: 79904990-79904990 on Build GRCh38
rs995476496 Chr.4: 79905000-79905000 on Build GRCh38
rs964388062 Chr.4: 79905082-79905082 on Build GRCh38
rs551925465 Chr.4: 79904991-79904992 on Build GRCh38
rs1042319474 Chr.4: 79904907-79904907 on Build GRCh38
rs865837648 Chr.4: 79904845-79904845 on Build GRCh38
rs768190397 Chr.4: 79904843-79904843 on Build GRCh38
rs939221710 Chr.4: 79904942-79904942 on Build GRCh38
rs987611494 Chr.4: 79904859-79904859 on Build GRCh38
rs899144164 Chr.4: 79905250-79905250 on Build GRCh38
rs904881756 Chr.4: 79905115-79905115 on Build GRCh38
rs924841599 Chr.4: 79904966-79904966 on Build GRCh38
rs1054647322 Chr.4: 79905006-79905006 on Build GRCh38
rs559781709 Chr.4: 79904879-79904879 on Build GRCh38
rs921310557 Chr.4: 79904938-79904938 on Build GRCh38
rs1026592501 Chr.4: 79905003-79905003 on Build GRCh38
rs533517816 Chr.4: 79905203-79905203 on Build GRCh38
rs902319267 Chr.4: 79904925-79904925 on Build GRCh38
rs950933867 Chr.4: 79904806-79904806 on Build GRCh38
rs1003608735 Chr.4: 79905123-79905123 on Build GRCh38
rs543254931 Chr.4: 79905204-79905204 on Build GRCh38
rs773364823 Chr.4: 79905068-79905068 on Build GRCh38
rs914909830 Chr.4: 79904884-79904884 on Build GRCh38
rs939180012 Chr.4: 79905189-79905189 on Build GRCh38
rs1051372319 Chr.4: 79904960-79904960 on Build GRCh38
rs946589406 Chr.4: 79904889-79904889 on Build GRCh38
rs893269822 Chr.4: 79905025-79905025 on Build GRCh38

View Full Product Details