Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5527412-5527935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTB
Entrez Gene ID
60
Gene Name
actin beta
Gene Aliases
BRWS1, PS1TP5BP1
UniGene
Hs.520640
Gene Chromosome Location
Chr.7: 5527148-5530601 on Build GRCh38
Transcripts
NM_001101.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205060 c.1123T>G p.F375V Chr.7: 5527753-5527753 on Build GRCh38
453255 c.1004G>C p.R335P Chr.7: 5527872-5527872 on Build GRCh38
4520641 c.1068G>C p.W356C Chr.7: 5527808-5527808 on Build GRCh38
747412 c.1062G>C p.Q354H Chr.7: 5527814-5527814 on Build GRCh38
6592853 c.1009T>C p.Y337H Chr.7: 5527867-5527867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772626920 Chr.7: 5527894-5527896 on Build GRCh38
rs372205322 Chr.7: 5527652-5527653 on Build GRCh38
rs528042634 Chr.7: 5527723-5527723 on Build GRCh38
rs751647886 Chr.7: 5527699-5527699 on Build GRCh38
rs761955678 Chr.7: 5527702-5527702 on Build GRCh38
rs11546911 Chr.7: 5527547-5527547 on Build GRCh38
rs577490709 Chr.7: 5527632-5527632 on Build GRCh38
rs765811140 Chr.7: 5527904-5527904 on Build GRCh38
rs774719514 Chr.7: 5527747-5527748 on Build GRCh38
rs887488041 Chr.7: 5527592-5527592 on Build GRCh38
rs779589316 Chr.7: 5527799-5527799 on Build GRCh38
rs920724265 Chr.7: 5527624-5527626 on Build GRCh38
rs917362860 Chr.7: 5527655-5527655 on Build GRCh38
rs776416632 Chr.7: 5527736-5527736 on Build GRCh38
rs766708549 Chr.7: 5527783-5527783 on Build GRCh38
rs200011987 Chr.7: 5527746-5527746 on Build GRCh38
rs13447411 Chr.7: 5527538-5527538 on Build GRCh38
rs546596340 Chr.7: 5527730-5527730 on Build GRCh38
rs11546927 Chr.7: 5527477-5527477 on Build GRCh38
rs11546928 Chr.7: 5527639-5527639 on Build GRCh38
rs774373985 Chr.7: 5527751-5527751 on Build GRCh38
rs768197623 Chr.7: 5527725-5527729 on Build GRCh38
rs766542363 Chr.7: 5527899-5527899 on Build GRCh38
rs950871994 Chr.7: 5527624-5527624 on Build GRCh38
rs756549406 Chr.7: 5527710-5527710 on Build GRCh38
rs560972961 Chr.7: 5527695-5527695 on Build GRCh38
rs138910287 Chr.7: 5527900-5527900 on Build GRCh38
rs760087387 Chr.7: 5527530-5527541 on Build GRCh38
rs186180658 Chr.7: 5527679-5527679 on Build GRCh38
rs770822774 Chr.7: 5527731-5527731 on Build GRCh38
rs780969261 Chr.7: 5527850-5527850 on Build GRCh38
rs1064790 Chr.7: 5527862-5527862 on Build GRCh38
rs748905451 Chr.7: 5527705-5527706 on Build GRCh38
rs370848417 Chr.7: 5527787-5527787 on Build GRCh38
rs886041305 Chr.7: 5527855-5527855 on Build GRCh38
rs747696590 Chr.7: 5527724-5527724 on Build GRCh38
rs886041790 Chr.7: 5527856-5527856 on Build GRCh38
rs548291214 Chr.7: 5527914-5527914 on Build GRCh38
rs58704474 Chr.7: 5527853-5527853 on Build GRCh38
rs994400629 Chr.7: 5527843-5527843 on Build GRCh38
rs11546905 Chr.7: 5527468-5527468 on Build GRCh38
rs574890214 Chr.7: 5527651-5527652 on Build GRCh38
rs879167389 Chr.7: 5527618-5527618 on Build GRCh38
rs879140023 Chr.7: 5527623-5527624 on Build GRCh38
rs886041309 Chr.7: 5527833-5527833 on Build GRCh38
rs11546918 Chr.7: 5527513-5527513 on Build GRCh38
rs11546902 Chr.7: 5527851-5527851 on Build GRCh38
rs758025941 Chr.7: 5527820-5527820 on Build GRCh38
rs777620729 Chr.7: 5527728-5527728 on Build GRCh38
rs773181139 Chr.7: 5527877-5527877 on Build GRCh38
rs13447410 Chr.7: 5527648-5527648 on Build GRCh38
rs368352689 Chr.7: 5527786-5527786 on Build GRCh38
rs767758078 Chr.7: 5527704-5527704 on Build GRCh38
rs370106412 Chr.7: 5527624-5527624 on Build GRCh38
rs769620514 Chr.7: 5527745-5527745 on Build GRCh38
rs147293109 Chr.7: 5527676-5527676 on Build GRCh38
rs11546906 Chr.7: 5527488-5527488 on Build GRCh38
rs773005303 Chr.7: 5527460-5527460 on Build GRCh38
rs370911789 Chr.7: 5527719-5527719 on Build GRCh38
rs182601851 Chr.7: 5527474-5527474 on Build GRCh38
rs754429468 Chr.7: 5527716-5527716 on Build GRCh38
rs1009378776 Chr.7: 5527906-5527907 on Build GRCh38
rs11546890 Chr.7: 5527491-5527491 on Build GRCh38
rs13447409 Chr.7: 5527832-5527832 on Build GRCh38
rs973656749 Chr.7: 5527545-5527545 on Build GRCh38
rs775082663 Chr.7: 5527721-5527721 on Build GRCh38
rs1057518142 Chr.7: 5527770-5527771 on Build GRCh38
rs71531321 Chr.7: 5527769-5527769 on Build GRCh38
rs780422687 Chr.7: 5527713-5527713 on Build GRCh38
rs182943508 Chr.7: 5527897-5527897 on Build GRCh38
rs11546941 Chr.7: 5527525-5527525 on Build GRCh38
rs750735339 Chr.7: 5527706-5527706 on Build GRCh38
rs563132910 Chr.7: 5527653-5527653 on Build GRCh38
rs190642491 Chr.7: 5527835-5527835 on Build GRCh38
rs11546935 Chr.7: 5527485-5527485 on Build GRCh38
rs527895375 Chr.7: 5527775-5527775 on Build GRCh38
rs76543773 Chr.7: 5527530-5527530 on Build GRCh38
rs372590587 Chr.7: 5527513-5527513 on Build GRCh38
rs11546903 Chr.7: 5527780-5527780 on Build GRCh38
rs558941094 Chr.7: 5527546-5527546 on Build GRCh38
rs7612 Chr.7: 5527481-5527481 on Build GRCh38
rs1008593723 Chr.7: 5527490-5527490 on Build GRCh38
rs201416978 Chr.7: 5527859-5527859 on Build GRCh38
rs1053587255 Chr.7: 5527456-5527456 on Build GRCh38
rs879017430 Chr.7: 5527651-5527651 on Build GRCh38
rs1065094 Chr.7: 5527661-5527661 on Build GRCh38
rs17136418 Chr.7: 5527646-5527646 on Build GRCh38
rs762140581 Chr.7: 5527820-5527832 on Build GRCh38
rs11546919 Chr.7: 5527516-5527516 on Build GRCh38
rs748194756 Chr.7: 5527865-5527865 on Build GRCh38
rs1018267990 Chr.7: 5527522-5527522 on Build GRCh38
rs761902382 Chr.7: 5527766-5527766 on Build GRCh38
rs544447349 Chr.7: 5527645-5527645 on Build GRCh38
rs757964810 Chr.7: 5527912-5527912 on Build GRCh38
rs772199214 Chr.7: 5527871-5527871 on Build GRCh38
rs771806128 Chr.7: 5527727-5527727 on Build GRCh38
rs759456782 Chr.7: 5527740-5527740 on Build GRCh38
rs371724879 Chr.7: 5527906-5527906 on Build GRCh38
rs367881583 Chr.7: 5527901-5527901 on Build GRCh38
rs760936273 Chr.7: 5527781-5527781 on Build GRCh38
rs777559854 Chr.7: 5527828-5527828 on Build GRCh38
rs770392180 Chr.7: 5527512-5527514 on Build GRCh38
rs767420899 Chr.7: 5527802-5527802 on Build GRCh38

View Full Product Details