Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 95977429-95977937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1I1
Entrez Gene ID
1780
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
DNCI1, DNCIC1
UniGene
Hs.440364
Gene Chromosome Location
Chr.7: 95772506-96110322 on Build GRCh38
Transcripts
NM_001135556.1 NM_001135557.1 NM_001278421.1 NM_001278422.1 NM_004411.4 XM_011515861.1 XM_011515862.1 XM_011515863.2 XM_017011804.1 XM_017011805.1 XM_017011806.1 XM_017011807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2865621 c.585G>T p.Q195H Chr.7: 95977555-95977555 on Build GRCh38
5830768 c.461C>A p.S154Y Chr.7: 95977542-95977542 on Build GRCh38
5608319 c.567G>A p.V189V Chr.7: 95977537-95977537 on Build GRCh38
1093374 c.572C>A p.S191Y Chr.7: 95977542-95977542 on Build GRCh38
7241077 c.575A>T p.K192I Chr.7: 95977545-95977545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004575837 Chr.7: 95977513-95977513 on Build GRCh38
rs370031174 Chr.7: 95977539-95977539 on Build GRCh38
rs574134074 Chr.7: 95977687-95977687 on Build GRCh38
rs550248010 Chr.7: 95977454-95977454 on Build GRCh38
rs763454385 Chr.7: 95977804-95977804 on Build GRCh38
rs937354436 Chr.7: 95977766-95977766 on Build GRCh38
rs545659003 Chr.7: 95977778-95977778 on Build GRCh38
rs755332664 Chr.7: 95977687-95977687 on Build GRCh38
rs200097900 Chr.7: 95977485-95977485 on Build GRCh38
rs779940439 Chr.7: 95977591-95977591 on Build GRCh38
rs773803907 Chr.7: 95977628-95977628 on Build GRCh38
rs967298326 Chr.7: 95977842-95977842 on Build GRCh38
rs377029695 Chr.7: 95977676-95977676 on Build GRCh38
rs913062404 Chr.7: 95977622-95977622 on Build GRCh38
rs185161249 Chr.7: 95977481-95977481 on Build GRCh38
rs770007831 Chr.7: 95977625-95977625 on Build GRCh38
rs573503779 Chr.7: 95977716-95977716 on Build GRCh38
rs150145925 Chr.7: 95977597-95977597 on Build GRCh38
rs550736919 Chr.7: 95977823-95977823 on Build GRCh38
rs769156675 Chr.7: 95977464-95977464 on Build GRCh38
rs748574401 Chr.7: 95977623-95977623 on Build GRCh38
rs950293896 Chr.7: 95977808-95977808 on Build GRCh38
rs952157685 Chr.7: 95977756-95977756 on Build GRCh38
rs755320715 Chr.7: 95977609-95977609 on Build GRCh38
rs376550986 Chr.7: 95977476-95977476 on Build GRCh38
rs987342572 Chr.7: 95977614-95977614 on Build GRCh38
rs141450323 Chr.7: 95977575-95977575 on Build GRCh38
rs546530856 Chr.7: 95977508-95977508 on Build GRCh38
rs376286232 Chr.7: 95977489-95977489 on Build GRCh38
rs762550680 Chr.7: 95977515-95977515 on Build GRCh38
rs559687992 Chr.7: 95977711-95977711 on Build GRCh38
rs775935455 Chr.7: 95977739-95977739 on Build GRCh38
rs752551956 Chr.7: 95977561-95977561 on Build GRCh38
rs1029333730 Chr.7: 95977585-95977585 on Build GRCh38
rs993575878 Chr.7: 95977669-95977669 on Build GRCh38
rs765771171 Chr.7: 95977533-95977533 on Build GRCh38
rs777195827 Chr.7: 95977468-95977468 on Build GRCh38
rs572664287 Chr.7: 95977822-95977822 on Build GRCh38
rs946144145 Chr.7: 95977667-95977667 on Build GRCh38
rs35314029 Chr.7: 95977565-95977565 on Build GRCh38
rs533685174 Chr.7: 95977501-95977502 on Build GRCh38
rs753670141 Chr.7: 95977579-95977579 on Build GRCh38
rs777478919 Chr.7: 95977494-95977495 on Build GRCh38
rs897668376 Chr.7: 95977663-95977663 on Build GRCh38
rs538004157 Chr.7: 95977672-95977672 on Build GRCh38
rs202183816 Chr.7: 95977551-95977551 on Build GRCh38
rs542663285 Chr.7: 95977706-95977706 on Build GRCh38
rs190596245 Chr.7: 95977584-95977584 on Build GRCh38
rs781534363 Chr.7: 95977613-95977613 on Build GRCh38
rs976748379 Chr.7: 95977837-95977837 on Build GRCh38
rs565169517 Chr.7: 95977796-95977796 on Build GRCh38
rs914044208 Chr.7: 95977841-95977841 on Build GRCh38
rs531027301 Chr.7: 95977807-95977807 on Build GRCh38

View Full Product Details