Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128938125-128938619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF5
Entrez Gene ID
3663
Gene Name
interferon regulatory factor 5
Gene Aliases
SLEB10
UniGene
Hs.521181
Gene Chromosome Location
Chr.7: 128937737-128950042 on Build GRCh38
Transcripts
NM_001098629.2 NM_001098630.2 NM_001242452.2 NM_032643.4 XM_006715974.2 XM_011516160.1 XM_011516161.1 XM_011516162.1 XM_011516164.1

SNP Detail

SNP ID Chromosome Location
rs545570324 Chr.7: 128938405-128938405 on Build GRCh38
rs541777175 Chr.7: 128938327-128938327 on Build GRCh38
rs898529810 Chr.7: 128938516-128938516 on Build GRCh38
rs893421503 Chr.7: 128938303-128938303 on Build GRCh38
rs373955841 Chr.7: 128938237-128938237 on Build GRCh38
rs548791514 Chr.7: 128938203-128938203 on Build GRCh38
rs997955105 Chr.7: 128938391-128938391 on Build GRCh38
rs111387050 Chr.7: 128938317-128938317 on Build GRCh38
rs992337755 Chr.7: 128938254-128938254 on Build GRCh38
rs1029474575 Chr.7: 128938396-128938396 on Build GRCh38
rs34629663 Chr.7: 128938174-128938174 on Build GRCh38
rs145527852 Chr.7: 128938583-128938583 on Build GRCh38
rs926973628 Chr.7: 128938585-128938585 on Build GRCh38
rs891097558 Chr.7: 128938187-128938187 on Build GRCh38
rs1055385812 Chr.7: 128938295-128938295 on Build GRCh38
rs142029795 Chr.7: 128938338-128938338 on Build GRCh38
rs985742692 Chr.7: 128938406-128938406 on Build GRCh38
rs573355619 Chr.7: 128938250-128938250 on Build GRCh38
rs528165961 Chr.7: 128938493-128938493 on Build GRCh38
rs780719595 Chr.7: 128938568-128938568 on Build GRCh38
rs6953165 Chr.7: 128938156-128938156 on Build GRCh38
rs573927804 Chr.7: 128938307-128938307 on Build GRCh38
rs147568564 Chr.7: 128938272-128938272 on Build GRCh38
rs962983704 Chr.7: 128938451-128938451 on Build GRCh38
rs74317073 Chr.7: 128938429-128938429 on Build GRCh38
rs2004640 Chr.7: 128938247-128938247 on Build GRCh38
rs985408755 Chr.7: 128938208-128938208 on Build GRCh38
rs540007794 Chr.7: 128938238-128938238 on Build GRCh38
rs1006904410 Chr.7: 128938188-128938188 on Build GRCh38
rs979776756 Chr.7: 128938306-128938306 on Build GRCh38
rs971448385 Chr.7: 128938275-128938275 on Build GRCh38
rs41298401 Chr.7: 128938253-128938253 on Build GRCh38
rs746423919 Chr.7: 128938587-128938587 on Build GRCh38
rs951521419 Chr.7: 128938565-128938565 on Build GRCh38
rs910511173 Chr.7: 128938219-128938219 on Build GRCh38
rs1018751531 Chr.7: 128938193-128938193 on Build GRCh38
rs945869348 Chr.7: 128938411-128938411 on Build GRCh38
rs935754448 Chr.7: 128938373-128938373 on Build GRCh38
rs530650191 Chr.7: 128938356-128938356 on Build GRCh38
rs879264098 Chr.7: 128938464-128938464 on Build GRCh38
rs938232464 Chr.7: 128938289-128938289 on Build GRCh38
rs1054242720 Chr.7: 128938376-128938376 on Build GRCh38
rs1025693395 Chr.7: 128938557-128938557 on Build GRCh38
rs995566069 Chr.7: 128938551-128938551 on Build GRCh38
rs575774110 Chr.7: 128938220-128938220 on Build GRCh38
rs535782578 Chr.7: 128938180-128938180 on Build GRCh38
rs1007242079 Chr.7: 128938479-128938479 on Build GRCh38
rs567967697 Chr.7: 128938569-128938569 on Build GRCh38
rs149688821 Chr.7: 128938567-128938567 on Build GRCh38
rs902247454 Chr.7: 128938358-128938358 on Build GRCh38
rs973851559 Chr.7: 128938243-128938244 on Build GRCh38
rs1051010104 Chr.7: 128938185-128938185 on Build GRCh38

View Full Product Details