Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134575239-134575737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1B15
Entrez Gene ID
441282
Gene Name
aldo-keto reductase family 1 member B15
Gene Aliases
AKR1B10L, AKR1B10L, AK1R1B7, AKR1R1B7
UniGene
Hs.729418
Gene Chromosome Location
Chr.7: 134549097-134579841 on Build GRCh38
Transcripts
NM_001080538.2 XM_011516241.2 XM_017012224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7412141 c.599T>G p.V200G Chr.7: 134575451-134575451 on Build GRCh38
3748915 c.560T>G p.V187G Chr.7: 134575466-134575466 on Build GRCh38
1448198 c.484T>G p.F162V Chr.7: 134575474-134575474 on Build GRCh38
318636 c.569C>T p.A190V Chr.7: 134575421-134575421 on Build GRCh38
5641570 c.480A>C p.S160S Chr.7: 134575470-134575470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375750709 Chr.7: 134575441-134575441 on Build GRCh38
rs373999079 Chr.7: 134575552-134575552 on Build GRCh38
rs541813811 Chr.7: 134575365-134575365 on Build GRCh38
rs577000621 Chr.7: 134575265-134575265 on Build GRCh38
rs766138823 Chr.7: 134575318-134575318 on Build GRCh38
rs767756439 Chr.7: 134575390-134575390 on Build GRCh38
rs566688385 Chr.7: 134575641-134575641 on Build GRCh38
rs762052584 Chr.7: 134575422-134575422 on Build GRCh38
rs753868277 Chr.7: 134575642-134575642 on Build GRCh38
rs760429574 Chr.7: 134575316-134575316 on Build GRCh38
rs997439739 Chr.7: 134575300-134575300 on Build GRCh38
rs756743854 Chr.7: 134575429-134575429 on Build GRCh38
rs2347380 Chr.7: 134575715-134575715 on Build GRCh38
rs779487942 Chr.7: 134575584-134575584 on Build GRCh38
rs372221214 Chr.7: 134575311-134575311 on Build GRCh38
rs376943062 Chr.7: 134575477-134575477 on Build GRCh38
rs745443235 Chr.7: 134575430-134575430 on Build GRCh38
rs373724981 Chr.7: 134575490-134575490 on Build GRCh38
rs867905735 Chr.7: 134575480-134575480 on Build GRCh38
rs34885589 Chr.7: 134575661-134575662 on Build GRCh38
rs961688009 Chr.7: 134575322-134575322 on Build GRCh38
rs372666205 Chr.7: 134575376-134575376 on Build GRCh38
rs763806992 Chr.7: 134575542-134575542 on Build GRCh38
rs757189417 Chr.7: 134575488-134575488 on Build GRCh38
rs2347379 Chr.7: 134575711-134575711 on Build GRCh38
rs765486166 Chr.7: 134575569-134575569 on Build GRCh38
rs1002556017 Chr.7: 134575713-134575713 on Build GRCh38
rs759817570 Chr.7: 134575535-134575535 on Build GRCh38
rs3963353 Chr.7: 134575478-134575478 on Build GRCh38
rs1627864 Chr.7: 134575603-134575603 on Build GRCh38
rs776211531 Chr.7: 134575319-134575319 on Build GRCh38
rs752397346 Chr.7: 134575476-134575476 on Build GRCh38
rs369029578 Chr.7: 134575469-134575469 on Build GRCh38
rs773120093 Chr.7: 134575516-134575516 on Build GRCh38
rs746992591 Chr.7: 134575428-134575428 on Build GRCh38
rs782539 Chr.7: 134575354-134575354 on Build GRCh38
rs190889650 Chr.7: 134575600-134575600 on Build GRCh38
rs773347979 Chr.7: 134575456-134575456 on Build GRCh38
rs754682285 Chr.7: 134575592-134575592 on Build GRCh38
rs563118086 Chr.7: 134575431-134575431 on Build GRCh38
rs972004803 Chr.7: 134575342-134575342 on Build GRCh38
rs189253733 Chr.7: 134575482-134575482 on Build GRCh38
rs767472081 Chr.7: 134575564-134575564 on Build GRCh38
rs759884740 Chr.7: 134575536-134575536 on Build GRCh38
rs3963352 Chr.7: 134575470-134575470 on Build GRCh38
rs767701287 Chr.7: 134575451-134575451 on Build GRCh38
rs73724979 Chr.7: 134575712-134575712 on Build GRCh38
rs748600907 Chr.7: 134575510-134575510 on Build GRCh38
rs1048795566 Chr.7: 134575707-134575707 on Build GRCh38
rs540136299 Chr.7: 134575348-134575348 on Build GRCh38
rs181174304 Chr.7: 134575491-134575491 on Build GRCh38
rs530499864 Chr.7: 134575530-134575530 on Build GRCh38
rs370266956 Chr.7: 134575513-134575513 on Build GRCh38
rs755628322 Chr.7: 134575577-134575577 on Build GRCh38
rs750661134 Chr.7: 134575393-134575393 on Build GRCh38
rs1627777 Chr.7: 134575632-134575632 on Build GRCh38
rs992370981 Chr.7: 134575524-134575524 on Build GRCh38
rs769130202 Chr.7: 134575505-134575505 on Build GRCh38
rs914004150 Chr.7: 134575531-134575531 on Build GRCh38
rs372387354 Chr.7: 134575405-134575405 on Build GRCh38
rs185828325 Chr.7: 134575599-134575599 on Build GRCh38
rs571084916 Chr.7: 134575306-134575306 on Build GRCh38
rs979409454 Chr.7: 134575576-134575576 on Build GRCh38
rs752553310 Chr.7: 134575417-134575417 on Build GRCh38
rs181629071 Chr.7: 134575380-134575380 on Build GRCh38
rs1043941624 Chr.7: 134575268-134575268 on Build GRCh38
rs1732026 Chr.7: 134575685-134575685 on Build GRCh38
rs904082734 Chr.7: 134575283-134575283 on Build GRCh38
rs778795589 Chr.7: 134575714-134575714 on Build GRCh38
rs369342124 Chr.7: 134575437-134575437 on Build GRCh38
rs376060073 Chr.7: 134575378-134575378 on Build GRCh38
rs4035285 Chr.7: 134575440-134575440 on Build GRCh38
rs4035286 Chr.7: 134575496-134575496 on Build GRCh38
rs1732025 Chr.7: 134575701-134575701 on Build GRCh38
rs559180903 Chr.7: 134575486-134575486 on Build GRCh38
rs1027287636 Chr.7: 134575479-134575479 on Build GRCh38
rs753526051 Chr.7: 134575588-134575588 on Build GRCh38
rs992254147 Chr.7: 134575403-134575403 on Build GRCh38
rs368354343 Chr.7: 134575384-134575384 on Build GRCh38
rs375475867 Chr.7: 134575351-134575351 on Build GRCh38
rs557257673 Chr.7: 134575272-134575272 on Build GRCh38
rs186057065 Chr.7: 134575423-134575423 on Build GRCh38
rs1028451605 Chr.7: 134575301-134575301 on Build GRCh38
rs550698469 Chr.7: 134575567-134575567 on Build GRCh38
rs199704947 Chr.7: 134575492-134575492 on Build GRCh38
rs752806230 Chr.7: 134575590-134575591 on Build GRCh38
rs866171696 Chr.7: 134575425-134575425 on Build GRCh38
rs568084043 Chr.7: 134575521-134575521 on Build GRCh38
rs774923025 Chr.7: 134575508-134575508 on Build GRCh38
rs376997972 Chr.7: 134575502-134575502 on Build GRCh38
rs761951313 Chr.7: 134575387-134575387 on Build GRCh38
rs761686077 Chr.7: 134575557-134575557 on Build GRCh38
rs763310650 Chr.7: 134575383-134575383 on Build GRCh38
rs945390811 Chr.7: 134575406-134575406 on Build GRCh38
rs751647404 Chr.7: 134575386-134575386 on Build GRCh38

View Full Product Details