Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134891317-134891828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALD1
Entrez Gene ID
800
Gene Name
caldesmon 1
Gene Aliases
CDM, H-CAD, HCAD, L-CAD, LCAD, NAG22
UniGene
Hs.490203
Gene Chromosome Location
Chr.7: 134779413-134970729 on Build GRCh38
Transcripts
NM_004342.6 NM_033138.3 NM_033139.3 NM_033140.3 NM_033157.3 XM_017012650.1 XM_017012651.1 XM_017012652.1 XM_017012653.1 XM_017012654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1247257 c.15C>T p.S5S Chr.7: 134891635-134891635 on Build GRCh38
7341168 c.31C>A p.R11S Chr.7: 134891651-134891651 on Build GRCh38
6619051 c.32G>A p.R11H Chr.7: 134891652-134891652 on Build GRCh38
7359319 c.17G>T p.G6V Chr.7: 134891637-134891637 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs967143769 Chr.7: 134891577-134891577 on Build GRCh38
rs770169917 Chr.7: 134891588-134891588 on Build GRCh38
rs755470852 Chr.7: 134891505-134891505 on Build GRCh38
rs911132641 Chr.7: 134891579-134891579 on Build GRCh38
rs1031212365 Chr.7: 134891550-134891550 on Build GRCh38
rs774093441 Chr.7: 134891598-134891598 on Build GRCh38
rs869255250 Chr.7: 134891746-134891751 on Build GRCh38
rs777805352 Chr.7: 134891674-134891674 on Build GRCh38
rs1053514539 Chr.7: 134891731-134891731 on Build GRCh38
rs751601069 Chr.7: 134891681-134891681 on Build GRCh38
rs371332466 Chr.7: 134891572-134891572 on Build GRCh38
rs769081176 Chr.7: 134891706-134891707 on Build GRCh38
rs765638052 Chr.7: 134891651-134891651 on Build GRCh38
rs565642394 Chr.7: 134891389-134891389 on Build GRCh38
rs748990540 Chr.7: 134891589-134891589 on Build GRCh38
rs941530848 Chr.7: 134891704-134891704 on Build GRCh38
rs757352524 Chr.7: 134891688-134891688 on Build GRCh38
rs551050301 Chr.7: 134891499-134891499 on Build GRCh38
rs977966716 Chr.7: 134891523-134891523 on Build GRCh38
rs890555144 Chr.7: 134891421-134891421 on Build GRCh38
rs796965110 Chr.7: 134891769-134891769 on Build GRCh38
rs994620526 Chr.7: 134891538-134891538 on Build GRCh38
rs562945349 Chr.7: 134891785-134891785 on Build GRCh38
rs768513971 Chr.7: 134891597-134891597 on Build GRCh38
rs375797843 Chr.7: 134891618-134891618 on Build GRCh38
rs376167771 Chr.7: 134891716-134891716 on Build GRCh38
rs796165154 Chr.7: 134891640-134891640 on Build GRCh38
rs922368355 Chr.7: 134891661-134891661 on Build GRCh38
rs545270213 Chr.7: 134891765-134891768 on Build GRCh38
rs1046747552 Chr.7: 134891781-134891781 on Build GRCh38
rs760374572 Chr.7: 134891617-134891617 on Build GRCh38
rs945052731 Chr.7: 134891777-134891777 on Build GRCh38
rs958831798 Chr.7: 134891565-134891565 on Build GRCh38
rs753135479 Chr.7: 134891656-134891656 on Build GRCh38
rs776128097 Chr.7: 134891664-134891664 on Build GRCh38
rs758959249 Chr.7: 134891662-134891662 on Build GRCh38
rs368688174 Chr.7: 134891582-134891582 on Build GRCh38
rs1054072101 Chr.7: 134891382-134891382 on Build GRCh38
rs900745482 Chr.7: 134891383-134891383 on Build GRCh38
rs929703880 Chr.7: 134891663-134891663 on Build GRCh38
rs759761507 Chr.7: 134891632-134891632 on Build GRCh38
rs767831408 Chr.7: 134891711-134891712 on Build GRCh38
rs779338929 Chr.7: 134891709-134891709 on Build GRCh38
rs12531510 Chr.7: 134891796-134891796 on Build GRCh38
rs201422064 Chr.7: 134891689-134891689 on Build GRCh38
rs892153461 Chr.7: 134891744-134891744 on Build GRCh38
rs756601183 Chr.7: 134891455-134891455 on Build GRCh38
rs200143478 Chr.7: 134891562-134891562 on Build GRCh38
rs114103910 Chr.7: 134891429-134891429 on Build GRCh38
rs559607001 Chr.7: 134891390-134891390 on Build GRCh38
rs867614112 Chr.7: 134891435-134891435 on Build GRCh38
rs765576423 Chr.7: 134891747-134891748 on Build GRCh38
rs751163669 Chr.7: 134891403-134891403 on Build GRCh38
rs753548867 Chr.7: 134891626-134891626 on Build GRCh38
rs992514185 Chr.7: 134891571-134891571 on Build GRCh38
rs561902864 Chr.7: 134891436-134891436 on Build GRCh38
rs771695787 Chr.7: 134891612-134891612 on Build GRCh38
rs577062522 Chr.7: 134891722-134891722 on Build GRCh38
rs33997290 Chr.7: 134891768-134891768 on Build GRCh38
rs77382246 Chr.7: 134891715-134891715 on Build GRCh38
rs199881302 Chr.7: 134891729-134891729 on Build GRCh38
rs899030985 Chr.7: 134891519-134891519 on Build GRCh38
rs990013785 Chr.7: 134891646-134891646 on Build GRCh38
rs781427024 Chr.7: 134891583-134891583 on Build GRCh38
rs75054986 Chr.7: 134891407-134891407 on Build GRCh38
rs137987656 Chr.7: 134891492-134891492 on Build GRCh38
rs540840644 Chr.7: 134891746-134891746 on Build GRCh38
rs114957651 Chr.7: 134891581-134891581 on Build GRCh38
rs1014940689 Chr.7: 134891504-134891504 on Build GRCh38
rs201096071 Chr.7: 134891608-134891608 on Build GRCh38
rs894514532 Chr.7: 134891483-134891483 on Build GRCh38
rs749653457 Chr.7: 134891706-134891709 on Build GRCh38
rs6953822 Chr.7: 134891690-134891690 on Build GRCh38
rs942359918 Chr.7: 134891348-134891348 on Build GRCh38
rs758431023 Chr.7: 134891746-134891748 on Build GRCh38
rs368180740 Chr.7: 134891724-134891724 on Build GRCh38
rs62479555 Chr.7: 134891370-134891370 on Build GRCh38
rs1034363713 Chr.7: 134891465-134891465 on Build GRCh38
rs768332503 Chr.7: 134891697-134891697 on Build GRCh38
rs774857666 Chr.7: 134891711-134891711 on Build GRCh38
rs569914986 Chr.7: 134891547-134891547 on Build GRCh38
rs772900229 Chr.7: 134891614-134891614 on Build GRCh38
rs868656654 Chr.7: 134891637-134891637 on Build GRCh38
rs10607357 Chr.7: 134891767-134891768 on Build GRCh38
rs762081711 Chr.7: 134891710-134891711 on Build GRCh38
rs910008481 Chr.7: 134891694-134891695 on Build GRCh38

View Full Product Details