Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135134565-135135069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL3
Entrez Gene ID
340351
Gene Name
ATP/GTP binding protein like 3
Gene Aliases
CCP3
UniGene
Hs.648616
Gene Chromosome Location
Chr.7: 134985169-135135782 on Build GRCh38
Transcripts
NM_178563.3 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1 NM_001305630.1 XM_017012595.1

Gene Symbol
C7orf49
Entrez Gene ID
78996
Gene Name
chromosome 7 open reading frame 49
Gene Aliases
MRI
UniGene
Hs.643113
Gene Chromosome Location
Chr.7: 135092303-135170826 on Build GRCh38
Transcripts
NM_178563.3 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1 NM_001305630.1 XM_017012595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4649062 c.2533T>C p.W845R Chr.7: 135135031-135135031 on Build GRCh38
5574383 c.2568C>A p.S856R Chr.7: 135135066-135135066 on Build GRCh38
1448234 c.2527C>A p.Q843K Chr.7: 135135025-135135025 on Build GRCh38
4649063 c.2698T>C p.W900R Chr.7: 135135031-135135031 on Build GRCh38
5574384 c.2733C>A p.S911R Chr.7: 135135066-135135066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056280883 Chr.7: 135134873-135134873 on Build GRCh38
rs11765077 Chr.7: 135134714-135134714 on Build GRCh38
rs768611754 Chr.7: 135134864-135134864 on Build GRCh38
rs967676613 Chr.7: 135134693-135134693 on Build GRCh38
rs767394407 Chr.7: 135134665-135134666 on Build GRCh38
rs560482469 Chr.7: 135134718-135134718 on Build GRCh38
rs556335506 Chr.7: 135134631-135134631 on Build GRCh38
rs761245376 Chr.7: 135134881-135134881 on Build GRCh38
rs200672886 Chr.7: 135134666-135134667 on Build GRCh38
rs530876622 Chr.7: 135134839-135134839 on Build GRCh38
rs774104669 Chr.7: 135134850-135134850 on Build GRCh38
rs760068106 Chr.7: 135135000-135135000 on Build GRCh38
rs6953478 Chr.7: 135134640-135134640 on Build GRCh38
rs148526262 Chr.7: 135134849-135134849 on Build GRCh38
rs761424217 Chr.7: 135134924-135134924 on Build GRCh38
rs772797432 Chr.7: 135134934-135134934 on Build GRCh38
rs758476920 Chr.7: 135135013-135135013 on Build GRCh38
rs977025978 Chr.7: 135134716-135134721 on Build GRCh38
rs1017717921 Chr.7: 135134987-135134987 on Build GRCh38
rs1022071690 Chr.7: 135134650-135134650 on Build GRCh38
rs752757788 Chr.7: 135135009-135135009 on Build GRCh38
rs183850209 Chr.7: 135134866-135134866 on Build GRCh38
rs893666876 Chr.7: 135134893-135134893 on Build GRCh38
rs942980469 Chr.7: 135134708-135134708 on Build GRCh38
rs539117778 Chr.7: 135134936-135134936 on Build GRCh38
rs116331035 Chr.7: 135134832-135134832 on Build GRCh38
rs892280382 Chr.7: 135134933-135134933 on Build GRCh38
rs1022019458 Chr.7: 135134953-135134953 on Build GRCh38
rs967828793 Chr.7: 135134958-135134958 on Build GRCh38
rs909947169 Chr.7: 135134804-135134804 on Build GRCh38
rs779012183 Chr.7: 135134666-135134666 on Build GRCh38
rs766890067 Chr.7: 135134988-135134988 on Build GRCh38
rs1047109076 Chr.7: 135134816-135134816 on Build GRCh38
rs144725955 Chr.7: 135134790-135134790 on Build GRCh38
rs552665313 Chr.7: 135135008-135135008 on Build GRCh38
rs1007218712 Chr.7: 135134970-135134970 on Build GRCh38
rs1056313149 Chr.7: 135134823-135134827 on Build GRCh38
rs951494812 Chr.7: 135134749-135134749 on Build GRCh38
rs866294061 Chr.7: 135134979-135134979 on Build GRCh38
rs777865214 Chr.7: 135134821-135134821 on Build GRCh38
rs571346751 Chr.7: 135134698-135134698 on Build GRCh38
rs911475096 Chr.7: 135134706-135134706 on Build GRCh38
rs548418384 Chr.7: 135134810-135134810 on Build GRCh38
rs754281649 Chr.7: 135134993-135134993 on Build GRCh38
rs758586171 Chr.7: 135134791-135134791 on Build GRCh38
rs1010635774 Chr.7: 135134967-135134967 on Build GRCh38
rs922785949 Chr.7: 135134764-135134764 on Build GRCh38
rs1029883211 Chr.7: 135135042-135135045 on Build GRCh38

View Full Product Details