Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 16992744-16993247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF20
Entrez Gene ID
26281
Gene Name
fibroblast growth factor 20
Gene Aliases
FGF-20, RHDA2
UniGene
Hs.199905
Gene Chromosome Location
Chr.8: 16992825-17002165 on Build GRCh38
Transcripts
NM_019851.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834343 c.586G>A p.D196N Chr.8: 16993122-16993122 on Build GRCh38
3834344 c.558A>G p.K186K Chr.8: 16993150-16993150 on Build GRCh38
6472460 c.569T>C p.F190S Chr.8: 16993139-16993139 on Build GRCh38
6663530 c.517G>A p.G173R Chr.8: 16993191-16993191 on Build GRCh38
3924935 c.501G>A p.V167V Chr.8: 16993207-16993207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113374862 Chr.8: 16992918-16992918 on Build GRCh38
rs116309944 Chr.8: 16992918-16992918 on Build GRCh38
rs774693591 Chr.8: 16993062-16993062 on Build GRCh38
rs999003744 Chr.8: 16992787-16992787 on Build GRCh38
rs773678490 Chr.8: 16993199-16993199 on Build GRCh38
rs369979222 Chr.8: 16993064-16993064 on Build GRCh38
rs572374220 Chr.8: 16992926-16992926 on Build GRCh38
rs376283820 Chr.8: 16993124-16993124 on Build GRCh38
rs556236317 Chr.8: 16993153-16993153 on Build GRCh38
rs1040204264 Chr.8: 16993108-16993108 on Build GRCh38
rs1041831813 Chr.8: 16992785-16992785 on Build GRCh38
rs1721100 Chr.8: 16992989-16992989 on Build GRCh38
rs376980441 Chr.8: 16993192-16993192 on Build GRCh38
rs376046538 Chr.8: 16993173-16993173 on Build GRCh38
rs780961429 Chr.8: 16993080-16993080 on Build GRCh38
rs1017087794 Chr.8: 16993035-16993035 on Build GRCh38
rs569516194 Chr.8: 16992959-16992959 on Build GRCh38
rs1045775155 Chr.8: 16992946-16992946 on Build GRCh38
rs755939016 Chr.8: 16993161-16993161 on Build GRCh38
rs769791216 Chr.8: 16993051-16993051 on Build GRCh38
rs764058382 Chr.8: 16993138-16993138 on Build GRCh38
rs749155129 Chr.8: 16993168-16993168 on Build GRCh38
rs975825506 Chr.8: 16992882-16992882 on Build GRCh38
rs751701947 Chr.8: 16992849-16992849 on Build GRCh38
rs780521897 Chr.8: 16993069-16993069 on Build GRCh38
rs372796901 Chr.8: 16993098-16993098 on Build GRCh38
rs182573765 Chr.8: 16992976-16992976 on Build GRCh38
rs951641387 Chr.8: 16992885-16992885 on Build GRCh38
rs760365627 Chr.8: 16993220-16993220 on Build GRCh38
rs984994210 Chr.8: 16993042-16993042 on Build GRCh38
rs774471972 Chr.8: 16993062-16993062 on Build GRCh38
rs921238581 Chr.8: 16993167-16993167 on Build GRCh38
rs762222483 Chr.8: 16993193-16993193 on Build GRCh38
rs754685284 Chr.8: 16993078-16993078 on Build GRCh38
rs1024379139 Chr.8: 16992777-16992777 on Build GRCh38
rs777035608 Chr.8: 16993134-16993134 on Build GRCh38
rs564045462 Chr.8: 16992911-16992911 on Build GRCh38
rs1029805434 Chr.8: 16992858-16992858 on Build GRCh38
rs375846209 Chr.8: 16993076-16993076 on Build GRCh38
rs747943357 Chr.8: 16993083-16993083 on Build GRCh38
rs147590629 Chr.8: 16992917-16992917 on Build GRCh38
rs17550360 Chr.8: 16993092-16993092 on Build GRCh38
rs777595585 Chr.8: 16993084-16993084 on Build GRCh38
rs574070314 Chr.8: 16992887-16992887 on Build GRCh38
rs764711524 Chr.8: 16993042-16993043 on Build GRCh38
rs769148271 Chr.8: 16992886-16992886 on Build GRCh38
rs753990768 Chr.8: 16993160-16993160 on Build GRCh38
rs1051138244 Chr.8: 16993188-16993188 on Build GRCh38
rs112602726 Chr.8: 16992952-16992952 on Build GRCh38
rs752318885 Chr.8: 16993217-16993217 on Build GRCh38
rs530611016 Chr.8: 16992903-16992903 on Build GRCh38
rs566436899 Chr.8: 16992922-16992922 on Build GRCh38
rs773213151 Chr.8: 16993055-16993055 on Build GRCh38
rs17515351 Chr.8: 16992829-16992829 on Build GRCh38
rs768410520 Chr.8: 16993034-16993034 on Build GRCh38
rs767173525 Chr.8: 16993159-16993159 on Build GRCh38
rs758002395 Chr.8: 16993022-16993022 on Build GRCh38
rs998865931 Chr.8: 16992850-16992850 on Build GRCh38
rs759071753 Chr.8: 16993211-16993211 on Build GRCh38
rs904484059 Chr.8: 16992804-16992804 on Build GRCh38
rs1000193361 Chr.8: 16992825-16992825 on Build GRCh38
rs777667380 Chr.8: 16993165-16993165 on Build GRCh38
rs113126203 Chr.8: 16993038-16993038 on Build GRCh38
rs865776582 Chr.8: 16993075-16993075 on Build GRCh38
rs771137602 Chr.8: 16993059-16993059 on Build GRCh38
rs900067949 Chr.8: 16992845-16992845 on Build GRCh38
rs946124140 Chr.8: 16992781-16992781 on Build GRCh38
rs764646776 Chr.8: 16993127-16993127 on Build GRCh38
rs746261060 Chr.8: 16992908-16992908 on Build GRCh38
rs142106513 Chr.8: 16992924-16992924 on Build GRCh38
rs373460171 Chr.8: 16992931-16992931 on Build GRCh38
rs573963889 Chr.8: 16993077-16993077 on Build GRCh38
rs975773082 Chr.8: 16993020-16993020 on Build GRCh38
rs770340289 Chr.8: 16993196-16993196 on Build GRCh38
rs370810066 Chr.8: 16993137-16993137 on Build GRCh38
rs761133332 Chr.8: 16993126-16993126 on Build GRCh38
rs369523536 Chr.8: 16993025-16993025 on Build GRCh38
rs937947933 Chr.8: 16992918-16992919 on Build GRCh38
rs767120553 Chr.8: 16993216-16993216 on Build GRCh38
rs1033200361 Chr.8: 16992975-16992975 on Build GRCh38
rs575400716 Chr.8: 16993057-16993057 on Build GRCh38
rs749890928 Chr.8: 16993070-16993070 on Build GRCh38
rs774535729 Chr.8: 16993095-16993095 on Build GRCh38
rs10089600 Chr.8: 16993185-16993185 on Build GRCh38
rs12720208 Chr.8: 16992890-16992890 on Build GRCh38
rs2640901 Chr.8: 16992907-16992907 on Build GRCh38
rs746061786 Chr.8: 16993096-16993096 on Build GRCh38
rs746767850 Chr.8: 16993030-16993030 on Build GRCh38
rs762432192 Chr.8: 16993135-16993135 on Build GRCh38
rs965682514 Chr.8: 16993019-16993019 on Build GRCh38
rs779979768 Chr.8: 16992906-16992907 on Build GRCh38
rs772360437 Chr.8: 16993184-16993184 on Build GRCh38
rs377266598 Chr.8: 16993046-16993046 on Build GRCh38
rs936045906 Chr.8: 16993107-16993107 on Build GRCh38
rs6982917 Chr.8: 16992961-16992961 on Build GRCh38
rs560503402 Chr.8: 16992854-16992854 on Build GRCh38
rs534476986 Chr.8: 16992986-16992986 on Build GRCh38
rs965734689 Chr.8: 16992873-16992873 on Build GRCh38
rs578238179 Chr.8: 16992806-16992806 on Build GRCh38
rs1004331933 Chr.8: 16993024-16993024 on Build GRCh38
rs767969554 Chr.8: 16993066-16993066 on Build GRCh38
rs904598532 Chr.8: 16992925-16992925 on Build GRCh38
rs372623598 Chr.8: 16993067-16993067 on Build GRCh38
rs533487160 Chr.8: 16992889-16992889 on Build GRCh38
rs373023438 Chr.8: 16993174-16993174 on Build GRCh38
rs140595599 Chr.8: 16992984-16992984 on Build GRCh38
rs998417008 Chr.8: 16992977-16992977 on Build GRCh38

View Full Product Details