Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22120932-22121440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HR
Entrez Gene ID
55806
Gene Name
hair growth associated
Gene Aliases
ALUNC, AU, HSA277165, HYPT4, MUHH, MUHH1
UniGene
Hs.272367
Gene Chromosome Location
Chr.8: 22114419-22131053 on Build GRCh38
Transcripts
NM_005144.4 NM_018411.4 XM_005273569.1 XM_006716367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3898846 c.2341G>A p.A781T Chr.8: 22121091-22121091 on Build GRCh38
3898847 c.2239C>T p.R747C Chr.8: 22121193-22121193 on Build GRCh38
7175645 c.2318A>T p.H773L Chr.8: 22121114-22121114 on Build GRCh38
3898845 c.2355G>A p.P785P Chr.8: 22121077-22121077 on Build GRCh38
5109917 c.2212G>A p.D738N Chr.8: 22121220-22121220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911761521 Chr.8: 22121373-22121373 on Build GRCh38
rs112532092 Chr.8: 22121051-22121051 on Build GRCh38
rs962568434 Chr.8: 22121244-22121244 on Build GRCh38
rs574322741 Chr.8: 22121137-22121137 on Build GRCh38
rs886062807 Chr.8: 22121220-22121220 on Build GRCh38
rs146360676 Chr.8: 22121002-22121002 on Build GRCh38
rs781478489 Chr.8: 22121088-22121088 on Build GRCh38
rs1039786928 Chr.8: 22121381-22121381 on Build GRCh38
rs867523769 Chr.8: 22120959-22120959 on Build GRCh38
rs758171223 Chr.8: 22121268-22121268 on Build GRCh38
rs767931701 Chr.8: 22121258-22121258 on Build GRCh38
rs547812095 Chr.8: 22121299-22121299 on Build GRCh38
rs949383553 Chr.8: 22121416-22121416 on Build GRCh38
rs373625154 Chr.8: 22121058-22121058 on Build GRCh38
rs760095677 Chr.8: 22121181-22121181 on Build GRCh38
rs151306827 Chr.8: 22121077-22121077 on Build GRCh38
rs745393302 Chr.8: 22121108-22121108 on Build GRCh38
rs994258864 Chr.8: 22120961-22120961 on Build GRCh38
rs772166094 Chr.8: 22121050-22121050 on Build GRCh38
rs139027208 Chr.8: 22121214-22121214 on Build GRCh38
rs925436907 Chr.8: 22121357-22121357 on Build GRCh38
rs910950666 Chr.8: 22121309-22121309 on Build GRCh38
rs375913911 Chr.8: 22121086-22121086 on Build GRCh38
rs751142062 Chr.8: 22121275-22121275 on Build GRCh38
rs960749402 Chr.8: 22120955-22120955 on Build GRCh38
rs770987134 Chr.8: 22121253-22121253 on Build GRCh38
rs560464452 Chr.8: 22121310-22121310 on Build GRCh38
rs747921321 Chr.8: 22121210-22121210 on Build GRCh38
rs543527133 Chr.8: 22121283-22121283 on Build GRCh38
rs77689228 Chr.8: 22121221-22121221 on Build GRCh38
rs765297729 Chr.8: 22121003-22121003 on Build GRCh38
rs145225497 Chr.8: 22121134-22121134 on Build GRCh38
rs574361484 Chr.8: 22121215-22121215 on Build GRCh38
rs781003868 Chr.8: 22121205-22121205 on Build GRCh38
rs191763994 Chr.8: 22121001-22121001 on Build GRCh38
rs201308362 Chr.8: 22121192-22121192 on Build GRCh38
rs747006592 Chr.8: 22121135-22121135 on Build GRCh38
rs775866528 Chr.8: 22121259-22121259 on Build GRCh38
rs942855695 Chr.8: 22121091-22121091 on Build GRCh38
rs749884432 Chr.8: 22121190-22121190 on Build GRCh38
rs745796869 Chr.8: 22121047-22121047 on Build GRCh38
rs746916573 Chr.8: 22121194-22121194 on Build GRCh38
rs999446705 Chr.8: 22121227-22121227 on Build GRCh38
rs751738208 Chr.8: 22121363-22121363 on Build GRCh38
rs576462393 Chr.8: 22121261-22121261 on Build GRCh38
rs899830500 Chr.8: 22121387-22121387 on Build GRCh38
rs745641192 Chr.8: 22121015-22121015 on Build GRCh38
rs139655818 Chr.8: 22121314-22121314 on Build GRCh38
rs770472945 Chr.8: 22121029-22121029 on Build GRCh38
rs778445391 Chr.8: 22121103-22121103 on Build GRCh38
rs770717791 Chr.8: 22120962-22120962 on Build GRCh38
rs1043289162 Chr.8: 22121402-22121402 on Build GRCh38
rs370739747 Chr.8: 22121254-22121254 on Build GRCh38
rs146456920 Chr.8: 22121154-22121154 on Build GRCh38
rs765951873 Chr.8: 22121273-22121273 on Build GRCh38
rs570581049 Chr.8: 22121362-22121362 on Build GRCh38
rs759459355 Chr.8: 22120968-22120968 on Build GRCh38
rs751956380 Chr.8: 22121018-22121018 on Build GRCh38
rs368840485 Chr.8: 22121027-22121027 on Build GRCh38
rs756910909 Chr.8: 22121101-22121101 on Build GRCh38
rs564376330 Chr.8: 22121260-22121260 on Build GRCh38
rs140534514 Chr.8: 22121078-22121078 on Build GRCh38
rs150013665 Chr.8: 22121114-22121114 on Build GRCh38
rs761995462 Chr.8: 22121067-22121067 on Build GRCh38
rs760558633 Chr.8: 22121063-22121063 on Build GRCh38
rs991154565 Chr.8: 22121087-22121087 on Build GRCh38
rs376900561 Chr.8: 22121297-22121297 on Build GRCh38
rs750084580 Chr.8: 22121265-22121265 on Build GRCh38
rs142343325 Chr.8: 22121136-22121136 on Build GRCh38
rs368256327 Chr.8: 22121166-22121166 on Build GRCh38
rs184050689 Chr.8: 22121317-22121317 on Build GRCh38
rs765024062 Chr.8: 22121278-22121278 on Build GRCh38
rs1012276789 Chr.8: 22121042-22121042 on Build GRCh38
rs57903149 Chr.8: 22120956-22120956 on Build GRCh38
rs553608835 Chr.8: 22121316-22121316 on Build GRCh38
rs748681773 Chr.8: 22121028-22121028 on Build GRCh38
rs1016775425 Chr.8: 22121024-22121024 on Build GRCh38
rs780368795 Chr.8: 22121016-22121016 on Build GRCh38
rs376714428 Chr.8: 22121284-22121284 on Build GRCh38
rs753516082 Chr.8: 22121092-22121092 on Build GRCh38
rs766897426 Chr.8: 22121085-22121085 on Build GRCh38
rs896768051 Chr.8: 22121412-22121412 on Build GRCh38
rs775465022 Chr.8: 22120979-22120979 on Build GRCh38
rs367903979 Chr.8: 22120992-22120992 on Build GRCh38
rs750923997 Chr.8: 22121007-22121007 on Build GRCh38
rs752059018 Chr.8: 22121169-22121169 on Build GRCh38
rs755716128 Chr.8: 22121025-22121025 on Build GRCh38
rs1052671567 Chr.8: 22121380-22121380 on Build GRCh38
rs370033127 Chr.8: 22121064-22121064 on Build GRCh38
rs779765704 Chr.8: 22121193-22121193 on Build GRCh38
rs200306863 Chr.8: 22121183-22121183 on Build GRCh38
rs756883750 Chr.8: 22121187-22121187 on Build GRCh38
rs987274714 Chr.8: 22121333-22121333 on Build GRCh38
rs763036949 Chr.8: 22121155-22121155 on Build GRCh38
rs754908515 Chr.8: 22121201-22121201 on Build GRCh38
rs778344701 Chr.8: 22121041-22121041 on Build GRCh38
rs764431917 Chr.8: 22120996-22120996 on Build GRCh38
rs765334789 Chr.8: 22121074-22121074 on Build GRCh38
rs769944653 Chr.8: 22121149-22121149 on Build GRCh38
rs772672266 Chr.8: 22120952-22120952 on Build GRCh38
rs947388354 Chr.8: 22121396-22121396 on Build GRCh38
rs904066099 Chr.8: 22121109-22121109 on Build GRCh38
rs772074243 Chr.8: 22121113-22121113 on Build GRCh38
rs58080661 Chr.8: 22121065-22121065 on Build GRCh38
rs774352039 Chr.8: 22120966-22120966 on Build GRCh38
rs767407285 Chr.8: 22120971-22120971 on Build GRCh38
rs1038060015 Chr.8: 22121140-22121140 on Build GRCh38
rs371514025 Chr.8: 22121189-22121189 on Build GRCh38

View Full Product Details