Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179385058-179385587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXDND1
Entrez Gene ID
126859
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
C1orf125
UniGene
Hs.658505
Gene Chromosome Location
Chr.1: 179365705-179554735 on Build GRCh38
Transcripts
NM_144696.5 NR_073544.1 XM_011509165.1 XM_011509166.2 XM_011509167.2 XM_011509168.1 XM_011509169.1 XM_011509170.1 XM_011509171.1 XM_011509174.1 XM_011509175.1 XM_011509176.1 XM_011509178.2 XM_011509179.2 XM_011509180.1 XM_017000257.1 XM_017000258.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5891570 c.806T>C p.L269P Chr.1: 179385302-179385302 on Build GRCh38
7234346 c.863+10G>T p.? Chr.1: 179385369-179385369 on Build GRCh38
5378646 c.811C>T p.R271* Chr.1: 179385307-179385307 on Build GRCh38
3725316 c.764T>G p.L255W Chr.1: 179385260-179385260 on Build GRCh38
6459704 c.854C>T p.S285F Chr.1: 179385350-179385350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749311397 Chr.1: 179385398-179385398 on Build GRCh38
rs750630646 Chr.1: 179385366-179385366 on Build GRCh38
rs751616896 Chr.1: 179385348-179385348 on Build GRCh38
rs535982484 Chr.1: 179385205-179385205 on Build GRCh38
rs560301086 Chr.1: 179385451-179385451 on Build GRCh38
rs982413595 Chr.1: 179385471-179385471 on Build GRCh38
rs974629511 Chr.1: 179385420-179385420 on Build GRCh38
rs368746160 Chr.1: 179385305-179385305 on Build GRCh38
rs140324387 Chr.1: 179385422-179385422 on Build GRCh38
rs188823983 Chr.1: 179385381-179385381 on Build GRCh38
rs866821341 Chr.1: 179385354-179385354 on Build GRCh38
rs890492737 Chr.1: 179385256-179385256 on Build GRCh38
rs144830533 Chr.1: 179385138-179385138 on Build GRCh38
rs1051315212 Chr.1: 179385489-179385489 on Build GRCh38
rs183872681 Chr.1: 179385139-179385139 on Build GRCh38
rs774832484 Chr.1: 179385316-179385316 on Build GRCh38
rs752056204 Chr.1: 179385227-179385227 on Build GRCh38
rs746095759 Chr.1: 179385259-179385259 on Build GRCh38
rs762573163 Chr.1: 179385328-179385328 on Build GRCh38
rs1046502784 Chr.1: 179385149-179385149 on Build GRCh38
rs369480240 Chr.1: 179385202-179385202 on Build GRCh38
rs544858689 Chr.1: 179385287-179385287 on Build GRCh38
rs191687362 Chr.1: 179385137-179385137 on Build GRCh38
rs529247119 Chr.1: 179385461-179385461 on Build GRCh38
rs941569808 Chr.1: 179385395-179385395 on Build GRCh38
rs747665105 Chr.1: 179385240-179385240 on Build GRCh38
rs139773041 Chr.1: 179385360-179385360 on Build GRCh38
rs556255231 Chr.1: 179385254-179385254 on Build GRCh38
rs937660380 Chr.1: 179385159-179385159 on Build GRCh38
rs758300489 Chr.1: 179385242-179385242 on Build GRCh38
rs767435848 Chr.1: 179385362-179385362 on Build GRCh38
rs937730332 Chr.1: 179385523-179385523 on Build GRCh38
rs202024026 Chr.1: 179385204-179385204 on Build GRCh38
rs769017403 Chr.1: 179385286-179385286 on Build GRCh38
rs772429676 Chr.1: 179385296-179385296 on Build GRCh38
rs927675051 Chr.1: 179385155-179385155 on Build GRCh38
rs890165098 Chr.1: 179385525-179385525 on Build GRCh38
rs199966358 Chr.1: 179385289-179385289 on Build GRCh38
rs369737277 Chr.1: 179385368-179385368 on Build GRCh38
rs371174580 Chr.1: 179385214-179385214 on Build GRCh38
rs1021108171 Chr.1: 179385431-179385431 on Build GRCh38
rs540890458 Chr.1: 179385436-179385436 on Build GRCh38
rs757180091 Chr.1: 179385255-179385255 on Build GRCh38
rs998091467 Chr.1: 179385222-179385222 on Build GRCh38
rs776331360 Chr.1: 179385218-179385218 on Build GRCh38
rs900073106 Chr.1: 179385118-179385118 on Build GRCh38
rs751290512 Chr.1: 179385252-179385252 on Build GRCh38
rs762209849 Chr.1: 179385212-179385212 on Build GRCh38
rs755153551 Chr.1: 179385409-179385409 on Build GRCh38
rs749422483 Chr.1: 179385279-179385279 on Build GRCh38
rs888193081 Chr.1: 179385197-179385197 on Build GRCh38
rs781564646 Chr.1: 179385258-179385258 on Build GRCh38
rs752633154 Chr.1: 179385241-179385241 on Build GRCh38
rs763929568 Chr.1: 179385330-179385330 on Build GRCh38
rs779338477 Chr.1: 179385238-179385238 on Build GRCh38
rs1017702941 Chr.1: 179385331-179385331 on Build GRCh38
rs928198049 Chr.1: 179385521-179385521 on Build GRCh38
rs144196620 Chr.1: 179385487-179385487 on Build GRCh38
rs771514964 Chr.1: 179385270-179385270 on Build GRCh38
rs374786188 Chr.1: 179385313-179385313 on Build GRCh38
rs542756993 Chr.1: 179385470-179385470 on Build GRCh38
rs575915608 Chr.1: 179385264-179385264 on Build GRCh38
rs375232831 Chr.1: 179385201-179385201 on Build GRCh38
rs758979423 Chr.1: 179385228-179385228 on Build GRCh38
rs7541905 Chr.1: 179385169-179385169 on Build GRCh38
rs764766519 Chr.1: 179385308-179385308 on Build GRCh38
rs764132304 Chr.1: 179385247-179385247 on Build GRCh38
rs774544416 Chr.1: 179385209-179385209 on Build GRCh38
rs565266822 Chr.1: 179385488-179385489 on Build GRCh38
rs559609813 Chr.1: 179385157-179385157 on Build GRCh38
rs149782250 Chr.1: 179385307-179385307 on Build GRCh38

View Full Product Details