Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108943570-108944076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNAL1
Entrez Gene ID
8727
Gene Name
catenin alpha like 1
Gene Aliases
ACRP, CLLP, alpha-CATU
UniGene
Hs.58488
Gene Chromosome Location
Chr.9: 108942569-109013594 on Build GRCh38
Transcripts
NM_001286974.1 NM_003798.3 XM_005252291.3 XM_017015250.1 XM_017015251.1 XM_011518816.2

Gene Symbol
FAM206A
Entrez Gene ID
54942
Gene Name
family with sequence similarity 206 member A
Gene Aliases
C9orf6, CG-8, Simiate
UniGene
Hs.29276
Gene Chromosome Location
Chr.9: 108934393-108950745 on Build GRCh38
Transcripts
NM_001286974.1 NM_003798.3 XM_005252291.3 XM_017015250.1 XM_017015251.1 XM_011518816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6575761 c.1975G>A p.E659K Chr.9: 108943783-108943783 on Build GRCh38
6575762 c.1975G>A p.E659K Chr.9: 108943783-108943783 on Build GRCh38
6312133 c.2001C>A p.C667* Chr.9: 108943757-108943757 on Build GRCh38
7306429 c.1885-7C>T p.? Chr.9: 108944025-108944025 on Build GRCh38
4675391 c.1951G>A p.D651N Chr.9: 108943807-108943807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761403846 Chr.9: 108943762-108943762 on Build GRCh38
rs763814785 Chr.9: 108943757-108943757 on Build GRCh38
rs777140399 Chr.9: 108943862-108943862 on Build GRCh38
rs1036501332 Chr.9: 108943714-108943714 on Build GRCh38
rs778130522 Chr.9: 108943979-108943980 on Build GRCh38
rs749583352 Chr.9: 108943805-108943805 on Build GRCh38
rs1031600118 Chr.9: 108943990-108943990 on Build GRCh38
rs752506638 Chr.9: 108944010-108944010 on Build GRCh38
rs758462515 Chr.9: 108944036-108944036 on Build GRCh38
rs769205844 Chr.9: 108943860-108943860 on Build GRCh38
rs1035182236 Chr.9: 108943712-108943712 on Build GRCh38
rs764734016 Chr.9: 108943765-108943765 on Build GRCh38
rs749912261 Chr.9: 108943772-108943772 on Build GRCh38
rs956804727 Chr.9: 108943887-108943887 on Build GRCh38
rs750436951 Chr.9: 108943663-108943663 on Build GRCh38
rs775876992 Chr.9: 108943746-108943746 on Build GRCh38
rs933076364 Chr.9: 108943763-108943763 on Build GRCh38
rs180957286 Chr.9: 108943670-108943670 on Build GRCh38
rs372641856 Chr.9: 108943808-108943808 on Build GRCh38
rs762412532 Chr.9: 108943972-108943972 on Build GRCh38
rs975894282 Chr.9: 108943912-108943912 on Build GRCh38
rs777963085 Chr.9: 108943797-108943797 on Build GRCh38
rs548302043 Chr.9: 108943853-108943853 on Build GRCh38
rs562577268 Chr.9: 108943702-108943702 on Build GRCh38
rs772470823 Chr.9: 108943728-108943728 on Build GRCh38
rs769299785 Chr.9: 108944047-108944047 on Build GRCh38
rs778981030 Chr.9: 108943659-108943659 on Build GRCh38
rs184673433 Chr.9: 108943796-108943796 on Build GRCh38
rs374610805 Chr.9: 108944041-108944041 on Build GRCh38
rs751341151 Chr.9: 108943791-108943791 on Build GRCh38
rs761160038 Chr.9: 108943751-108943751 on Build GRCh38
rs754763124 Chr.9: 108943994-108943994 on Build GRCh38
rs751557242 Chr.9: 108943983-108943983 on Build GRCh38
rs746320248 Chr.9: 108943721-108943721 on Build GRCh38
rs375950356 Chr.9: 108943976-108943976 on Build GRCh38
rs774716122 Chr.9: 108943708-108943708 on Build GRCh38
rs370231485 Chr.9: 108943859-108943859 on Build GRCh38
rs923137690 Chr.9: 108943932-108943932 on Build GRCh38
rs757454468 Chr.9: 108944017-108944017 on Build GRCh38
rs746799511 Chr.9: 108943675-108943675 on Build GRCh38
rs904001329 Chr.9: 108943870-108943870 on Build GRCh38
rs772737081 Chr.9: 108943969-108943969 on Build GRCh38
rs747449893 Chr.9: 108944043-108944043 on Build GRCh38
rs767237003 Chr.9: 108944008-108944008 on Build GRCh38
rs867899058 Chr.9: 108943747-108943747 on Build GRCh38
rs772027723 Chr.9: 108943914-108943914 on Build GRCh38
rs940571000 Chr.9: 108943693-108943693 on Build GRCh38
rs867477994 Chr.9: 108944002-108944002 on Build GRCh38
rs778012892 Chr.9: 108943676-108943676 on Build GRCh38
rs544431140 Chr.9: 108943690-108943690 on Build GRCh38
rs758303330 Chr.9: 108943773-108943773 on Build GRCh38
rs373556295 Chr.9: 108943652-108943652 on Build GRCh38
rs766320045 Chr.9: 108943977-108943977 on Build GRCh38
rs576450579 Chr.9: 108943918-108943918 on Build GRCh38
rs764818762 Chr.9: 108943948-108943948 on Build GRCh38
rs776483894 Chr.9: 108943759-108943759 on Build GRCh38
rs758269751 Chr.9: 108943668-108943668 on Build GRCh38
rs372722408 Chr.9: 108943849-108943849 on Build GRCh38
rs761499232 Chr.9: 108943930-108943930 on Build GRCh38
rs374521418 Chr.9: 108944022-108944022 on Build GRCh38
rs941700867 Chr.9: 108943618-108943618 on Build GRCh38
rs778996913 Chr.9: 108943839-108943839 on Build GRCh38
rs780120157 Chr.9: 108943784-108943784 on Build GRCh38
rs909126348 Chr.9: 108943987-108943987 on Build GRCh38
rs763779451 Chr.9: 108943656-108943656 on Build GRCh38
rs984710867 Chr.9: 108943646-108943646 on Build GRCh38
rs566502080 Chr.9: 108944041-108944041 on Build GRCh38
rs866167967 Chr.9: 108943807-108943807 on Build GRCh38
rs141504242 Chr.9: 108943782-108943782 on Build GRCh38
rs376478832 Chr.9: 108944031-108944031 on Build GRCh38
rs867581007 Chr.9: 108943872-108943872 on Build GRCh38

View Full Product Details