Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69336021-69336522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A2
Entrez Gene ID
9413
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C9orf61, X123
UniGene
Hs.118003
Gene Chromosome Location
Chr.9: 69324572-69392461 on Build GRCh38
Transcripts
NM_001127608.1 NM_004816.3 XM_005252307.4 XM_017015324.1 XM_017015325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6318349 c.12G>T p.L4L Chr.9: 69336269-69336269 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs967653797 Chr.9: 69336416-69336416 on Build GRCh38
rs774795206 Chr.9: 69336303-69336303 on Build GRCh38
rs781081118 Chr.9: 69336261-69336261 on Build GRCh38
rs766703134 Chr.9: 69336249-69336249 on Build GRCh38
rs977746712 Chr.9: 69336424-69336424 on Build GRCh38
rs375246573 Chr.9: 69336272-69336272 on Build GRCh38
rs1032759909 Chr.9: 69336415-69336415 on Build GRCh38
rs923668125 Chr.9: 69336443-69336443 on Build GRCh38
rs773540467 Chr.9: 69336488-69336488 on Build GRCh38
rs374786319 Chr.9: 69336298-69336298 on Build GRCh38
rs756579062 Chr.9: 69336292-69336292 on Build GRCh38
rs1026371490 Chr.9: 69336212-69336212 on Build GRCh38
rs560522220 Chr.9: 69336187-69336187 on Build GRCh38
rs964438866 Chr.9: 69336387-69336387 on Build GRCh38
rs374449715 Chr.9: 69336208-69336208 on Build GRCh38
rs945589517 Chr.9: 69336469-69336469 on Build GRCh38
rs746526715 Chr.9: 69336139-69336139 on Build GRCh38
rs1022170402 Chr.9: 69336375-69336375 on Build GRCh38
rs140737122 Chr.9: 69336043-69336043 on Build GRCh38
rs933785164 Chr.9: 69336463-69336463 on Build GRCh38
rs369539393 Chr.9: 69336289-69336289 on Build GRCh38
rs768002526 Chr.9: 69336295-69336295 on Build GRCh38
rs990178838 Chr.9: 69336109-69336109 on Build GRCh38
rs982290931 Chr.9: 69336478-69336478 on Build GRCh38
rs533462148 Chr.9: 69336114-69336114 on Build GRCh38
rs936942179 Chr.9: 69336108-69336108 on Build GRCh38
rs974425725 Chr.9: 69336410-69336415 on Build GRCh38
rs116026385 Chr.9: 69336219-69336219 on Build GRCh38
rs749634708 Chr.9: 69336299-69336299 on Build GRCh38
rs751665125 Chr.9: 69336253-69336253 on Build GRCh38
rs1042081658 Chr.9: 69336048-69336048 on Build GRCh38
rs567570788 Chr.9: 69336317-69336317 on Build GRCh38
rs545604993 Chr.9: 69336176-69336176 on Build GRCh38
rs779612815 Chr.9: 69336159-69336162 on Build GRCh38
rs549285630 Chr.9: 69336220-69336220 on Build GRCh38
rs373172464 Chr.9: 69336241-69336241 on Build GRCh38
rs934800813 Chr.9: 69336062-69336062 on Build GRCh38
rs938156777 Chr.9: 69336490-69336490 on Build GRCh38
rs142255232 Chr.9: 69336129-69336129 on Build GRCh38
rs1018703365 Chr.9: 69336357-69336357 on Build GRCh38
rs1008702117 Chr.9: 69336301-69336301 on Build GRCh38
rs762214776 Chr.9: 69336284-69336285 on Build GRCh38
rs750400713 Chr.9: 69336333-69336333 on Build GRCh38
rs1007850437 Chr.9: 69336137-69336137 on Build GRCh38
rs746596964 Chr.9: 69336318-69336318 on Build GRCh38
rs866355457 Chr.9: 69336080-69336080 on Build GRCh38
rs755239749 Chr.9: 69336258-69336258 on Build GRCh38
rs760384341 Chr.9: 69336210-69336210 on Build GRCh38
rs372038515 Chr.9: 69336234-69336234 on Build GRCh38
rs758326626 Chr.9: 69336244-69336244 on Build GRCh38
rs189174562 Chr.9: 69336300-69336300 on Build GRCh38
rs538057755 Chr.9: 69336337-69336337 on Build GRCh38
rs376768911 Chr.9: 69336428-69336431 on Build GRCh38
rs923727726 Chr.9: 69336060-69336060 on Build GRCh38
rs768111745 Chr.9: 69336140-69336140 on Build GRCh38
rs765308667 Chr.9: 69336235-69336235 on Build GRCh38

View Full Product Details