Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50080516-50081019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN5
Entrez Gene ID
1184
Gene Name
chloride voltage-gated channel 5
Gene Aliases
CLC5, CLCK2, ClC-5, DENTS, NPHL1, NPHL2, XLRH, XRN, hCIC-K2
UniGene
Hs.166486
Gene Chromosome Location
Chr.X: 49922615-50099235 on Build GRCh38
Transcripts
NM_000084.4 NM_001127898.3 NM_001127899.3 NM_001282163.1 XM_017029257.1 XM_017029258.1

SNP Detail

SNP ID Chromosome Location
rs376001902 Chr.X: 50080607-50080607 on Build GRCh38
rs782304700 Chr.X: 50080987-50080987 on Build GRCh38
rs186590815 Chr.X: 50080973-50080973 on Build GRCh38
rs973397689 Chr.X: 50080863-50080863 on Build GRCh38
rs782582465 Chr.X: 50080849-50080849 on Build GRCh38
rs782014828 Chr.X: 50080609-50080609 on Build GRCh38
rs782280603 Chr.X: 50080739-50080739 on Build GRCh38
rs781799509 Chr.X: 50080646-50080646 on Build GRCh38
rs782084593 Chr.X: 50080629-50080629 on Build GRCh38
rs782584473 Chr.X: 50080698-50080698 on Build GRCh38
rs782050861 Chr.X: 50080605-50080605 on Build GRCh38
rs782286395 Chr.X: 50080941-50080941 on Build GRCh38
rs181775862 Chr.X: 50080905-50080905 on Build GRCh38
rs781909097 Chr.X: 50080667-50080667 on Build GRCh38
rs374421323 Chr.X: 50080744-50080744 on Build GRCh38
rs982326037 Chr.X: 50080907-50080907 on Build GRCh38
rs782217593 Chr.X: 50080672-50080672 on Build GRCh38
rs781973587 Chr.X: 50080766-50080766 on Build GRCh38
rs782620034 Chr.X: 50080549-50080549 on Build GRCh38
rs1029626741 Chr.X: 50080625-50080625 on Build GRCh38
rs34800648 Chr.X: 50080626-50080626 on Build GRCh38
rs782431747 Chr.X: 50080741-50080741 on Build GRCh38
rs140312372 Chr.X: 50080686-50080686 on Build GRCh38
rs782322018 Chr.X: 50080608-50080608 on Build GRCh38
rs376106973 Chr.X: 50080758-50080758 on Build GRCh38
rs368674238 Chr.X: 50080679-50080679 on Build GRCh38
rs781959302 Chr.X: 50080586-50080586 on Build GRCh38
rs782463692 Chr.X: 50080662-50080662 on Build GRCh38
rs782335406 Chr.X: 50080575-50080575 on Build GRCh38
rs142498911 Chr.X: 50080624-50080624 on Build GRCh38
rs782306046 Chr.X: 50080695-50080695 on Build GRCh38
rs782258497 Chr.X: 50080560-50080560 on Build GRCh38
rs782288269 Chr.X: 50080882-50080882 on Build GRCh38
rs200390605 Chr.X: 50080663-50080663 on Build GRCh38
rs782605316 Chr.X: 50080685-50080685 on Build GRCh38
rs782746421 Chr.X: 50080630-50080630 on Build GRCh38
rs782261029 Chr.X: 50080752-50080752 on Build GRCh38
rs782415635 Chr.X: 50080900-50080900 on Build GRCh38

View Full Product Details