Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70041690-70042191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AWAT2
Entrez Gene ID
158835
Gene Name
acyl-CoA wax alcohol acyltransferase 2
Gene Aliases
ARAT, DC4, DGAT2L4, MFAT, WS
UniGene
Hs.661507
Gene Chromosome Location
Chr.X: 70040542-70049970 on Build GRCh38
Transcripts
NM_001002254.1 XM_011530876.2

SNP Detail

SNP ID Chromosome Location
rs41310609 Chr.X: 70042005-70042005 on Build GRCh38
rs756874028 Chr.X: 70041832-70041832 on Build GRCh38
rs780959816 Chr.X: 70041839-70041839 on Build GRCh38
rs192369503 Chr.X: 70041747-70041747 on Build GRCh38
rs756822595 Chr.X: 70042160-70042160 on Build GRCh38
rs1051164678 Chr.X: 70041783-70041783 on Build GRCh38
rs935064811 Chr.X: 70041750-70041750 on Build GRCh38
rs369460902 Chr.X: 70041763-70041763 on Build GRCh38
rs41312130 Chr.X: 70042046-70042046 on Build GRCh38
rs757205004 Chr.X: 70041845-70041845 on Build GRCh38
rs749685250 Chr.X: 70041880-70041880 on Build GRCh38
rs867514850 Chr.X: 70041857-70041857 on Build GRCh38
rs754113344 Chr.X: 70041990-70041990 on Build GRCh38
rs755375421 Chr.X: 70041800-70041800 on Build GRCh38
rs902301847 Chr.X: 70041725-70041725 on Build GRCh38
rs935958994 Chr.X: 70042006-70042006 on Build GRCh38
rs753721903 Chr.X: 70041805-70041806 on Build GRCh38
rs746470905 Chr.X: 70041829-70041829 on Build GRCh38
rs753296093 Chr.X: 70042137-70042137 on Build GRCh38
rs769617781 Chr.X: 70041876-70041876 on Build GRCh38
rs771302360 Chr.X: 70041896-70041896 on Build GRCh38
rs779507862 Chr.X: 70041803-70041803 on Build GRCh38
rs755394818 Chr.X: 70041996-70041996 on Build GRCh38
rs1053918966 Chr.X: 70041756-70041756 on Build GRCh38
rs200995684 Chr.X: 70041891-70041891 on Build GRCh38
rs369771963 Chr.X: 70041987-70041987 on Build GRCh38
rs1053147728 Chr.X: 70042025-70042025 on Build GRCh38
rs866925711 Chr.X: 70042053-70042053 on Build GRCh38
rs1040785583 Chr.X: 70041720-70041720 on Build GRCh38
rs187693487 Chr.X: 70041719-70041719 on Build GRCh38
rs756470607 Chr.X: 70041770-70041770 on Build GRCh38
rs767707063 Chr.X: 70041765-70041765 on Build GRCh38
rs768210433 Chr.X: 70042034-70042034 on Build GRCh38
rs745582657 Chr.X: 70041865-70041865 on Build GRCh38
rs780915226 Chr.X: 70041960-70041960 on Build GRCh38
rs2281933 Chr.X: 70041968-70041968 on Build GRCh38
rs750687942 Chr.X: 70041768-70041768 on Build GRCh38
rs766716184 Chr.X: 70041772-70041772 on Build GRCh38
rs201120184 Chr.X: 70042054-70042054 on Build GRCh38
rs754243709 Chr.X: 70041799-70041799 on Build GRCh38
rs1023849414 Chr.X: 70041882-70041882 on Build GRCh38
rs760829389 Chr.X: 70041980-70041980 on Build GRCh38
rs774323186 Chr.X: 70041929-70041929 on Build GRCh38
rs976569578 Chr.X: 70041804-70041804 on Build GRCh38
rs767653991 Chr.X: 70041955-70041955 on Build GRCh38
rs761853431 Chr.X: 70041936-70041936 on Build GRCh38

View Full Product Details