Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231040026-231040531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM89A
Entrez Gene ID
375061
Gene Name
family with sequence similarity 89 member A
Gene Aliases
C1orf153
UniGene
Hs.38516
Gene Chromosome Location
Chr.1: 231018958-231040249 on Build GRCh38
Transcripts
NM_198552.2

SNP Detail

SNP ID Chromosome Location
rs1037676653 Chr.1: 231040214-231040214 on Build GRCh38
rs868367716 Chr.1: 231040345-231040345 on Build GRCh38
rs926036562 Chr.1: 231040357-231040363 on Build GRCh38
rs973486864 Chr.1: 231040340-231040340 on Build GRCh38
rs968359760 Chr.1: 231040264-231040264 on Build GRCh38
rs768536088 Chr.1: 231040154-231040154 on Build GRCh38
rs1040877430 Chr.1: 231040244-231040244 on Build GRCh38
rs1009836318 Chr.1: 231040146-231040146 on Build GRCh38
rs1015120004 Chr.1: 231040308-231040308 on Build GRCh38
rs536705796 Chr.1: 231040339-231040339 on Build GRCh38
rs931205864 Chr.1: 231040224-231040224 on Build GRCh38
rs775562202 Chr.1: 231040065-231040065 on Build GRCh38
rs766589274 Chr.1: 231040095-231040095 on Build GRCh38
rs571948912 Chr.1: 231040135-231040135 on Build GRCh38
rs547963369 Chr.1: 231040277-231040277 on Build GRCh38
rs993437890 Chr.1: 231040462-231040462 on Build GRCh38
rs868582324 Chr.1: 231040102-231040102 on Build GRCh38
rs765479760 Chr.1: 231040087-231040087 on Build GRCh38
rs375041957 Chr.1: 231040319-231040319 on Build GRCh38
rs898398332 Chr.1: 231040079-231040079 on Build GRCh38
rs776515561 Chr.1: 231040166-231040166 on Build GRCh38
rs748848078 Chr.1: 231040124-231040124 on Build GRCh38
rs1001595617 Chr.1: 231040402-231040402 on Build GRCh38
rs960349203 Chr.1: 231040296-231040296 on Build GRCh38
rs891640100 Chr.1: 231040384-231040384 on Build GRCh38
rs777880088 Chr.1: 231040135-231040135 on Build GRCh38
rs1008101084 Chr.1: 231040118-231040118 on Build GRCh38
rs762049785 Chr.1: 231040077-231040077 on Build GRCh38
rs745500241 Chr.1: 231040136-231040136 on Build GRCh38
rs931967085 Chr.1: 231040349-231040349 on Build GRCh38
rs1021214192 Chr.1: 231040259-231040259 on Build GRCh38
rs776642566 Chr.1: 231040076-231040076 on Build GRCh38
rs557655387 Chr.1: 231040440-231040440 on Build GRCh38
rs896034345 Chr.1: 231040460-231040460 on Build GRCh38
rs760461220 Chr.1: 231040069-231040069 on Build GRCh38
rs552138002 Chr.1: 231040100-231040100 on Build GRCh38
rs538629968 Chr.1: 231040407-231040407 on Build GRCh38
rs761927073 Chr.1: 231040173-231040173 on Build GRCh38
rs995435993 Chr.1: 231040088-231040088 on Build GRCh38
rs968915711 Chr.1: 231040158-231040158 on Build GRCh38
rs889741281 Chr.1: 231040241-231040241 on Build GRCh38
rs567666700 Chr.1: 231040303-231040303 on Build GRCh38
rs750585922 Chr.1: 231040092-231040092 on Build GRCh38
rs777461342 Chr.1: 231040105-231040105 on Build GRCh38
rs973601356 Chr.1: 231040493-231040494 on Build GRCh38
rs962086820 Chr.1: 231040318-231040319 on Build GRCh38
rs556307245 Chr.1: 231040354-231040354 on Build GRCh38
rs1015059692 Chr.1: 231040477-231040478 on Build GRCh38
rs960483853 Chr.1: 231040192-231040192 on Build GRCh38
rs769848650 Chr.1: 231040184-231040184 on Build GRCh38
rs866545541 Chr.1: 231040075-231040075 on Build GRCh38
rs940779647 Chr.1: 231040202-231040202 on Build GRCh38
rs891520769 Chr.1: 231040251-231040251 on Build GRCh38
rs1008463542 Chr.1: 231040242-231040242 on Build GRCh38
rs940666764 Chr.1: 231040055-231040055 on Build GRCh38
rs773884421 Chr.1: 231040507-231040507 on Build GRCh38
rs532061224 Chr.1: 231040090-231040090 on Build GRCh38
rs1035007360 Chr.1: 231040284-231040284 on Build GRCh38
rs943985247 Chr.1: 231040358-231040358 on Build GRCh38
rs1037734265 Chr.1: 231040062-231040062 on Build GRCh38
rs993387737 Chr.1: 231040297-231040297 on Build GRCh38
rs758853714 Chr.1: 231040093-231040093 on Build GRCh38
rs775088122 Chr.1: 231040140-231040140 on Build GRCh38
rs1021261788 Chr.1: 231040155-231040155 on Build GRCh38
rs980772507 Chr.1: 231040163-231040163 on Build GRCh38
rs1014515636 Chr.1: 231040461-231040461 on Build GRCh38
rs771903868 Chr.1: 231040139-231040139 on Build GRCh38
rs569678958 Chr.1: 231040386-231040386 on Build GRCh38
rs1050104580 Chr.1: 231040228-231040228 on Build GRCh38
rs867368647 Chr.1: 231040323-231040323 on Build GRCh38
rs746989013 Chr.1: 231040145-231040145 on Build GRCh38
rs756806421 Chr.1: 231040125-231040125 on Build GRCh38
rs577511007 Chr.1: 231040480-231040480 on Build GRCh38
rs1009957065 Chr.1: 231040255-231040255 on Build GRCh38
rs907947135 Chr.1: 231040200-231040200 on Build GRCh38
rs920715830 Chr.1: 231040344-231040344 on Build GRCh38
rs562456714 Chr.1: 231040089-231040089 on Build GRCh38
rs752124042 Chr.1: 231040098-231040098 on Build GRCh38
rs955816746 Chr.1: 231040143-231040143 on Build GRCh38
rs1027725459 Chr.1: 231040500-231040500 on Build GRCh38
rs867214222 Chr.1: 231040064-231040064 on Build GRCh38
rs920601549 Chr.1: 231040219-231040219 on Build GRCh38
rs945762447 Chr.1: 231040389-231040389 on Build GRCh38
rs755496867 Chr.1: 231040101-231040101 on Build GRCh38
rs1030378092 Chr.1: 231040464-231040464 on Build GRCh38
rs1040825076 Chr.1: 231040369-231040369 on Build GRCh38
rs867505392 Chr.1: 231040071-231040071 on Build GRCh38
rs764171929 Chr.1: 231040070-231040070 on Build GRCh38
rs560610357 Chr.1: 231040275-231040275 on Build GRCh38
rs1042788311 Chr.1: 231040394-231040394 on Build GRCh38

View Full Product Details