Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65884778-65885269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIBP
Entrez Gene ID
9158
Gene Name
FGF1 intracellular binding protein
Gene Aliases
FGFIBP, FIBP-1
UniGene
Hs.7768
Gene Chromosome Location
Chr.11: 65883740-65888539 on Build GRCh38
Transcripts
NM_004214.4 NM_198897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3451917 c.715C>T p.Q239* Chr.11: 65885139-65885139 on Build GRCh38
5886720 c.777C>T p.S259S Chr.11: 65884998-65884998 on Build GRCh38
4943484 c.825G>A p.M275I Chr.11: 65884950-65884950 on Build GRCh38
1745139 c.669_676delCTCACAGA p.S224fs*2 Chr.11: 65885178-65885185 on Build GRCh38
325353 c.706G>T p.E236* Chr.11: 65885148-65885148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771514398 Chr.11: 65885236-65885236 on Build GRCh38
rs1028545902 Chr.11: 65885204-65885204 on Build GRCh38
rs1055790644 Chr.11: 65885064-65885064 on Build GRCh38
rs773967562 Chr.11: 65884850-65884850 on Build GRCh38
rs530389246 Chr.11: 65885134-65885134 on Build GRCh38
rs762228673 Chr.11: 65884980-65884981 on Build GRCh38
rs374597428 Chr.11: 65884886-65884886 on Build GRCh38
rs1031596283 Chr.11: 65884997-65884997 on Build GRCh38
rs775003757 Chr.11: 65884959-65884959 on Build GRCh38
rs373866174 Chr.11: 65885231-65885231 on Build GRCh38
rs749557354 Chr.11: 65884995-65884995 on Build GRCh38
rs543944553 Chr.11: 65884863-65884863 on Build GRCh38
rs377135404 Chr.11: 65885199-65885199 on Build GRCh38
rs373942573 Chr.11: 65885030-65885030 on Build GRCh38
rs912150483 Chr.11: 65885083-65885083 on Build GRCh38
rs944695642 Chr.11: 65884883-65884883 on Build GRCh38
rs1022817477 Chr.11: 65885233-65885233 on Build GRCh38
rs761375102 Chr.11: 65885044-65885044 on Build GRCh38
rs200458781 Chr.11: 65884892-65884892 on Build GRCh38
rs766790271 Chr.11: 65885058-65885058 on Build GRCh38
rs756394663 Chr.11: 65885053-65885053 on Build GRCh38
rs959910087 Chr.11: 65885005-65885005 on Build GRCh38
rs762334947 Chr.11: 65884920-65884920 on Build GRCh38
rs983989564 Chr.11: 65885066-65885066 on Build GRCh38
rs749090777 Chr.11: 65885212-65885212 on Build GRCh38
rs786204849 Chr.11: 65885181-65885181 on Build GRCh38
rs745631961 Chr.11: 65885019-65885019 on Build GRCh38
rs761261038 Chr.11: 65885194-65885194 on Build GRCh38
rs534999269 Chr.11: 65884821-65884821 on Build GRCh38
rs371052475 Chr.11: 65885158-65885158 on Build GRCh38
rs188776710 Chr.11: 65884909-65884909 on Build GRCh38
rs756076684 Chr.11: 65885208-65885208 on Build GRCh38
rs981922791 Chr.11: 65885221-65885221 on Build GRCh38
rs750468273 Chr.11: 65884876-65884876 on Build GRCh38
rs772985154 Chr.11: 65884884-65884884 on Build GRCh38
rs778497312 Chr.11: 65885214-65885214 on Build GRCh38
rs748224586 Chr.11: 65885070-65885070 on Build GRCh38
rs938655466 Chr.11: 65884855-65884855 on Build GRCh38
rs558864995 Chr.11: 65884804-65884804 on Build GRCh38
rs768379682 Chr.11: 65885213-65885213 on Build GRCh38
rs781240271 Chr.11: 65885109-65885109 on Build GRCh38
rs141444067 Chr.11: 65885206-65885206 on Build GRCh38
rs915664010 Chr.11: 65884838-65884838 on Build GRCh38
rs996467689 Chr.11: 65884916-65884916 on Build GRCh38
rs761285384 Chr.11: 65885166-65885166 on Build GRCh38
rs2231891 Chr.11: 65885065-65885065 on Build GRCh38
rs775253016 Chr.11: 65884918-65884918 on Build GRCh38
rs771602328 Chr.11: 65885178-65885178 on Build GRCh38
rs1039011992 Chr.11: 65884888-65884888 on Build GRCh38
rs754111636 Chr.11: 65885213-65885214 on Build GRCh38
rs371059977 Chr.11: 65884911-65884911 on Build GRCh38
rs755231302 Chr.11: 65885061-65885061 on Build GRCh38
rs150145567 Chr.11: 65884887-65884887 on Build GRCh38
rs999067369 Chr.11: 65884969-65884969 on Build GRCh38
rs944977861 Chr.11: 65885084-65885084 on Build GRCh38
rs773468043 Chr.11: 65885074-65885074 on Build GRCh38
rs768025186 Chr.11: 65885032-65885058 on Build GRCh38
rs768081747 Chr.11: 65884972-65884972 on Build GRCh38
rs902562855 Chr.11: 65885163-65885163 on Build GRCh38
rs758599897 Chr.11: 65885094-65885094 on Build GRCh38
rs773661243 Chr.11: 65885186-65885186 on Build GRCh38
rs536284942 Chr.11: 65885220-65885220 on Build GRCh38
rs35265924 Chr.11: 65885113-65885113 on Build GRCh38
rs989701827 Chr.11: 65884807-65884807 on Build GRCh38
rs753951250 Chr.11: 65885059-65885059 on Build GRCh38
rs931322560 Chr.11: 65885085-65885085 on Build GRCh38
rs756523812 Chr.11: 65884977-65884977 on Build GRCh38
rs574968598 Chr.11: 65884887-65884887 on Build GRCh38
rs371679267 Chr.11: 65885006-65885006 on Build GRCh38
rs572735420 Chr.11: 65884815-65884815 on Build GRCh38
rs368306000 Chr.11: 65884979-65884979 on Build GRCh38
rs940196715 Chr.11: 65885141-65885141 on Build GRCh38
rs559273110 Chr.11: 65884938-65884938 on Build GRCh38
rs769666927 Chr.11: 65885022-65885022 on Build GRCh38
rs757768112 Chr.11: 65884965-65884965 on Build GRCh38
rs1053402237 Chr.11: 65885182-65885182 on Build GRCh38
rs528431626 Chr.11: 65885048-65885048 on Build GRCh38
rs375313477 Chr.11: 65884923-65884923 on Build GRCh38
rs780108813 Chr.11: 65884978-65884978 on Build GRCh38
rs141182394 Chr.11: 65885140-65885140 on Build GRCh38
rs747822282 Chr.11: 65885225-65885225 on Build GRCh38
rs554997037 Chr.11: 65884822-65884822 on Build GRCh38
rs959203362 Chr.11: 65885244-65885244 on Build GRCh38
rs966531260 Chr.11: 65885003-65885003 on Build GRCh38
rs750970017 Chr.11: 65885052-65885052 on Build GRCh38
rs895646526 Chr.11: 65885191-65885191 on Build GRCh38
rs146124046 Chr.11: 65885165-65885165 on Build GRCh38
rs766583455 Chr.11: 65885198-65885198 on Build GRCh38
rs751959829 Chr.11: 65884964-65884964 on Build GRCh38
rs746875538 Chr.11: 65885018-65885018 on Build GRCh38
rs921006817 Chr.11: 65884799-65884799 on Build GRCh38
rs759874580 Chr.11: 65885201-65885201 on Build GRCh38
rs779062281 Chr.11: 65885016-65885016 on Build GRCh38
rs995630046 Chr.11: 65884872-65884872 on Build GRCh38
rs766752054 Chr.11: 65885207-65885208 on Build GRCh38
rs761102835 Chr.11: 65885151-65885162 on Build GRCh38
rs927872693 Chr.11: 65884849-65884849 on Build GRCh38
rs550280360 Chr.11: 65885203-65885203 on Build GRCh38
rs774963051 Chr.11: 65885024-65885024 on Build GRCh38
rs929444881 Chr.11: 65884871-65884871 on Build GRCh38
rs969820406 Chr.11: 65884842-65884842 on Build GRCh38
rs574840533 Chr.11: 65884836-65884836 on Build GRCh38
rs1011027032 Chr.11: 65884893-65884893 on Build GRCh38
rs778018511 Chr.11: 65885207-65885207 on Build GRCh38
rs912100203 Chr.11: 65884877-65884877 on Build GRCh38
rs1046324269 Chr.11: 65884841-65884841 on Build GRCh38
rs768353666 Chr.11: 65884810-65884810 on Build GRCh38
rs540374414 Chr.11: 65884889-65884891 on Build GRCh38
rs539134005 Chr.11: 65885238-65885238 on Build GRCh38

View Full Product Details