Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65890283-65890781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85B
Entrez Gene ID
11007
Gene Name
coiled-coil domain containing 85B
Gene Aliases
DIPA
UniGene
Hs.66713
Gene Chromosome Location
Chr.11: 65890404-65891635 on Build GRCh38
Transcripts
NM_006848.2

SNP Detail

SNP ID Chromosome Location
rs886952836 Chr.11: 65890641-65890641 on Build GRCh38
rs752245770 Chr.11: 65890506-65890506 on Build GRCh38
rs1021273110 Chr.11: 65890669-65890669 on Build GRCh38
rs916109729 Chr.11: 65890560-65890560 on Build GRCh38
rs1019527950 Chr.11: 65890656-65890656 on Build GRCh38
rs1003816107 Chr.11: 65890642-65890642 on Build GRCh38
rs996977989 Chr.11: 65890667-65890667 on Build GRCh38
rs757957589 Chr.11: 65890476-65890476 on Build GRCh38
rs1003866930 Chr.11: 65890522-65890523 on Build GRCh38
rs1056393249 Chr.11: 65890481-65890481 on Build GRCh38
rs936601892 Chr.11: 65890404-65890404 on Build GRCh38
rs1031349670 Chr.11: 65890694-65890694 on Build GRCh38
rs1048924 Chr.11: 65890686-65890686 on Build GRCh38
rs552142507 Chr.11: 65890677-65890677 on Build GRCh38
rs752550577 Chr.11: 65890463-65890463 on Build GRCh38
rs938734313 Chr.11: 65890621-65890621 on Build GRCh38
rs965234229 Chr.11: 65890326-65890326 on Build GRCh38
rs763638034 Chr.11: 65890560-65890560 on Build GRCh38
rs532483669 Chr.11: 65890608-65890608 on Build GRCh38
rs563464728 Chr.11: 65890565-65890565 on Build GRCh38
rs968335527 Chr.11: 65890674-65890674 on Build GRCh38
rs928721095 Chr.11: 65890609-65890609 on Build GRCh38
rs561342117 Chr.11: 65890523-65890523 on Build GRCh38
rs996768515 Chr.11: 65890666-65890666 on Build GRCh38
rs916075159 Chr.11: 65890701-65890701 on Build GRCh38
rs938823993 Chr.11: 65890553-65890553 on Build GRCh38
rs993444993 Chr.11: 65890699-65890699 on Build GRCh38
rs1020882554 Chr.11: 65890679-65890679 on Build GRCh38
rs1037793535 Chr.11: 65890343-65890343 on Build GRCh38
rs960237952 Chr.11: 65890695-65890695 on Build GRCh38
rs949019356 Chr.11: 65890705-65890705 on Build GRCh38
rs993576261 Chr.11: 65890378-65890378 on Build GRCh38
rs989411464 Chr.11: 65890427-65890427 on Build GRCh38
rs1047817329 Chr.11: 65890614-65890614 on Build GRCh38
rs1048567986 Chr.11: 65890744-65890744 on Build GRCh38
rs908786091 Chr.11: 65890759-65890759 on Build GRCh38
rs899290240 Chr.11: 65890349-65890349 on Build GRCh38
rs112280459 Chr.11: 65890505-65890505 on Build GRCh38
rs543678586 Chr.11: 65890555-65890555 on Build GRCh38
rs528045467 Chr.11: 65890749-65890749 on Build GRCh38
rs975691429 Chr.11: 65890339-65890339 on Build GRCh38
rs534196882 Chr.11: 65890468-65890468 on Build GRCh38
rs991947608 Chr.11: 65890707-65890707 on Build GRCh38
rs373241568 Chr.11: 65890492-65890492 on Build GRCh38
rs1041791026 Chr.11: 65890456-65890456 on Build GRCh38
rs946704260 Chr.11: 65890622-65890639 on Build GRCh38
rs371368406 Chr.11: 65890625-65890625 on Build GRCh38
rs975443408 Chr.11: 65890496-65890496 on Build GRCh38
rs968473342 Chr.11: 65890512-65890512 on Build GRCh38
rs899786827 Chr.11: 65890664-65890664 on Build GRCh38
rs1008161852 Chr.11: 65890469-65890469 on Build GRCh38
rs11544733 Chr.11: 65890734-65890734 on Build GRCh38
rs886924962 Chr.11: 65890458-65890458 on Build GRCh38
rs938862801 Chr.11: 65890471-65890471 on Build GRCh38
rs558912774 Chr.11: 65890485-65890485 on Build GRCh38
rs913829302 Chr.11: 65890606-65890617 on Build GRCh38
rs900850001 Chr.11: 65890655-65890655 on Build GRCh38
rs973664159 Chr.11: 65890718-65890718 on Build GRCh38
rs931478296 Chr.11: 65890731-65890731 on Build GRCh38
rs920614579 Chr.11: 65890722-65890722 on Build GRCh38
rs931488243 Chr.11: 65890640-65890640 on Build GRCh38
rs1038143581 Chr.11: 65890564-65890564 on Build GRCh38
rs916024466 Chr.11: 65890639-65890639 on Build GRCh38
rs948998506 Chr.11: 65890562-65890562 on Build GRCh38
rs2276318 Chr.11: 65890442-65890442 on Build GRCh38
rs764110100 Chr.11: 65890607-65890608 on Build GRCh38
rs547590136 Chr.11: 65890504-65890504 on Build GRCh38
rs929401118 Chr.11: 65890606-65890606 on Build GRCh38
rs1019919895 Chr.11: 65890480-65890480 on Build GRCh38
rs977126657 Chr.11: 65890532-65890532 on Build GRCh38
rs993103635 Chr.11: 65890559-65890559 on Build GRCh38
rs146161054 Chr.11: 65890533-65890533 on Build GRCh38
rs966643299 Chr.11: 65890683-65890683 on Build GRCh38
rs946034771 Chr.11: 65890455-65890455 on Build GRCh38
rs576711490 Chr.11: 65890306-65890306 on Build GRCh38
rs921173304 Chr.11: 65890363-65890363 on Build GRCh38
rs1013298930 Chr.11: 65890321-65890321 on Build GRCh38
rs1008440374 Chr.11: 65890647-65890647 on Build GRCh38

View Full Product Details