Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67418353-67418856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARNS1
Entrez Gene ID
57571
Gene Name
carnosine synthase 1
Gene Aliases
ATPGD1
UniGene
Hs.502982
Gene Chromosome Location
Chr.11: 67415678-67425607 on Build GRCh38
Transcripts
NM_001166222.1 NM_020811.1 XM_011545189.2 XM_011545191.2 XM_017018054.1 XM_017018055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350607 c.364+9A>G p.? Chr.11: 67418529-67418529 on Build GRCh38
5668751 c.389C>T p.P130L Chr.11: 67418780-67418780 on Build GRCh38
6239327 c.318C>T p.P106P Chr.11: 67418474-67418474 on Build GRCh38
5437762 c.287C>T p.A96V Chr.11: 67418443-67418443 on Build GRCh38
6014531 c.284G>A p.G95E Chr.11: 67418440-67418440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774816159 Chr.11: 67418415-67418415 on Build GRCh38
rs758506028 Chr.11: 67418559-67418559 on Build GRCh38
rs903539800 Chr.11: 67418782-67418782 on Build GRCh38
rs78200432 Chr.11: 67418648-67418648 on Build GRCh38
rs772175734 Chr.11: 67418493-67418493 on Build GRCh38
rs984909596 Chr.11: 67418704-67418704 on Build GRCh38
rs1050540577 Chr.11: 67418755-67418755 on Build GRCh38
rs370093309 Chr.11: 67418470-67418470 on Build GRCh38
rs552741498 Chr.11: 67418694-67418694 on Build GRCh38
rs1024567408 Chr.11: 67418687-67418687 on Build GRCh38
rs61744660 Chr.11: 67418816-67418816 on Build GRCh38
rs901804004 Chr.11: 67418736-67418736 on Build GRCh38
rs772591037 Chr.11: 67418714-67418715 on Build GRCh38
rs1002921544 Chr.11: 67418416-67418416 on Build GRCh38
rs971386444 Chr.11: 67418691-67418691 on Build GRCh38
rs769646669 Chr.11: 67418809-67418809 on Build GRCh38
rs563065105 Chr.11: 67418717-67418717 on Build GRCh38
rs1028693257 Chr.11: 67418621-67418621 on Build GRCh38
rs748134171 Chr.11: 67418554-67418554 on Build GRCh38
rs927408061 Chr.11: 67418518-67418518 on Build GRCh38
rs968232075 Chr.11: 67418632-67418632 on Build GRCh38
rs528470629 Chr.11: 67418460-67418460 on Build GRCh38
rs750192824 Chr.11: 67418779-67418779 on Build GRCh38
rs995799299 Chr.11: 67418612-67418612 on Build GRCh38
rs371219030 Chr.11: 67418749-67418749 on Build GRCh38
rs947294141 Chr.11: 67418683-67418683 on Build GRCh38
rs777803378 Chr.11: 67418808-67418808 on Build GRCh38
rs551107070 Chr.11: 67418583-67418583 on Build GRCh38
rs1033574395 Chr.11: 67418654-67418654 on Build GRCh38
rs746269105 Chr.11: 67418713-67418713 on Build GRCh38
rs886339465 Chr.11: 67418563-67418563 on Build GRCh38
rs577311660 Chr.11: 67418701-67418701 on Build GRCh38
rs747111880 Chr.11: 67418631-67418631 on Build GRCh38
rs549231263 Chr.11: 67418399-67418399 on Build GRCh38
rs766159727 Chr.11: 67418797-67418797 on Build GRCh38
rs956246161 Chr.11: 67418681-67418681 on Build GRCh38
rs747732983 Chr.11: 67418409-67418409 on Build GRCh38
rs903151127 Chr.11: 67418693-67418693 on Build GRCh38
rs1005181542 Chr.11: 67418568-67418568 on Build GRCh38
rs1044460800 Chr.11: 67418498-67418498 on Build GRCh38
rs767415356 Chr.11: 67418521-67418521 on Build GRCh38
rs757424371 Chr.11: 67418812-67418812 on Build GRCh38
rs544657256 Chr.11: 67418702-67418702 on Build GRCh38
rs773990634 Chr.11: 67418714-67418714 on Build GRCh38
rs1035786488 Chr.11: 67418427-67418427 on Build GRCh38
rs868167 Chr.11: 67418800-67418800 on Build GRCh38
rs774084845 Chr.11: 67418388-67418389 on Build GRCh38
rs747921192 Chr.11: 67418395-67418395 on Build GRCh38
rs539328267 Chr.11: 67418570-67418570 on Build GRCh38
rs776961756 Chr.11: 67418710-67418710 on Build GRCh38
rs373347042 Chr.11: 67418443-67418443 on Build GRCh38
rs1790733 Chr.11: 67418529-67418529 on Build GRCh38
rs763685332 Chr.11: 67418751-67418751 on Build GRCh38
rs760547772 Chr.11: 67418780-67418780 on Build GRCh38
rs956515609 Chr.11: 67418459-67418459 on Build GRCh38
rs917225584 Chr.11: 67418463-67418463 on Build GRCh38
rs754924007 Chr.11: 67418806-67418806 on Build GRCh38
rs771670634 Chr.11: 67418712-67418713 on Build GRCh38
rs373811431 Chr.11: 67418474-67418474 on Build GRCh38
rs536862162 Chr.11: 67418603-67418603 on Build GRCh38
rs762314194 Chr.11: 67418719-67418719 on Build GRCh38
rs777910151 Chr.11: 67418562-67418562 on Build GRCh38
rs569429311 Chr.11: 67418602-67418602 on Build GRCh38
rs190065726 Chr.11: 67418686-67418686 on Build GRCh38
rs780060575 Chr.11: 67418550-67418550 on Build GRCh38
rs771187321 Chr.11: 67418706-67418706 on Build GRCh38
rs185221600 Chr.11: 67418435-67418435 on Build GRCh38
rs770163677 Chr.11: 67418716-67418716 on Build GRCh38
rs767325534 Chr.11: 67418771-67418771 on Build GRCh38
rs555213547 Chr.11: 67418641-67418641 on Build GRCh38
rs770209351 Chr.11: 67418820-67418820 on Build GRCh38
rs903202187 Chr.11: 67418394-67418394 on Build GRCh38
rs757029107 Chr.11: 67418501-67418501 on Build GRCh38
rs116762183 Chr.11: 67418574-67418574 on Build GRCh38
rs892988431 Chr.11: 67418673-67418673 on Build GRCh38
rs750282832 Chr.11: 67418526-67418526 on Build GRCh38
rs746154102 Chr.11: 67418817-67418817 on Build GRCh38
rs774072032 Chr.11: 67418753-67418753 on Build GRCh38
rs761525899 Chr.11: 67418754-67418754 on Build GRCh38

View Full Product Details