Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89669616-89670167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA33
Entrez Gene ID
124045
Gene Name
spermatogenesis associated 33
Gene Aliases
C16orf55
UniGene
Hs.655171
Gene Chromosome Location
Chr.16: 89657744-89670458 on Build GRCh38
Transcripts
NM_001271907.1 NM_001271908.1 NM_001271909.1 NM_001271910.1 NM_153025.2 NR_073515.1

SNP Detail

SNP ID Chromosome Location
rs564847348 Chr.16: 89670086-89670086 on Build GRCh38
rs35063026 Chr.16: 89669749-89669749 on Build GRCh38
rs74033828 Chr.16: 89670109-89670109 on Build GRCh38
rs999394932 Chr.16: 89670149-89670149 on Build GRCh38
rs537175883 Chr.16: 89669662-89669662 on Build GRCh38
rs537543334 Chr.16: 89670113-89670113 on Build GRCh38
rs1049396757 Chr.16: 89669971-89669971 on Build GRCh38
rs182901284 Chr.16: 89670070-89670070 on Build GRCh38
rs908040833 Chr.16: 89669673-89669673 on Build GRCh38
rs563345892 Chr.16: 89669942-89669942 on Build GRCh38
rs7197444 Chr.16: 89670112-89670112 on Build GRCh38
rs553391827 Chr.16: 89669802-89669802 on Build GRCh38
rs536220030 Chr.16: 89669682-89669682 on Build GRCh38
rs952121956 Chr.16: 89669665-89669665 on Build GRCh38
rs201580694 Chr.16: 89669815-89669815 on Build GRCh38
rs187093060 Chr.16: 89670134-89670134 on Build GRCh38
rs555716525 Chr.16: 89669667-89669667 on Build GRCh38
rs889497808 Chr.16: 89670053-89670053 on Build GRCh38
rs1657379 Chr.16: 89670103-89670103 on Build GRCh38
rs900614684 Chr.16: 89669789-89669789 on Build GRCh38
rs1043521505 Chr.16: 89670106-89670106 on Build GRCh38
rs1036712064 Chr.16: 89669690-89669690 on Build GRCh38
rs1037785241 Chr.16: 89669689-89669689 on Build GRCh38
rs603156 Chr.16: 89670122-89670122 on Build GRCh38
rs933632550 Chr.16: 89669960-89669960 on Build GRCh38
rs755113719 Chr.16: 89669674-89669674 on Build GRCh38
rs368725031 Chr.16: 89669959-89669959 on Build GRCh38
rs373933615 Chr.16: 89669845-89669845 on Build GRCh38
rs604868 Chr.16: 89669769-89669769 on Build GRCh38
rs1046876 Chr.16: 89669637-89669637 on Build GRCh38
rs953994904 Chr.16: 89669938-89669938 on Build GRCh38
rs2434871 Chr.16: 89669944-89669944 on Build GRCh38
rs896897602 Chr.16: 89669726-89669726 on Build GRCh38
rs546828075 Chr.16: 89670076-89670076 on Build GRCh38
rs778248072 Chr.16: 89669917-89669917 on Build GRCh38
rs931967484 Chr.16: 89670124-89670124 on Build GRCh38
rs976735454 Chr.16: 89669869-89669869 on Build GRCh38
rs560932824 Chr.16: 89669986-89669986 on Build GRCh38
rs535320951 Chr.16: 89669755-89669755 on Build GRCh38
rs549255209 Chr.16: 89670133-89670133 on Build GRCh38
rs78790307 Chr.16: 89669926-89669926 on Build GRCh38
rs779211015 Chr.16: 89670032-89670032 on Build GRCh38
rs545631816 Chr.16: 89669895-89669895 on Build GRCh38
rs532146153 Chr.16: 89670097-89670097 on Build GRCh38
rs11648172 Chr.16: 89669935-89669935 on Build GRCh38
rs2437952 Chr.16: 89669893-89669893 on Build GRCh38
rs746716853 Chr.16: 89670037-89670037 on Build GRCh38
rs756671041 Chr.16: 89669913-89669913 on Build GRCh38
rs946470343 Chr.16: 89670056-89670056 on Build GRCh38

View Full Product Details