Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39760847-39761373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF3
Entrez Gene ID
22806
Gene Name
IKAROS family zinc finger 3
Gene Aliases
AIO, AIOLOS, ZNFN1A3
UniGene
Hs.371680
Gene Chromosome Location
Chr.17: 39757715-39864188 on Build GRCh38
Transcripts
NM_001257408.1 NM_001257409.1 NM_001257410.1 NM_001257411.1 NM_001257412.1 NM_001257413.1 NM_001257414.1 NM_001284514.1 NM_001284515.1 NM_001284516.1 NM_012481.4 NM_183228.2 NM_183229.2 NM_183230.2 NM_183231.2 NM_183232.2

SNP Detail

SNP ID Chromosome Location
rs77905879 Chr.17: 39761114-39761114 on Build GRCh38
rs948635665 Chr.17: 39761239-39761239 on Build GRCh38
rs959737073 Chr.17: 39761338-39761338 on Build GRCh38
rs982130830 Chr.17: 39760960-39760960 on Build GRCh38
rs895757704 Chr.17: 39761238-39761238 on Build GRCh38
rs964486995 Chr.17: 39760893-39760893 on Build GRCh38
rs904307983 Chr.17: 39760939-39760939 on Build GRCh38
rs946982571 Chr.17: 39761079-39761079 on Build GRCh38
rs369606415 Chr.17: 39761195-39761195 on Build GRCh38
rs764455923 Chr.17: 39761192-39761192 on Build GRCh38
rs1035008290 Chr.17: 39761319-39761319 on Build GRCh38
rs999952485 Chr.17: 39761292-39761292 on Build GRCh38
rs972752015 Chr.17: 39761124-39761124 on Build GRCh38
rs1029892474 Chr.17: 39760977-39760977 on Build GRCh38
rs572398501 Chr.17: 39761151-39761151 on Build GRCh38
rs887577863 Chr.17: 39761284-39761284 on Build GRCh38
rs1055724150 Chr.17: 39760997-39760998 on Build GRCh38
rs908553781 Chr.17: 39760936-39760936 on Build GRCh38
rs538677278 Chr.17: 39761113-39761113 on Build GRCh38
rs1012729356 Chr.17: 39760921-39760921 on Build GRCh38
rs77203621 Chr.17: 39761191-39761191 on Build GRCh38
rs372547176 Chr.17: 39761258-39761258 on Build GRCh38
rs760063502 Chr.17: 39760892-39760892 on Build GRCh38
rs1004808016 Chr.17: 39761298-39761298 on Build GRCh38
rs747023676 Chr.17: 39760976-39760976 on Build GRCh38
rs982546945 Chr.17: 39761318-39761319 on Build GRCh38
rs536044306 Chr.17: 39761152-39761152 on Build GRCh38
rs1009014809 Chr.17: 39761136-39761136 on Build GRCh38
rs537966462 Chr.17: 39760929-39760929 on Build GRCh38
rs904316341 Chr.17: 39761282-39761282 on Build GRCh38
rs779707221 Chr.17: 39760900-39760900 on Build GRCh38
rs371287000 Chr.17: 39761022-39761024 on Build GRCh38
rs954424975 Chr.17: 39761017-39761017 on Build GRCh38
rs576433503 Chr.17: 39761210-39761210 on Build GRCh38
rs1014891053 Chr.17: 39761305-39761305 on Build GRCh38
rs955659116 Chr.17: 39760960-39760960 on Build GRCh38
rs761084937 Chr.17: 39761128-39761128 on Build GRCh38
rs977423434 Chr.17: 39760971-39760971 on Build GRCh38
rs772493271 Chr.17: 39760957-39760957 on Build GRCh38
rs149512316 Chr.17: 39761017-39761019 on Build GRCh38
rs1027472148 Chr.17: 39760905-39760905 on Build GRCh38
rs1057425317 Chr.17: 39761197-39761197 on Build GRCh38
rs1019070504 Chr.17: 39760883-39760883 on Build GRCh38
rs1044291855 Chr.17: 39761249-39761249 on Build GRCh38
rs999944229 Chr.17: 39760952-39760952 on Build GRCh38
rs571272390 Chr.17: 39761062-39761062 on Build GRCh38
rs549463413 Chr.17: 39761043-39761043 on Build GRCh38
rs754020463 Chr.17: 39761259-39761259 on Build GRCh38
rs1027916779 Chr.17: 39761268-39761268 on Build GRCh38
rs971293415 Chr.17: 39761313-39761314 on Build GRCh38

View Full Product Details