Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147648054-147648570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP6
Entrez Gene ID
51205
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
ACPL1, LPAP, PACPL1
UniGene
Hs.562154
Gene Chromosome Location
Chr.1: 147629658-147670543 on Build GRCh38
Transcripts
NM_001323625.1 NM_016361.4 NR_136633.1 NR_136634.1 NR_136635.1 NR_136636.1 XM_011509601.2 XM_017001422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4658941 c.987T>G p.Y329* Chr.1: 147648402-147648402 on Build GRCh38
3740616 c.1061C>T p.P354L Chr.1: 147648328-147648328 on Build GRCh38
895317 c.1060C>G p.P354A Chr.1: 147648329-147648329 on Build GRCh38
331088 c.1036G>C p.G346R Chr.1: 147648353-147648353 on Build GRCh38
5377526 c.1018C>T p.P340S Chr.1: 147648371-147648371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372030217 Chr.1: 147648393-147648393 on Build GRCh38
rs782806936 Chr.1: 147648354-147648354 on Build GRCh38
rs782022454 Chr.1: 147648297-147648297 on Build GRCh38
rs782475765 Chr.1: 147648269-147648269 on Build GRCh38
rs782008577 Chr.1: 147648419-147648419 on Build GRCh38
rs150472235 Chr.1: 147648446-147648446 on Build GRCh38
rs782019991 Chr.1: 147648146-147648146 on Build GRCh38
rs782611985 Chr.1: 147648401-147648401 on Build GRCh38
rs782082534 Chr.1: 147648443-147648443 on Build GRCh38
rs782386072 Chr.1: 147648407-147648407 on Build GRCh38
rs368571661 Chr.1: 147648375-147648375 on Build GRCh38
rs944838009 Chr.1: 147648120-147648120 on Build GRCh38
rs782297294 Chr.1: 147648302-147648302 on Build GRCh38
rs912135918 Chr.1: 147648477-147648477 on Build GRCh38
rs897571119 Chr.1: 147648143-147648143 on Build GRCh38
rs587641218 Chr.1: 147648252-147648252 on Build GRCh38
rs782315254 Chr.1: 147648474-147648474 on Build GRCh38
rs143417376 Chr.1: 147648259-147648259 on Build GRCh38
rs376534452 Chr.1: 147648236-147648236 on Build GRCh38
rs1051133920 Chr.1: 147648102-147648102 on Build GRCh38
rs989980947 Chr.1: 147648397-147648397 on Build GRCh38
rs782044515 Chr.1: 147648227-147648227 on Build GRCh38
rs897997502 Chr.1: 147648519-147648519 on Build GRCh38
rs782057047 Chr.1: 147648228-147648228 on Build GRCh38
rs782647726 Chr.1: 147648200-147648200 on Build GRCh38
rs369372085 Chr.1: 147648451-147648451 on Build GRCh38
rs782079243 Chr.1: 147648323-147648323 on Build GRCh38
rs782315322 Chr.1: 147648318-147648318 on Build GRCh38
rs116353121 Chr.1: 147648394-147648394 on Build GRCh38
rs782281061 Chr.1: 147648201-147648201 on Build GRCh38
rs781817606 Chr.1: 147648250-147648250 on Build GRCh38
rs782643752 Chr.1: 147648370-147648370 on Build GRCh38
rs375150545 Chr.1: 147648432-147648432 on Build GRCh38
rs782239307 Chr.1: 147648405-147648405 on Build GRCh38
rs781900285 Chr.1: 147648359-147648359 on Build GRCh38
rs376211817 Chr.1: 147648342-147648342 on Build GRCh38
rs587674018 Chr.1: 147648510-147648510 on Build GRCh38
rs994626464 Chr.1: 147648194-147648194 on Build GRCh38
rs989487526 Chr.1: 147648087-147648087 on Build GRCh38
rs587599700 Chr.1: 147648486-147648486 on Build GRCh38
rs373845128 Chr.1: 147648199-147648199 on Build GRCh38
rs17853316 Chr.1: 147648249-147648249 on Build GRCh38
rs782697677 Chr.1: 147648264-147648264 on Build GRCh38
rs781939778 Chr.1: 147648319-147648319 on Build GRCh38
rs915231346 Chr.1: 147648095-147648095 on Build GRCh38
rs782129830 Chr.1: 147648223-147648223 on Build GRCh38
rs781875815 Chr.1: 147648450-147648450 on Build GRCh38
rs782395582 Chr.1: 147648276-147648276 on Build GRCh38
rs372793677 Chr.1: 147648425-147648425 on Build GRCh38
rs782420040 Chr.1: 147648202-147648202 on Build GRCh38
rs782144310 Chr.1: 147648218-147648218 on Build GRCh38
rs782743074 Chr.1: 147648327-147648327 on Build GRCh38
rs760427695 Chr.1: 147648098-147648098 on Build GRCh38
rs116116332 Chr.1: 147648126-147648126 on Build GRCh38
rs782815602 Chr.1: 147648294-147648294 on Build GRCh38
rs782537051 Chr.1: 147648255-147648255 on Build GRCh38
rs376790669 Chr.1: 147648368-147648368 on Build GRCh38
rs782212974 Chr.1: 147648212-147648212 on Build GRCh38
rs782720686 Chr.1: 147648240-147648240 on Build GRCh38
rs781940205 Chr.1: 147648441-147648441 on Build GRCh38
rs782358266 Chr.1: 147648215-147648215 on Build GRCh38
rs201634452 Chr.1: 147648328-147648328 on Build GRCh38
rs781986500 Chr.1: 147648216-147648216 on Build GRCh38
rs915219308 Chr.1: 147648414-147648414 on Build GRCh38
rs192579981 Chr.1: 147648108-147648108 on Build GRCh38
rs931996531 Chr.1: 147648449-147648449 on Build GRCh38
rs782049544 Chr.1: 147648361-147648361 on Build GRCh38
rs146616655 Chr.1: 147648251-147648251 on Build GRCh38
rs782583619 Chr.1: 147648389-147648389 on Build GRCh38

View Full Product Details