Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 61760496-61761012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf107
Entrez Gene ID
219621
Gene Name
chromosome 10 open reading frame 107
Gene Aliases
bA63A2.1
UniGene
Hs.673160
Gene Chromosome Location
Chr.10: 61662927-61766766 on Build GRCh38
Transcripts
NM_173554.2 XM_005269598.3 XM_005269599.4 XM_005269600.4 XM_006717690.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2157978 c.436G>A p.E146K Chr.10: 61760887-61760887 on Build GRCh38
2157979 c.463T>C p.S155P Chr.10: 61760914-61760914 on Build GRCh38
2157980 c.481G>T p.V161F Chr.10: 61760932-61760932 on Build GRCh38
1128027 c.489T>C p.F163F Chr.10: 61760940-61760940 on Build GRCh38
1185093 c.424G>T p.E142* Chr.10: 61760875-61760875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776859799 Chr.10: 61760866-61760866 on Build GRCh38
rs531051840 Chr.10: 61760902-61760902 on Build GRCh38
rs368412334 Chr.10: 61760704-61760704 on Build GRCh38
rs988668301 Chr.10: 61760749-61760749 on Build GRCh38
rs981094884 Chr.10: 61760681-61760681 on Build GRCh38
rs184833850 Chr.10: 61760554-61760554 on Build GRCh38
rs10994912 Chr.10: 61760859-61760859 on Build GRCh38
rs375217844 Chr.10: 61760927-61760927 on Build GRCh38
rs761814732 Chr.10: 61760869-61760869 on Build GRCh38
rs768775624 Chr.10: 61760535-61760535 on Build GRCh38
rs575554989 Chr.10: 61760598-61760598 on Build GRCh38
rs947192032 Chr.10: 61760786-61760786 on Build GRCh38
rs970137864 Chr.10: 61760680-61760680 on Build GRCh38
rs759389754 Chr.10: 61760912-61760912 on Build GRCh38
rs761066544 Chr.10: 61760842-61760842 on Build GRCh38
rs768842663 Chr.10: 61760851-61760851 on Build GRCh38
rs758799202 Chr.10: 61760973-61760973 on Build GRCh38
rs189556265 Chr.10: 61760656-61760656 on Build GRCh38
rs1043797018 Chr.10: 61760799-61760799 on Build GRCh38
rs1026513647 Chr.10: 61760650-61760650 on Build GRCh38
rs770044523 Chr.10: 61760777-61760777 on Build GRCh38
rs952173199 Chr.10: 61760655-61760655 on Build GRCh38
rs376917011 Chr.10: 61760878-61760878 on Build GRCh38
rs994147397 Chr.10: 61760608-61760608 on Build GRCh38
rs180741631 Chr.10: 61760764-61760764 on Build GRCh38
rs528372548 Chr.10: 61760711-61760711 on Build GRCh38
rs752415805 Chr.10: 61760922-61760922 on Build GRCh38
rs1007262132 Chr.10: 61760691-61760691 on Build GRCh38
rs138221401 Chr.10: 61760884-61760884 on Build GRCh38
rs902132172 Chr.10: 61760605-61760605 on Build GRCh38
rs61850528 Chr.10: 61760580-61760580 on Build GRCh38
rs756765492 Chr.10: 61760942-61760942 on Build GRCh38
rs946823910 Chr.10: 61760531-61760531 on Build GRCh38
rs890145834 Chr.10: 61760686-61760686 on Build GRCh38
rs745447908 Chr.10: 61760960-61760960 on Build GRCh38
rs889127562 Chr.10: 61760565-61760565 on Build GRCh38
rs145707790 Chr.10: 61760835-61760835 on Build GRCh38
rs1992625 Chr.10: 61760940-61760940 on Build GRCh38
rs115694572 Chr.10: 61760836-61760836 on Build GRCh38
rs562600873 Chr.10: 61760857-61760857 on Build GRCh38
rs546009761 Chr.10: 61760639-61760639 on Build GRCh38
rs1022694943 Chr.10: 61760669-61760669 on Build GRCh38
rs541915713 Chr.10: 61760576-61760576 on Build GRCh38
rs928289625 Chr.10: 61760710-61760710 on Build GRCh38
rs1007524091 Chr.10: 61760570-61760570 on Build GRCh38
rs1043765658 Chr.10: 61760532-61760532 on Build GRCh38
rs1017349372 Chr.10: 61760744-61760744 on Build GRCh38
rs201465999 Chr.10: 61760816-61760816 on Build GRCh38
rs778270785 Chr.10: 61760945-61760945 on Build GRCh38
rs934622589 Chr.10: 61760729-61760729 on Build GRCh38
rs767532516 Chr.10: 61760913-61760913 on Build GRCh38
rs780658531 Chr.10: 61760978-61760978 on Build GRCh38

View Full Product Details