Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53983429-53984002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG8
Entrez Gene ID
59283
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 8
Gene Aliases
-
UniGene
Hs.631595
Gene Chromosome Location
Chr.19: 53963036-53990215 on Build GRCh38
Transcripts
NM_031895.5

SNP Detail

SNP ID Chromosome Location
rs1047365583 Chr.19: 53983933-53983933 on Build GRCh38
rs982061414 Chr.19: 53983566-53983566 on Build GRCh38
rs549979974 Chr.19: 53983782-53983782 on Build GRCh38
rs568194732 Chr.19: 53983852-53983852 on Build GRCh38
rs528985168 Chr.19: 53983870-53983870 on Build GRCh38
rs1033640363 Chr.19: 53983787-53983787 on Build GRCh38
rs926656942 Chr.19: 53983585-53983585 on Build GRCh38
rs894558689 Chr.19: 53983969-53983969 on Build GRCh38
rs1032174820 Chr.19: 53983463-53983463 on Build GRCh38
rs117787324 Chr.19: 53983708-53983708 on Build GRCh38
rs773285750 Chr.19: 53983722-53983722 on Build GRCh38
rs577325621 Chr.19: 53983523-53983523 on Build GRCh38
rs995778686 Chr.19: 53983630-53983630 on Build GRCh38
rs970658960 Chr.19: 53983767-53983767 on Build GRCh38
rs973428237 Chr.19: 53983828-53983828 on Build GRCh38
rs983610768 Chr.19: 53983853-53983853 on Build GRCh38
rs776141014 Chr.19: 53983687-53983687 on Build GRCh38
rs978016920 Chr.19: 53983776-53983776 on Build GRCh38
rs920726219 Chr.19: 53983619-53983619 on Build GRCh38
rs954846467 Chr.19: 53983822-53983822 on Build GRCh38
rs912056038 Chr.19: 53983868-53983868 on Build GRCh38
rs775029991 Chr.19: 53983617-53983617 on Build GRCh38
rs950883474 Chr.19: 53983851-53983851 on Build GRCh38
rs115618479 Chr.19: 53983644-53983644 on Build GRCh38
rs570122549 Chr.19: 53983742-53983742 on Build GRCh38
rs1003450996 Chr.19: 53983786-53983786 on Build GRCh38
rs932334356 Chr.19: 53983911-53983911 on Build GRCh38
rs957899002 Chr.19: 53983465-53983465 on Build GRCh38
rs768391416 Chr.19: 53983672-53983672 on Build GRCh38
rs893601820 Chr.19: 53983718-53983718 on Build GRCh38
rs7257865 Chr.19: 53983799-53983799 on Build GRCh38
rs1040724028 Chr.19: 53983679-53983679 on Build GRCh38
rs185969955 Chr.19: 53983686-53983686 on Build GRCh38
rs1048419776 Chr.19: 53983653-53983653 on Build GRCh38
rs181390662 Chr.19: 53983661-53983661 on Build GRCh38
rs988429645 Chr.19: 53983486-53983486 on Build GRCh38
rs1053514403 Chr.19: 53983947-53983947 on Build GRCh38
rs1012961399 Chr.19: 53983975-53983975 on Build GRCh38
rs73937632 Chr.19: 53983698-53983698 on Build GRCh38
rs996632041 Chr.19: 53983447-53983447 on Build GRCh38
rs1042187750 Chr.19: 53983967-53983967 on Build GRCh38
rs767727989 Chr.19: 53983905-53983905 on Build GRCh38
rs746318834 Chr.19: 53983540-53983540 on Build GRCh38
rs73937633 Chr.19: 53983955-53983955 on Build GRCh38
rs1009422168 Chr.19: 53983750-53983750 on Build GRCh38
rs534231701 Chr.19: 53983957-53983957 on Build GRCh38
rs935064746 Chr.19: 53983479-53983479 on Build GRCh38
rs547121078 Chr.19: 53983884-53983884 on Build GRCh38
rs529609279 Chr.19: 53983610-53983610 on Build GRCh38
rs912241650 Chr.19: 53983488-53983488 on Build GRCh38
rs7254456 Chr.19: 53983519-53983519 on Build GRCh38
rs986531252 Chr.19: 53983847-53983847 on Build GRCh38
rs560530175 Chr.19: 53983670-53983670 on Build GRCh38
rs920716993 Chr.19: 53983846-53983846 on Build GRCh38
rs7257802 Chr.19: 53983557-53983557 on Build GRCh38
rs923725441 Chr.19: 53983473-53983473 on Build GRCh38
rs893531706 Chr.19: 53983954-53983954 on Build GRCh38
rs1039343319 Chr.19: 53983876-53983876 on Build GRCh38
rs117568575 Chr.19: 53983751-53983751 on Build GRCh38
rs1022328431 Chr.19: 53983748-53983748 on Build GRCh38
rs757615013 Chr.19: 53983515-53983515 on Build GRCh38
rs564140892 Chr.19: 53983717-53983717 on Build GRCh38

View Full Product Details