Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 2729908-2730501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV2
Entrez Gene ID
169522
Gene Name
potassium voltage-gated channel modifier subfamily V member 2
Gene Aliases
KV11.1, Kv8.2, RCD3B
UniGene
Hs.622675
Gene Chromosome Location
Chr.9: 2717526-2730037 on Build GRCh38
Transcripts
NM_133497.3

SNP Detail

SNP ID Chromosome Location
rs940927166 Chr.9: 2730269-2730269 on Build GRCh38
rs981155436 Chr.9: 2730200-2730200 on Build GRCh38
rs760755859 Chr.9: 2730031-2730040 on Build GRCh38
rs1011579428 Chr.9: 2730255-2730255 on Build GRCh38
rs746989765 Chr.9: 2730006-2730006 on Build GRCh38
rs1028595546 Chr.9: 2730052-2730053 on Build GRCh38
rs1021382687 Chr.9: 2730381-2730381 on Build GRCh38
rs1036002063 Chr.9: 2730210-2730210 on Build GRCh38
rs989528023 Chr.9: 2730099-2730099 on Build GRCh38
rs900507267 Chr.9: 2730026-2730026 on Build GRCh38
rs934476153 Chr.9: 2729981-2729981 on Build GRCh38
rs567151801 Chr.9: 2729996-2729996 on Build GRCh38
rs978192351 Chr.9: 2730122-2730122 on Build GRCh38
rs554878962 Chr.9: 2730147-2730147 on Build GRCh38
rs1028982449 Chr.9: 2730249-2730249 on Build GRCh38
rs60723920 Chr.9: 2730035-2730035 on Build GRCh38
rs541207530 Chr.9: 2730013-2730013 on Build GRCh38
rs535730174 Chr.9: 2730016-2730016 on Build GRCh38
rs866471820 Chr.9: 2729943-2729943 on Build GRCh38
rs41313448 Chr.9: 2730315-2730315 on Build GRCh38
rs1005048357 Chr.9: 2730128-2730128 on Build GRCh38
rs780101028 Chr.9: 2730440-2730440 on Build GRCh38
rs894121306 Chr.9: 2729982-2729982 on Build GRCh38
rs888130659 Chr.9: 2730209-2730209 on Build GRCh38
rs561182653 Chr.9: 2730456-2730457 on Build GRCh38
rs542515501 Chr.9: 2730380-2730380 on Build GRCh38
rs928465529 Chr.9: 2729968-2729968 on Build GRCh38
rs551029643 Chr.9: 2730475-2730475 on Build GRCh38
rs886690146 Chr.9: 2730261-2730261 on Build GRCh38
rs113149553 Chr.9: 2730002-2730002 on Build GRCh38
rs995264267 Chr.9: 2730182-2730182 on Build GRCh38
rs1002944016 Chr.9: 2730304-2730304 on Build GRCh38
rs1022601091 Chr.9: 2730187-2730187 on Build GRCh38
rs978159642 Chr.9: 2730282-2730282 on Build GRCh38
rs773041500 Chr.9: 2730369-2730369 on Build GRCh38
rs938559254 Chr.9: 2730142-2730142 on Build GRCh38
rs754400546 Chr.9: 2730144-2730144 on Build GRCh38
rs113975253 Chr.9: 2729942-2729942 on Build GRCh38
rs574862166 Chr.9: 2730065-2730065 on Build GRCh38
rs914406449 Chr.9: 2730234-2730234 on Build GRCh38
rs1037839379 Chr.9: 2730275-2730275 on Build GRCh38
rs41315999 Chr.9: 2730279-2730279 on Build GRCh38
rs969966828 Chr.9: 2729966-2729966 on Build GRCh38
rs966747828 Chr.9: 2730121-2730121 on Build GRCh38
rs981779617 Chr.9: 2729967-2729967 on Build GRCh38
rs114811489 Chr.9: 2730286-2730286 on Build GRCh38
rs921929888 Chr.9: 2730240-2730240 on Build GRCh38
rs766562253 Chr.9: 2730452-2730453 on Build GRCh38
rs762865456 Chr.9: 2730370-2730370 on Build GRCh38
rs374693766 Chr.9: 2729939-2729939 on Build GRCh38
rs955990102 Chr.9: 2730223-2730223 on Build GRCh38
rs922004000 Chr.9: 2730404-2730404 on Build GRCh38
rs768798207 Chr.9: 2730074-2730074 on Build GRCh38
rs1051807711 Chr.9: 2730257-2730257 on Build GRCh38
rs758666130 Chr.9: 2730378-2730378 on Build GRCh38
rs530904659 Chr.9: 2730415-2730415 on Build GRCh38
rs1048129374 Chr.9: 2729993-2729993 on Build GRCh38
rs544065378 Chr.9: 2730449-2730456 on Build GRCh38
rs905640777 Chr.9: 2730299-2730299 on Build GRCh38
rs975244191 Chr.9: 2730238-2730238 on Build GRCh38
rs907886616 Chr.9: 2730455-2730455 on Build GRCh38
rs543824045 Chr.9: 2730139-2730139 on Build GRCh38
rs1048016789 Chr.9: 2730196-2730196 on Build GRCh38
rs947512932 Chr.9: 2730003-2730003 on Build GRCh38
rs1010115207 Chr.9: 2730372-2730372 on Build GRCh38
rs915251594 Chr.9: 2730112-2730112 on Build GRCh38
rs960143495 Chr.9: 2730296-2730296 on Build GRCh38
rs965531069 Chr.9: 2730031-2730035 on Build GRCh38
rs988601236 Chr.9: 2730230-2730230 on Build GRCh38
rs868713990 Chr.9: 2730018-2730018 on Build GRCh38
rs890007105 Chr.9: 2730251-2730251 on Build GRCh38
rs577350762 Chr.9: 2730191-2730191 on Build GRCh38
rs899344515 Chr.9: 2730277-2730277 on Build GRCh38
rs184267658 Chr.9: 2730385-2730385 on Build GRCh38
rs142869197 Chr.9: 2730448-2730449 on Build GRCh38
rs57109373 Chr.9: 2729989-2729989 on Build GRCh38
rs987840606 Chr.9: 2730426-2730426 on Build GRCh38
rs1057063752 Chr.9: 2729956-2729956 on Build GRCh38
rs974625422 Chr.9: 2730389-2730389 on Build GRCh38
rs757753549 Chr.9: 2730213-2730213 on Build GRCh38
rs933305502 Chr.9: 2730241-2730241 on Build GRCh38
rs755464261 Chr.9: 2730449-2730452 on Build GRCh38
rs947437985 Chr.9: 2730236-2730236 on Build GRCh38
rs574999979 Chr.9: 2730422-2730422 on Build GRCh38

View Full Product Details