Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31995607-31996108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4A
Entrez Gene ID
720
Gene Name
complement component 4A (Rodgers blood group)
Gene Aliases
C4, C4A2, C4A3, C4A4, C4A6, C4AD, C4S, CO4, CPAMD2, RG
UniGene
Hs.534847
Gene Chromosome Location
Chr.6: 31982057-32002680 on Build GRCh38
Transcripts
NM_001252204.1 NM_007293.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624961 c.3346T>C p.S1116P Chr.6: 31996070-31996070 on Build GRCh38
1329779 c.3332A>C p.Q1111P Chr.6: 31996056-31996056 on Build GRCh38
3624963 c.3365C>T p.P1122L Chr.6: 31996089-31996089 on Build GRCh38
3624959 c.3178C>T p.R1060W Chr.6: 31995742-31995742 on Build GRCh38
3861935 c.3215G>A p.R1072Q Chr.6: 31995779-31995779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756132144 Chr.6: 31995922-31995922 on Build GRCh38
rs759901298 Chr.6: 31995795-31995795 on Build GRCh38
rs751583416 Chr.6: 31995846-31995846 on Build GRCh38
rs747843856 Chr.6: 31995960-31995960 on Build GRCh38
rs201206908 Chr.6: 31996082-31996082 on Build GRCh38
rs746517517 Chr.6: 31995997-31995997 on Build GRCh38
rs754198198 Chr.6: 31995690-31995690 on Build GRCh38
rs765460604 Chr.6: 31995708-31995708 on Build GRCh38
rs758502569 Chr.6: 31995710-31995710 on Build GRCh38
rs1000726010 Chr.6: 31995990-31995990 on Build GRCh38
rs573825689 Chr.6: 31996086-31996086 on Build GRCh38
rs749075087 Chr.6: 31995743-31995743 on Build GRCh38
rs753789771 Chr.6: 31995940-31995940 on Build GRCh38
rs527733843 Chr.6: 31995851-31995851 on Build GRCh38
rs374875689 Chr.6: 31995797-31995797 on Build GRCh38
rs748217591 Chr.6: 31995827-31995827 on Build GRCh38
rs946150601 Chr.6: 31995663-31995663 on Build GRCh38
rs757987556 Chr.6: 31995967-31995967 on Build GRCh38
rs777297715 Chr.6: 31996075-31996075 on Build GRCh38
rs767658336 Chr.6: 31996036-31996036 on Build GRCh38
rs770189869 Chr.6: 31996010-31996010 on Build GRCh38
rs768510893 Chr.6: 31995749-31995749 on Build GRCh38
rs567824865 Chr.6: 31995895-31995895 on Build GRCh38
rs1039335725 Chr.6: 31996001-31996001 on Build GRCh38
rs747757875 Chr.6: 31995764-31995764 on Build GRCh38
rs777664867 Chr.6: 31995712-31995712 on Build GRCh38
rs777151814 Chr.6: 31996007-31996007 on Build GRCh38
rs752848113 Chr.6: 31995709-31995709 on Build GRCh38
rs750422695 Chr.6: 31995727-31995727 on Build GRCh38
rs572605375 Chr.6: 31996012-31996012 on Build GRCh38
rs759813561 Chr.6: 31995698-31995698 on Build GRCh38
rs764275000 Chr.6: 31995831-31995831 on Build GRCh38
rs778829969 Chr.6: 31995953-31995953 on Build GRCh38
rs79299066 Chr.6: 31996083-31996083 on Build GRCh38
rs367681824 Chr.6: 31995993-31995993 on Build GRCh38
rs763063069 Chr.6: 31995807-31995807 on Build GRCh38
rs772097613 Chr.6: 31995933-31995934 on Build GRCh38
rs202091437 Chr.6: 31996067-31996067 on Build GRCh38
rs758158004 Chr.6: 31996072-31996072 on Build GRCh38
rs774574186 Chr.6: 31996015-31996015 on Build GRCh38
rs779962386 Chr.6: 31995742-31995742 on Build GRCh38
rs753877618 Chr.6: 31996050-31996050 on Build GRCh38
rs143990092 Chr.6: 31995728-31995728 on Build GRCh38
rs374886374 Chr.6: 31996011-31996011 on Build GRCh38
rs371801898 Chr.6: 31996003-31996003 on Build GRCh38
rs766430084 Chr.6: 31995924-31995924 on Build GRCh38
rs140362734 Chr.6: 31996009-31996009 on Build GRCh38
rs368406231 Chr.6: 31996054-31996054 on Build GRCh38
rs765190546 Chr.6: 31996057-31996057 on Build GRCh38
rs771330838 Chr.6: 31995779-31995779 on Build GRCh38
rs775710806 Chr.6: 31995799-31995799 on Build GRCh38
rs766806798 Chr.6: 31995670-31995670 on Build GRCh38
rs778742093 Chr.6: 31995750-31995750 on Build GRCh38
rs147162052 Chr.6: 31995782-31995782 on Build GRCh38
rs201134904 Chr.6: 31995778-31995778 on Build GRCh38
rs55798441 Chr.6: 31995667-31995667 on Build GRCh38
rs538548145 Chr.6: 31995899-31995899 on Build GRCh38
rs772570568 Chr.6: 31995773-31995773 on Build GRCh38
rs771664862 Chr.6: 31995765-31995765 on Build GRCh38
rs761801471 Chr.6: 31996021-31996021 on Build GRCh38
rs556455246 Chr.6: 31995951-31995951 on Build GRCh38
rs549402002 Chr.6: 31995852-31995852 on Build GRCh38

View Full Product Details