Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35576893-35577369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP5
Entrez Gene ID
2289
Gene Name
FK506 binding protein 5
Gene Aliases
AIG6, FKBP51, FKBP54, P54, PPIase, Ptg-10
UniGene
Hs.407190
Gene Chromosome Location
Chr.6: 35573585-35728583 on Build GRCh38
Transcripts
NM_001145775.2 NM_001145776.1 NM_004117.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5061736 c.1227C>T p.Y409Y Chr.6: 35577033-35577033 on Build GRCh38
2155558 c.1217G>A p.R406H Chr.6: 35577043-35577043 on Build GRCh38
1754758 c.1057G>A p.G353S Chr.6: 35577203-35577203 on Build GRCh38
3921572 c.1034G>A p.G345E Chr.6: 35577226-35577226 on Build GRCh38
6664271 c.1179G>A p.M393I Chr.6: 35577081-35577081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771692610 Chr.6: 35577078-35577078 on Build GRCh38
rs180682572 Chr.6: 35577084-35577084 on Build GRCh38
rs376199256 Chr.6: 35577060-35577060 on Build GRCh38
rs1011778638 Chr.6: 35576926-35576926 on Build GRCh38
rs34866878 Chr.6: 35577165-35577165 on Build GRCh38
rs747242817 Chr.6: 35577002-35577002 on Build GRCh38
rs770520834 Chr.6: 35577091-35577091 on Build GRCh38
rs767683901 Chr.6: 35577140-35577140 on Build GRCh38
rs866291676 Chr.6: 35577036-35577036 on Build GRCh38
rs764892232 Chr.6: 35577049-35577049 on Build GRCh38
rs868641354 Chr.6: 35577101-35577101 on Build GRCh38
rs373264047 Chr.6: 35577054-35577054 on Build GRCh38
rs144913669 Chr.6: 35577033-35577033 on Build GRCh38
rs755396867 Chr.6: 35577069-35577069 on Build GRCh38
rs760776253 Chr.6: 35577142-35577142 on Build GRCh38
rs536210621 Chr.6: 35577296-35577296 on Build GRCh38
rs751820100 Chr.6: 35577050-35577050 on Build GRCh38
rs771627884 Chr.6: 35577032-35577032 on Build GRCh38
rs776885920 Chr.6: 35576948-35576949 on Build GRCh38
rs759626624 Chr.6: 35577165-35577165 on Build GRCh38
rs146787424 Chr.6: 35577112-35577112 on Build GRCh38
rs762377537 Chr.6: 35577053-35577053 on Build GRCh38
rs762696275 Chr.6: 35577255-35577255 on Build GRCh38
rs547900322 Chr.6: 35577336-35577336 on Build GRCh38
rs746377759 Chr.6: 35576961-35576961 on Build GRCh38
rs371249911 Chr.6: 35577259-35577259 on Build GRCh38
rs374789777 Chr.6: 35577208-35577208 on Build GRCh38
rs1024217045 Chr.6: 35577131-35577131 on Build GRCh38
rs759488963 Chr.6: 35577106-35577106 on Build GRCh38
rs780579204 Chr.6: 35577062-35577062 on Build GRCh38
rs762796041 Chr.6: 35577138-35577138 on Build GRCh38
rs941107659 Chr.6: 35576919-35576919 on Build GRCh38
rs1020672211 Chr.6: 35576965-35576990 on Build GRCh38
rs766400491 Chr.6: 35577160-35577160 on Build GRCh38
rs759431987 Chr.6: 35577044-35577044 on Build GRCh38
rs148272640 Chr.6: 35577043-35577043 on Build GRCh38
rs734369 Chr.6: 35577075-35577075 on Build GRCh38
rs76042921 Chr.6: 35576945-35576945 on Build GRCh38
rs755375739 Chr.6: 35577182-35577182 on Build GRCh38
rs1011078955 Chr.6: 35576951-35576976 on Build GRCh38
rs747844338 Chr.6: 35577077-35577077 on Build GRCh38
rs757380566 Chr.6: 35576951-35576951 on Build GRCh38
rs749701989 Chr.6: 35576989-35576989 on Build GRCh38
rs779473333 Chr.6: 35577187-35577187 on Build GRCh38
rs375912702 Chr.6: 35577266-35577266 on Build GRCh38
rs753049321 Chr.6: 35577198-35577198 on Build GRCh38
rs776731286 Chr.6: 35577095-35577095 on Build GRCh38
rs552172467 Chr.6: 35577015-35577015 on Build GRCh38
rs375321658 Chr.6: 35576949-35576949 on Build GRCh38
rs547741786 Chr.6: 35577251-35577251 on Build GRCh38
rs140740382 Chr.6: 35577211-35577211 on Build GRCh38
rs73417616 Chr.6: 35576928-35576928 on Build GRCh38
rs747710519 Chr.6: 35577028-35577028 on Build GRCh38
rs542669044 Chr.6: 35576948-35576948 on Build GRCh38
rs370284375 Chr.6: 35577141-35577141 on Build GRCh38
rs777543188 Chr.6: 35577083-35577083 on Build GRCh38
rs141262636 Chr.6: 35577072-35577072 on Build GRCh38
rs965428955 Chr.6: 35577011-35577011 on Build GRCh38
rs369801947 Chr.6: 35576968-35576968 on Build GRCh38
rs200398524 Chr.6: 35577238-35577238 on Build GRCh38
rs143893856 Chr.6: 35577164-35577164 on Build GRCh38
rs774087038 Chr.6: 35577026-35577026 on Build GRCh38
rs139316938 Chr.6: 35577133-35577133 on Build GRCh38
rs890065415 Chr.6: 35576929-35576929 on Build GRCh38
rs1019638390 Chr.6: 35576973-35576973 on Build GRCh38
rs775476348 Chr.6: 35577254-35577254 on Build GRCh38
rs199684698 Chr.6: 35577245-35577245 on Build GRCh38
rs781072865 Chr.6: 35576952-35576952 on Build GRCh38
rs750561820 Chr.6: 35577055-35577055 on Build GRCh38
rs760245959 Chr.6: 35577038-35577038 on Build GRCh38
rs768872131 Chr.6: 35577000-35577000 on Build GRCh38
rs916481623 Chr.6: 35577162-35577162 on Build GRCh38
rs769677713 Chr.6: 35577252-35577252 on Build GRCh38
rs760831586 Chr.6: 35577267-35577267 on Build GRCh38
rs777596648 Chr.6: 35577210-35577210 on Build GRCh38
rs756681379 Chr.6: 35576964-35576964 on Build GRCh38
rs766453660 Chr.6: 35577269-35577269 on Build GRCh38
rs918776070 Chr.6: 35576939-35576939 on Build GRCh38
rs749756951 Chr.6: 35577068-35577068 on Build GRCh38

View Full Product Details