Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 76173557-76174068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PBDC1
Entrez Gene ID
51260
Gene Name
polysaccharide biosynthesis domain containing 1
Gene Aliases
CXorf26
UniGene
Hs.370100
Gene Chromosome Location
Chr.X: 76172929-76178310 on Build GRCh38
Transcripts
NM_001300888.1 NM_016500.3

SNP Detail

SNP ID Chromosome Location
rs782687277 Chr.X: 76173626-76173626 on Build GRCh38
rs374917281 Chr.X: 76173957-76173957 on Build GRCh38
rs782587598 Chr.X: 76173650-76173650 on Build GRCh38
rs782792916 Chr.X: 76173636-76173636 on Build GRCh38
rs782698732 Chr.X: 76173822-76173822 on Build GRCh38
rs781971437 Chr.X: 76173687-76173687 on Build GRCh38
rs781867907 Chr.X: 76173606-76173606 on Build GRCh38
rs781911481 Chr.X: 76173648-76173648 on Build GRCh38
rs1025097212 Chr.X: 76173696-76173696 on Build GRCh38
rs980757445 Chr.X: 76173727-76173727 on Build GRCh38
rs782227532 Chr.X: 76173764-76173764 on Build GRCh38
rs930516960 Chr.X: 76173758-76173758 on Build GRCh38
rs781984016 Chr.X: 76173675-76173675 on Build GRCh38
rs781952411 Chr.X: 76173700-76173700 on Build GRCh38
rs782389065 Chr.X: 76173658-76173658 on Build GRCh38
rs907689973 Chr.X: 76173862-76173862 on Build GRCh38
rs782594593 Chr.X: 76173681-76173681 on Build GRCh38
rs782546549 Chr.X: 76173622-76173622 on Build GRCh38
rs919942005 Chr.X: 76173742-76173742 on Build GRCh38
rs11541301 Chr.X: 76173612-76173612 on Build GRCh38
rs782756723 Chr.X: 76173604-76173604 on Build GRCh38
rs782161107 Chr.X: 76173592-76173592 on Build GRCh38
rs376142225 Chr.X: 76173697-76173697 on Build GRCh38
rs782524493 Chr.X: 76173639-76173639 on Build GRCh38
rs782077897 Chr.X: 76173611-76173611 on Build GRCh38
rs1040516646 Chr.X: 76174031-76174031 on Build GRCh38
rs782655502 Chr.X: 76173902-76173902 on Build GRCh38
rs782449196 Chr.X: 76173676-76173676 on Build GRCh38
rs371393851 Chr.X: 76173590-76173590 on Build GRCh38
rs782607971 Chr.X: 76173641-76173641 on Build GRCh38
rs369977828 Chr.X: 76173709-76173709 on Build GRCh38
rs782418057 Chr.X: 76173673-76173673 on Build GRCh38
rs782232694 Chr.X: 76173647-76173647 on Build GRCh38
rs782500583 Chr.X: 76173804-76173804 on Build GRCh38

View Full Product Details