Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97555110-97555704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36B
Entrez Gene ID
57730
Gene Name
ankyrin repeat domain 36B
Gene Aliases
KIAA1641
UniGene
Hs.532921
Gene Chromosome Location
Chr.2: 97492663-97590245 on Build GRCh38
Transcripts
NM_025190.3 XM_005263995.4 XM_006712661.1 XM_011511555.1 XM_011511556.1 XM_011511557.2 XM_011511558.1 XM_017004578.1 XM_017004579.1 XM_017004580.1 XM_017004581.1 XM_017004582.1 XM_017004583.1 XM_017004584.1 XM_017004585.1 XM_017004586.1 XM_017004587.1 XM_017004588.1 XM_017004589.1 XM_017004590.1 XM_017004592.1 XM_017004593.1 XM_017004594.1 XM_017004595.1

SNP Detail

SNP ID Chromosome Location
rs908513538 Chr.2: 97555579-97555579 on Build GRCh38
rs778296696 Chr.2: 97555179-97555179 on Build GRCh38
rs1003285322 Chr.2: 97555285-97555285 on Build GRCh38
rs775505185 Chr.2: 97555234-97555234 on Build GRCh38
rs1003144684 Chr.2: 97555405-97555405 on Build GRCh38
rs549954810 Chr.2: 97555353-97555353 on Build GRCh38
rs535391677 Chr.2: 97555442-97555442 on Build GRCh38
rs565169940 Chr.2: 97555411-97555412 on Build GRCh38
rs532945084 Chr.2: 97555349-97555349 on Build GRCh38
rs921275367 Chr.2: 97555135-97555135 on Build GRCh38
rs776829547 Chr.2: 97555216-97555216 on Build GRCh38
rs577670080 Chr.2: 97555524-97555524 on Build GRCh38
rs767496263 Chr.2: 97555289-97555289 on Build GRCh38
rs1044817860 Chr.2: 97555669-97555669 on Build GRCh38
rs755773524 Chr.2: 97555159-97555159 on Build GRCh38
rs569602888 Chr.2: 97555440-97555440 on Build GRCh38
rs904938091 Chr.2: 97555671-97555671 on Build GRCh38
rs745992157 Chr.2: 97555222-97555222 on Build GRCh38
rs771603933 Chr.2: 97555198-97555198 on Build GRCh38
rs12712057 Chr.2: 97555344-97555344 on Build GRCh38
rs770048331 Chr.2: 97555223-97555223 on Build GRCh38
rs369872022 Chr.2: 97555513-97555513 on Build GRCh38
rs904908413 Chr.2: 97555371-97555371 on Build GRCh38
rs534626359 Chr.2: 97555493-97555493 on Build GRCh38
rs565306964 Chr.2: 97555476-97555476 on Build GRCh38
rs749922233 Chr.2: 97555304-97555304 on Build GRCh38
rs747478337 Chr.2: 97555183-97555183 on Build GRCh38
rs879430747 Chr.2: 97555634-97555634 on Build GRCh38
rs559821229 Chr.2: 97555178-97555178 on Build GRCh38
rs878923814 Chr.2: 97555608-97555608 on Build GRCh38
rs1036623959 Chr.2: 97555313-97555313 on Build GRCh38
rs750043329 Chr.2: 97555428-97555428 on Build GRCh38
rs749979583 Chr.2: 97555153-97555153 on Build GRCh38
rs748542021 Chr.2: 97555172-97555172 on Build GRCh38
rs763247940 Chr.2: 97555238-97555238 on Build GRCh38
rs866146945 Chr.2: 97555456-97555456 on Build GRCh38
rs775862485 Chr.2: 97555651-97555651 on Build GRCh38
rs779631372 Chr.2: 97555161-97555161 on Build GRCh38
rs534635511 Chr.2: 97555451-97555451 on Build GRCh38
rs371779590 Chr.2: 97555282-97555282 on Build GRCh38
rs554919643 Chr.2: 97555450-97555450 on Build GRCh38
rs772310214 Chr.2: 97555570-97555570 on Build GRCh38
rs528575927 Chr.2: 97555245-97555245 on Build GRCh38
rs898958450 Chr.2: 97555314-97555314 on Build GRCh38
rs369782940 Chr.2: 97555419-97555420 on Build GRCh38
rs1036246718 Chr.2: 97555600-97555600 on Build GRCh38
rs757212026 Chr.2: 97555132-97555132 on Build GRCh38
rs192028456 Chr.2: 97555307-97555307 on Build GRCh38
rs774165483 Chr.2: 97555274-97555274 on Build GRCh38
rs955385912 Chr.2: 97555526-97555526 on Build GRCh38
rs920456962 Chr.2: 97555606-97555606 on Build GRCh38
rs766977221 Chr.2: 97555138-97555138 on Build GRCh38
rs1015557813 Chr.2: 97555317-97555317 on Build GRCh38
rs543651348 Chr.2: 97555542-97555542 on Build GRCh38
rs1051703819 Chr.2: 97555279-97555279 on Build GRCh38
rs968211487 Chr.2: 97555507-97555507 on Build GRCh38
rs930476376 Chr.2: 97555630-97555630 on Build GRCh38

View Full Product Details