Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32116617-32117110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF6B
Entrez Gene ID
1388
Gene Name
activating transcription factor 6 beta
Gene Aliases
CREB-RP, CREBL1, G13
UniGene
Hs.42853
Gene Chromosome Location
Chr.6: 32115268-32128240 on Build GRCh38
Transcripts
NM_001136153.1 NM_004381.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1443565 c.1640C>T p.P547L Chr.6: 32117082-32117082 on Build GRCh38
1077816 c.1686-1G>A p.? Chr.6: 32116816-32116816 on Build GRCh38
4531360 c.1780G>T p.V594F Chr.6: 32116721-32116721 on Build GRCh38
1077815 c.1727G>A p.R576H Chr.6: 32116774-32116774 on Build GRCh38
4354348 c.1715G>A p.R572H Chr.6: 32116786-32116786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768242668 Chr.6: 32117083-32117083 on Build GRCh38
rs773705127 Chr.6: 32116863-32116864 on Build GRCh38
rs148987710 Chr.6: 32116708-32116708 on Build GRCh38
rs908331260 Chr.6: 32116759-32116759 on Build GRCh38
rs563678991 Chr.6: 32116786-32116786 on Build GRCh38
rs182341643 Chr.6: 32116754-32116754 on Build GRCh38
rs778173486 Chr.6: 32116997-32116997 on Build GRCh38
rs768090887 Chr.6: 32116958-32116958 on Build GRCh38
rs767473804 Chr.6: 32116812-32116812 on Build GRCh38
rs199610969 Chr.6: 32116787-32116787 on Build GRCh38
rs1002257457 Chr.6: 32116910-32116910 on Build GRCh38
rs975676385 Chr.6: 32117048-32117048 on Build GRCh38
rs771386454 Chr.6: 32116758-32116758 on Build GRCh38
rs964277128 Chr.6: 32117018-32117018 on Build GRCh38
rs753847817 Chr.6: 32116819-32116819 on Build GRCh38
rs1034186043 Chr.6: 32116912-32116912 on Build GRCh38
rs879185701 Chr.6: 32116956-32116956 on Build GRCh38
rs748402065 Chr.6: 32116742-32116742 on Build GRCh38
rs763882495 Chr.6: 32116715-32116715 on Build GRCh38
rs1008400086 Chr.6: 32116976-32116976 on Build GRCh38
rs763947131 Chr.6: 32116791-32116791 on Build GRCh38
rs368079808 Chr.6: 32116891-32116891 on Build GRCh38
rs6926448 Chr.6: 32116770-32116770 on Build GRCh38
rs759245940 Chr.6: 32116683-32116683 on Build GRCh38
rs559716525 Chr.6: 32116739-32116739 on Build GRCh38
rs370013339 Chr.6: 32116704-32116704 on Build GRCh38
rs532100174 Chr.6: 32116954-32116954 on Build GRCh38
rs774702849 Chr.6: 32116738-32116738 on Build GRCh38
rs542518888 Chr.6: 32116774-32116774 on Build GRCh38
rs373179783 Chr.6: 32116741-32116741 on Build GRCh38
rs564307366 Chr.6: 32116885-32116885 on Build GRCh38
rs772391185 Chr.6: 32116662-32116662 on Build GRCh38
rs1053370350 Chr.6: 32116870-32116870 on Build GRCh38
rs751351799 Chr.6: 32116800-32116800 on Build GRCh38
rs758586518 Chr.6: 32116863-32116863 on Build GRCh38
rs375944911 Chr.6: 32116669-32116669 on Build GRCh38
rs747515748 Chr.6: 32116757-32116757 on Build GRCh38
rs749391389 Chr.6: 32116731-32116731 on Build GRCh38
rs572621235 Chr.6: 32116643-32116643 on Build GRCh38
rs905286862 Chr.6: 32116900-32116903 on Build GRCh38
rs779020679 Chr.6: 32116854-32116854 on Build GRCh38
rs541130150 Chr.6: 32116680-32116680 on Build GRCh38
rs774932427 Chr.6: 32117056-32117056 on Build GRCh38
rs757209389 Chr.6: 32116728-32116728 on Build GRCh38
rs768405340 Chr.6: 32116661-32116661 on Build GRCh38
rs893474227 Chr.6: 32116881-32116881 on Build GRCh38
rs780077430 Chr.6: 32116729-32116729 on Build GRCh38
rs762636214 Chr.6: 32117059-32117059 on Build GRCh38
rs765905521 Chr.6: 32116674-32116674 on Build GRCh38
rs755062242 Chr.6: 32116834-32116834 on Build GRCh38
rs758221215 Chr.6: 32116709-32116709 on Build GRCh38
rs765008480 Chr.6: 32116684-32116684 on Build GRCh38
rs200540433 Chr.6: 32116670-32116670 on Build GRCh38
rs772466130 Chr.6: 32116748-32116748 on Build GRCh38
rs781766408 Chr.6: 32117008-32117008 on Build GRCh38
rs747359590 Chr.6: 32116998-32116998 on Build GRCh38
rs890247176 Chr.6: 32116966-32116966 on Build GRCh38
rs751504502 Chr.6: 32116717-32116717 on Build GRCh38
rs547131167 Chr.6: 32117026-32117026 on Build GRCh38
rs552380430 Chr.6: 32116858-32116858 on Build GRCh38
rs764918512 Chr.6: 32116776-32116776 on Build GRCh38
rs1038347869 Chr.6: 32116882-32116882 on Build GRCh38
rs761788684 Chr.6: 32116803-32116803 on Build GRCh38

View Full Product Details