Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 32092573-32093166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP5
Entrez Gene ID
394
Gene Name
Rho GTPase activating protein 5
Gene Aliases
GFI2, RhoGAP5, p190-B, p190BRhoGAP
UniGene
Hs.592313
Gene Chromosome Location
Chr.14: 32076152-32159728 on Build GRCh38
Transcripts
NM_001030055.1 NM_001173.2 XM_005267635.3 XM_005267636.3 XM_017021287.1 XM_017021288.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7305438 c.2398C>A p.L800I Chr.14: 32093067-32093067 on Build GRCh38
7291304 c.2001A>G p.S667S Chr.14: 32092670-32092670 on Build GRCh38
955339 c.2387T>G p.L796R Chr.14: 32093056-32093056 on Build GRCh38
6140686 c.2140C>T p.L714F Chr.14: 32092809-32092809 on Build GRCh38
4050524 c.1953C>T p.A651A Chr.14: 32092622-32092622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146041775 Chr.14: 32092973-32092973 on Build GRCh38
rs374527700 Chr.14: 32093053-32093053 on Build GRCh38
rs756332483 Chr.14: 32093114-32093114 on Build GRCh38
rs781511877 Chr.14: 32092920-32092920 on Build GRCh38
rs750015707 Chr.14: 32092614-32092614 on Build GRCh38
rs768232197 Chr.14: 32093066-32093066 on Build GRCh38
rs770072111 Chr.14: 32092807-32092807 on Build GRCh38
rs759100156 Chr.14: 32092965-32092965 on Build GRCh38
rs762994955 Chr.14: 32093079-32093079 on Build GRCh38
rs767618337 Chr.14: 32093096-32093096 on Build GRCh38
rs757953107 Chr.14: 32092622-32092622 on Build GRCh38
rs776464378 Chr.14: 32093067-32093067 on Build GRCh38
rs866726213 Chr.14: 32092599-32092599 on Build GRCh38
rs200687781 Chr.14: 32092636-32092636 on Build GRCh38
rs765834943 Chr.14: 32092708-32092708 on Build GRCh38
rs142663482 Chr.14: 32092969-32092969 on Build GRCh38
rs752413698 Chr.14: 32092869-32092869 on Build GRCh38
rs765151303 Chr.14: 32093071-32093071 on Build GRCh38
rs975341372 Chr.14: 32092798-32092798 on Build GRCh38
rs747267938 Chr.14: 32092693-32092693 on Build GRCh38
rs755065497 Chr.14: 32092999-32092999 on Build GRCh38
rs752559864 Chr.14: 32092738-32092738 on Build GRCh38
rs765813535 Chr.14: 32092701-32092701 on Build GRCh38
rs773910735 Chr.14: 32092828-32092828 on Build GRCh38
rs150293728 Chr.14: 32092758-32092758 on Build GRCh38
rs773822886 Chr.14: 32092963-32092963 on Build GRCh38
rs370267449 Chr.14: 32092974-32092974 on Build GRCh38
rs755228783 Chr.14: 32092905-32092905 on Build GRCh38
rs374179893 Chr.14: 32092679-32092679 on Build GRCh38
rs767289867 Chr.14: 32092729-32092729 on Build GRCh38
rs377117868 Chr.14: 32092811-32092811 on Build GRCh38
rs141747142 Chr.14: 32093012-32093012 on Build GRCh38
rs377277190 Chr.14: 32092625-32092625 on Build GRCh38
rs764639550 Chr.14: 32092595-32092595 on Build GRCh38
rs771592334 Chr.14: 32092968-32092968 on Build GRCh38
rs566879891 Chr.14: 32093000-32093000 on Build GRCh38
rs189967153 Chr.14: 32092956-32092956 on Build GRCh38
rs758465255 Chr.14: 32092761-32092761 on Build GRCh38
rs200248752 Chr.14: 32093016-32093016 on Build GRCh38
rs764275676 Chr.14: 32093118-32093118 on Build GRCh38
rs752789750 Chr.14: 32093113-32093113 on Build GRCh38
rs751531664 Chr.14: 32092703-32092705 on Build GRCh38
rs762222399 Chr.14: 32092697-32092697 on Build GRCh38
rs545845262 Chr.14: 32092644-32092644 on Build GRCh38
rs749658149 Chr.14: 32093019-32093019 on Build GRCh38
rs749207325 Chr.14: 32092759-32092759 on Build GRCh38
rs763829353 Chr.14: 32092879-32092879 on Build GRCh38
rs774877467 Chr.14: 32093046-32093046 on Build GRCh38
rs372956846 Chr.14: 32092714-32092714 on Build GRCh38
rs369766568 Chr.14: 32092652-32092652 on Build GRCh38
rs148226075 Chr.14: 32092629-32092629 on Build GRCh38
rs373593195 Chr.14: 32092989-32092989 on Build GRCh38
rs746406520 Chr.14: 32093028-32093028 on Build GRCh38
rs61739165 Chr.14: 32092847-32092847 on Build GRCh38
rs748591050 Chr.14: 32092803-32092803 on Build GRCh38
rs372988874 Chr.14: 32092670-32092670 on Build GRCh38
rs760250754 Chr.14: 32092970-32092970 on Build GRCh38
rs773213438 Chr.14: 32093077-32093077 on Build GRCh38
rs771504460 Chr.14: 32093045-32093045 on Build GRCh38
rs778857617 Chr.14: 32092661-32092661 on Build GRCh38
rs748344975 Chr.14: 32092922-32092922 on Build GRCh38
rs770127574 Chr.14: 32092928-32092928 on Build GRCh38
rs546529140 Chr.14: 32092800-32092800 on Build GRCh38
rs761810198 Chr.14: 32092988-32092988 on Build GRCh38
rs753743151 Chr.14: 32092884-32092884 on Build GRCh38
rs146396734 Chr.14: 32092623-32092623 on Build GRCh38
rs768060067 Chr.14: 32092901-32092901 on Build GRCh38
rs772162945 Chr.14: 32092665-32092665 on Build GRCh38
rs371137927 Chr.14: 32093021-32093021 on Build GRCh38
rs763371966 Chr.14: 32092819-32092819 on Build GRCh38
rs769083114 Chr.14: 32092694-32092694 on Build GRCh38
rs777191722 Chr.14: 32092695-32092695 on Build GRCh38
rs749791343 Chr.14: 32092949-32092949 on Build GRCh38
rs754642213 Chr.14: 32092726-32092726 on Build GRCh38
rs148538118 Chr.14: 32093023-32093023 on Build GRCh38
rs767200354 Chr.14: 32092854-32092854 on Build GRCh38
rs778010378 Chr.14: 32092931-32092931 on Build GRCh38
rs367605056 Chr.14: 32092826-32092826 on Build GRCh38
rs141134108 Chr.14: 32092717-32092717 on Build GRCh38
rs755978813 Chr.14: 32092740-32092740 on Build GRCh38
rs139895818 Chr.14: 32093004-32093004 on Build GRCh38
rs773413531 Chr.14: 32092592-32092592 on Build GRCh38
rs191278032 Chr.14: 32092703-32092703 on Build GRCh38
rs553456371 Chr.14: 32092998-32092998 on Build GRCh38
rs201765085 Chr.14: 32093048-32093048 on Build GRCh38
rs61737878 Chr.14: 32093018-32093018 on Build GRCh38
rs769066633 Chr.14: 32092792-32092792 on Build GRCh38
rs757199082 Chr.14: 32092885-32092885 on Build GRCh38
rs186741221 Chr.14: 32092877-32092877 on Build GRCh38
rs747218960 Chr.14: 32092788-32092788 on Build GRCh38
rs780293390 Chr.14: 32092786-32092786 on Build GRCh38
rs748023246 Chr.14: 32092640-32092640 on Build GRCh38

View Full Product Details