Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74885324-74885850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLST
Entrez Gene ID
1743
Gene Name
dihydrolipoamide S-succinyltransferase
Gene Aliases
DLTS
UniGene
Hs.525459
Gene Chromosome Location
Chr.14: 74881891-74903747 on Build GRCh38
Transcripts
NM_001244883.1 NM_001933.4 NR_033814.1 NR_045209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641885 c.111C>T p.C37C Chr.14: 74885599-74885599 on Build GRCh38
1477769 c.98G>T p.G33V Chr.14: 74885586-74885586 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs999820024 Chr.14: 74885479-74885479 on Build GRCh38
rs1050456278 Chr.14: 74885367-74885367 on Build GRCh38
rs1049825632 Chr.14: 74885729-74885729 on Build GRCh38
rs760956799 Chr.14: 74885646-74885646 on Build GRCh38
rs541447476 Chr.14: 74885571-74885571 on Build GRCh38
rs370164953 Chr.14: 74885668-74885668 on Build GRCh38
rs754353111 Chr.14: 74885563-74885564 on Build GRCh38
rs555769465 Chr.14: 74885346-74885346 on Build GRCh38
rs781079626 Chr.14: 74885597-74885597 on Build GRCh38
rs961690259 Chr.14: 74885532-74885532 on Build GRCh38
rs932191172 Chr.14: 74885344-74885344 on Build GRCh38
rs765672184 Chr.14: 74885647-74885647 on Build GRCh38
rs763539320 Chr.14: 74885669-74885669 on Build GRCh38
rs774820919 Chr.14: 74885613-74885613 on Build GRCh38
rs184192386 Chr.14: 74885740-74885740 on Build GRCh38
rs896292032 Chr.14: 74885502-74885502 on Build GRCh38
rs374171608 Chr.14: 74885708-74885708 on Build GRCh38
rs564406773 Chr.14: 74885393-74885393 on Build GRCh38
rs763744622 Chr.14: 74885672-74885672 on Build GRCh38
rs61755652 Chr.14: 74885633-74885633 on Build GRCh38
rs1012084800 Chr.14: 74885759-74885760 on Build GRCh38
rs1034961234 Chr.14: 74885488-74885488 on Build GRCh38
rs572331471 Chr.14: 74885354-74885354 on Build GRCh38
rs765609125 Chr.14: 74885647-74885647 on Build GRCh38
rs752192577 Chr.14: 74885549-74885549 on Build GRCh38
rs757856651 Chr.14: 74885624-74885625 on Build GRCh38
rs901225575 Chr.14: 74885358-74885358 on Build GRCh38
rs938720829 Chr.14: 74885743-74885743 on Build GRCh38
rs947385660 Chr.14: 74885441-74885441 on Build GRCh38
rs1028306749 Chr.14: 74885406-74885406 on Build GRCh38
rs926423403 Chr.14: 74885628-74885628 on Build GRCh38
rs752262014 Chr.14: 74885821-74885821 on Build GRCh38
rs764816634 Chr.14: 74885548-74885548 on Build GRCh38
rs1012112183 Chr.14: 74885531-74885531 on Build GRCh38
rs984544378 Chr.14: 74885596-74885596 on Build GRCh38
rs757983422 Chr.14: 74885566-74885566 on Build GRCh38
rs887620381 Chr.14: 74885786-74885786 on Build GRCh38
rs1017595237 Chr.14: 74885475-74885475 on Build GRCh38
rs917422302 Chr.14: 74885603-74885603 on Build GRCh38
rs998040193 Chr.14: 74885689-74885689 on Build GRCh38
rs77722908 Chr.14: 74885791-74885791 on Build GRCh38
rs1056773098 Chr.14: 74885750-74885750 on Build GRCh38
rs954163125 Chr.14: 74885449-74885449 on Build GRCh38
rs767944255 Chr.14: 74885630-74885630 on Build GRCh38
rs547723720 Chr.14: 74885779-74885779 on Build GRCh38
rs763701838 Chr.14: 74885553-74885555 on Build GRCh38
rs1005961748 Chr.14: 74885765-74885765 on Build GRCh38
rs903191600 Chr.14: 74885476-74885476 on Build GRCh38
rs549990314 Chr.14: 74885820-74885820 on Build GRCh38
rs981845288 Chr.14: 74885619-74885619 on Build GRCh38
rs961999583 Chr.14: 74885806-74885806 on Build GRCh38
rs577823850 Chr.14: 74885447-74885447 on Build GRCh38
rs762352613 Chr.14: 74885624-74885624 on Build GRCh38
rs915105266 Chr.14: 74885645-74885645 on Build GRCh38
rs749366879 Chr.14: 74885605-74885605 on Build GRCh38
rs776824211 Chr.14: 74885514-74885514 on Build GRCh38
rs530814310 Chr.14: 74885589-74885589 on Build GRCh38
rs768785226 Chr.14: 74885608-74885608 on Build GRCh38
rs144519222 Chr.14: 74885602-74885602 on Build GRCh38
rs527250050 Chr.14: 74885772-74885772 on Build GRCh38
rs750855393 Chr.14: 74885659-74885660 on Build GRCh38
rs187811624 Chr.14: 74885392-74885392 on Build GRCh38
rs562789807 Chr.14: 74885387-74885387 on Build GRCh38
rs192940527 Chr.14: 74885661-74885661 on Build GRCh38
rs745680013 Chr.14: 74885599-74885599 on Build GRCh38
rs754623219 Chr.14: 74885547-74885547 on Build GRCh38
rs539744001 Chr.14: 74885793-74885793 on Build GRCh38
rs984165400 Chr.14: 74885469-74885469 on Build GRCh38
rs200699875 Chr.14: 74885587-74885587 on Build GRCh38
rs896436864 Chr.14: 74885756-74885760 on Build GRCh38
rs368214083 Chr.14: 74885593-74885593 on Build GRCh38

View Full Product Details