Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122995641-122996154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF5
Entrez Gene ID
64376
Gene Name
IKAROS family zinc finger 5
Gene Aliases
PEGASUS, ZNFN1A5
UniGene
Hs.501289
Gene Chromosome Location
Chr.10: 122990806-123008850 on Build GRCh38
Transcripts
NM_001271840.1 NR_073475.1 NR_073476.1 NR_073477.1 NR_073478.1 NR_073479.1 XM_006717947.3 XM_017016550.1 XM_017016551.1 XM_017016552.1 XM_017016553.1 XM_017016554.1 XM_017016555.1

Gene Symbol
PSTK
Entrez Gene ID
118672
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
C10orf89
UniGene
Hs.281004
Gene Chromosome Location
Chr.10: 122980040-122997519 on Build GRCh38
Transcripts
NM_001271840.1 NR_073475.1 NR_073476.1 NR_073477.1 NR_073478.1 NR_073479.1 XM_006717947.3 XM_017016550.1 XM_017016551.1 XM_017016552.1 XM_017016553.1 XM_017016554.1 XM_017016555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3435366 c.186C>T p.S62S Chr.10: 122996124-122996124 on Build GRCh38
4393059 c.294T>G p.I98M Chr.10: 122996016-122996016 on Build GRCh38
6684815 c.289C>T p.L97F Chr.10: 122996021-122996021 on Build GRCh38
1743554 c.290T>A p.L97H Chr.10: 122996020-122996020 on Build GRCh38
5636665 c.266A>G p.Y89C Chr.10: 122996044-122996044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140339345 Chr.10: 122995663-122995663 on Build GRCh38
rs562676818 Chr.10: 122995824-122995824 on Build GRCh38
rs1013084627 Chr.10: 122995800-122995800 on Build GRCh38
rs753604528 Chr.10: 122996100-122996100 on Build GRCh38
rs1052824712 Chr.10: 122995972-122995972 on Build GRCh38
rs375967070 Chr.10: 122996056-122996056 on Build GRCh38
rs930353310 Chr.10: 122995851-122995851 on Build GRCh38
rs1035760411 Chr.10: 122995774-122995774 on Build GRCh38
rs8192670 Chr.10: 122995861-122995861 on Build GRCh38
rs780766740 Chr.10: 122995833-122995833 on Build GRCh38
rs998958961 Chr.10: 122995692-122995692 on Build GRCh38
rs928202902 Chr.10: 122995782-122995782 on Build GRCh38
rs776917967 Chr.10: 122996042-122996042 on Build GRCh38
rs756653528 Chr.10: 122995773-122995773 on Build GRCh38
rs752768334 Chr.10: 122996058-122996058 on Build GRCh38
rs779707011 Chr.10: 122995981-122995981 on Build GRCh38
rs747607329 Chr.10: 122996126-122996126 on Build GRCh38
rs867920894 Chr.10: 122996071-122996071 on Build GRCh38
rs772732569 Chr.10: 122996127-122996127 on Build GRCh38
rs761056675 Chr.10: 122996028-122996028 on Build GRCh38
rs765356482 Chr.10: 122996057-122996057 on Build GRCh38
rs187796573 Chr.10: 122996103-122996103 on Build GRCh38
rs1010948953 Chr.10: 122995736-122995736 on Build GRCh38
rs1023680298 Chr.10: 122995819-122995819 on Build GRCh38
rs1031037843 Chr.10: 122995694-122995694 on Build GRCh38
rs919614075 Chr.10: 122995865-122995865 on Build GRCh38
rs944505958 Chr.10: 122995880-122995880 on Build GRCh38
rs748861946 Chr.10: 122995982-122995982 on Build GRCh38
rs369032187 Chr.10: 122995956-122995956 on Build GRCh38
rs373533220 Chr.10: 122996106-122996106 on Build GRCh38
rs969542968 Chr.10: 122995763-122995763 on Build GRCh38
rs755978407 Chr.10: 122996093-122996093 on Build GRCh38
rs748733524 Chr.10: 122996008-122996008 on Build GRCh38
rs999934312 Chr.10: 122995688-122995688 on Build GRCh38
rs75871484 Chr.10: 122995822-122995822 on Build GRCh38
rs773465857 Chr.10: 122996022-122996022 on Build GRCh38
rs769754710 Chr.10: 122995873-122995873 on Build GRCh38
rs746522607 Chr.10: 122995728-122995728 on Build GRCh38
rs1023663546 Chr.10: 122996092-122996092 on Build GRCh38
rs200896102 Chr.10: 122996018-122996018 on Build GRCh38
rs745608675 Chr.10: 122995967-122995967 on Build GRCh38
rs903125912 Chr.10: 122995682-122995682 on Build GRCh38
rs761741165 Chr.10: 122996064-122996064 on Build GRCh38
rs8192669 Chr.10: 122996078-122996078 on Build GRCh38
rs777344725 Chr.10: 122995660-122995660 on Build GRCh38
rs773986869 Chr.10: 122996006-122996006 on Build GRCh38
rs768331397 Chr.10: 122995988-122995988 on Build GRCh38
rs757171051 Chr.10: 122995955-122995955 on Build GRCh38
rs751544965 Chr.10: 122995943-122995943 on Build GRCh38
rs779691190 Chr.10: 122996094-122996094 on Build GRCh38
rs1023737080 Chr.10: 122995753-122995753 on Build GRCh38
rs201785739 Chr.10: 122995970-122995970 on Build GRCh38
rs145388999 Chr.10: 122995734-122995734 on Build GRCh38
rs866294934 Chr.10: 122995937-122995937 on Build GRCh38
rs548317241 Chr.10: 122995922-122995922 on Build GRCh38
rs778308007 Chr.10: 122996130-122996130 on Build GRCh38
rs766544603 Chr.10: 122996035-122996035 on Build GRCh38
rs767430220 Chr.10: 122996077-122996077 on Build GRCh38
rs1035202792 Chr.10: 122996117-122996117 on Build GRCh38
rs903093734 Chr.10: 122995936-122995936 on Build GRCh38
rs776948929 Chr.10: 122996037-122996037 on Build GRCh38

View Full Product Details