Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27634883-27635436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPN1
Entrez Gene ID
11321
Gene Name
GPN-loop GTPase 1
Gene Aliases
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1
UniGene
Hs.18259
Gene Chromosome Location
Chr.2: 27628648-27650846 on Build GRCh38
Transcripts
NM_001145047.1 NM_001145048.1 NM_001145049.1 NM_007266.3 NR_026735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1690337 c.442C>T p.P148S Chr.2: 27635152-27635152 on Build GRCh38
7424485 c.429+2T>G p.? Chr.2: 27634926-27634926 on Build GRCh38
3580738 c.473C>T p.S158L Chr.2: 27635183-27635183 on Build GRCh38
6212769 c.496T>C p.F166L Chr.2: 27635206-27635206 on Build GRCh38
6251582 c.395G>T p.W132L Chr.2: 27634890-27634890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039653711 Chr.2: 27635036-27635036 on Build GRCh38
rs1053476327 Chr.2: 27635048-27635048 on Build GRCh38
rs149103939 Chr.2: 27634947-27634947 on Build GRCh38
rs749472671 Chr.2: 27634911-27634911 on Build GRCh38
rs370148886 Chr.2: 27635098-27635098 on Build GRCh38
rs764213461 Chr.2: 27635142-27635142 on Build GRCh38
rs772961209 Chr.2: 27635205-27635205 on Build GRCh38
rs1033529248 Chr.2: 27635159-27635159 on Build GRCh38
rs910063077 Chr.2: 27635184-27635184 on Build GRCh38
rs569254787 Chr.2: 27635229-27635229 on Build GRCh38
rs76683865 Chr.2: 27635245-27635245 on Build GRCh38
rs769311750 Chr.2: 27635204-27635204 on Build GRCh38
rs147884292 Chr.2: 27634905-27634905 on Build GRCh38
rs768591135 Chr.2: 27635166-27635166 on Build GRCh38
rs1039935454 Chr.2: 27634907-27634907 on Build GRCh38
rs189276904 Chr.2: 27635317-27635317 on Build GRCh38
rs544242157 Chr.2: 27635280-27635280 on Build GRCh38
rs1041383912 Chr.2: 27634934-27634934 on Build GRCh38
rs753972498 Chr.2: 27635144-27635144 on Build GRCh38
rs753846808 Chr.2: 27635275-27635275 on Build GRCh38
rs776136212 Chr.2: 27634951-27634951 on Build GRCh38
rs1024212943 Chr.2: 27635073-27635073 on Build GRCh38
rs549521802 Chr.2: 27635301-27635301 on Build GRCh38
rs549596761 Chr.2: 27635300-27635301 on Build GRCh38
rs933659640 Chr.2: 27634925-27634925 on Build GRCh38
rs377432599 Chr.2: 27635097-27635097 on Build GRCh38
rs771889014 Chr.2: 27634931-27634931 on Build GRCh38
rs533399321 Chr.2: 27635370-27635370 on Build GRCh38
rs974950821 Chr.2: 27635408-27635408 on Build GRCh38
rs752961270 Chr.2: 27635287-27635289 on Build GRCh38
rs965216040 Chr.2: 27635348-27635349 on Build GRCh38
rs746085163 Chr.2: 27635244-27635244 on Build GRCh38
rs191969491 Chr.2: 27635083-27635083 on Build GRCh38
rs934194328 Chr.2: 27635265-27635265 on Build GRCh38
rs563206142 Chr.2: 27635349-27635349 on Build GRCh38
rs758917684 Chr.2: 27635009-27635009 on Build GRCh38
rs745626897 Chr.2: 27635155-27635155 on Build GRCh38
rs556437480 Chr.2: 27634984-27634984 on Build GRCh38
rs912079108 Chr.2: 27635013-27635013 on Build GRCh38
rs948594328 Chr.2: 27635413-27635413 on Build GRCh38
rs747754562 Chr.2: 27635195-27635195 on Build GRCh38
rs892220040 Chr.2: 27635059-27635059 on Build GRCh38
rs187152743 Chr.2: 27635278-27635278 on Build GRCh38
rs894329211 Chr.2: 27635367-27635367 on Build GRCh38
rs57185039 Chr.2: 27635316-27635317 on Build GRCh38
rs778030973 Chr.2: 27634908-27634908 on Build GRCh38
rs779251443 Chr.2: 27635148-27635148 on Build GRCh38
rs761715868 Chr.2: 27635286-27635287 on Build GRCh38
rs764767059 Chr.2: 27635270-27635270 on Build GRCh38
rs963292944 Chr.2: 27635170-27635170 on Build GRCh38
rs776050145 Chr.2: 27635168-27635168 on Build GRCh38
rs1054193568 Chr.2: 27635297-27635297 on Build GRCh38
rs758253873 Chr.2: 27635156-27635156 on Build GRCh38
rs571413552 Chr.2: 27635001-27635001 on Build GRCh38
rs762549282 Chr.2: 27635223-27635223 on Build GRCh38
rs866175048 Chr.2: 27635300-27635300 on Build GRCh38
rs767688223 Chr.2: 27635094-27635094 on Build GRCh38
rs775455347 Chr.2: 27634939-27634939 on Build GRCh38
rs867532768 Chr.2: 27635246-27635246 on Build GRCh38
rs778507785 Chr.2: 27634919-27634919 on Build GRCh38
rs746896927 Chr.2: 27635163-27635163 on Build GRCh38
rs35587616 Chr.2: 27635396-27635396 on Build GRCh38
rs545291861 Chr.2: 27635383-27635383 on Build GRCh38
rs759190290 Chr.2: 27635301-27635302 on Build GRCh38
rs759998196 Chr.2: 27635124-27635124 on Build GRCh38
rs935775883 Chr.2: 27635289-27635289 on Build GRCh38
rs943557635 Chr.2: 27635018-27635018 on Build GRCh38
rs924267959 Chr.2: 27635282-27635282 on Build GRCh38
rs757483208 Chr.2: 27635140-27635140 on Build GRCh38
rs894307602 Chr.2: 27635038-27635038 on Build GRCh38
rs367570364 Chr.2: 27635297-27635298 on Build GRCh38
rs745741590 Chr.2: 27634928-27634928 on Build GRCh38
rs761312699 Chr.2: 27634965-27634965 on Build GRCh38
rs866593660 Chr.2: 27635232-27635232 on Build GRCh38
rs186546771 Chr.2: 27635046-27635046 on Build GRCh38
rs182834315 Chr.2: 27635130-27635130 on Build GRCh38
rs567282738 Chr.2: 27634914-27634914 on Build GRCh38
rs368001974 Chr.2: 27635145-27635145 on Build GRCh38
rs371838887 Chr.2: 27635043-27635043 on Build GRCh38
rs994779583 Chr.2: 27635215-27635215 on Build GRCh38
rs4666009 Chr.2: 27635125-27635125 on Build GRCh38
rs201794640 Chr.2: 27634916-27634916 on Build GRCh38
rs571666168 Chr.2: 27635069-27635069 on Build GRCh38
rs779851096 Chr.2: 27635157-27635157 on Build GRCh38
rs964229236 Chr.2: 27635216-27635216 on Build GRCh38

View Full Product Details