Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13778263-13778838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf53
Entrez Gene ID
28974
Gene Name
chromosome 19 open reading frame 53
Gene Aliases
HSPC023, LYDG10
UniGene
Hs.231616
Gene Chromosome Location
Chr.19: 13774443-13778773 on Build GRCh38
Transcripts
NM_014047.2

SNP Detail

SNP ID Chromosome Location
rs1003995239 Chr.19: 13778398-13778398 on Build GRCh38
rs900295518 Chr.19: 13778293-13778293 on Build GRCh38
rs931698791 Chr.19: 13778707-13778707 on Build GRCh38
rs747782712 Chr.19: 13778305-13778305 on Build GRCh38
rs925073672 Chr.19: 13778717-13778717 on Build GRCh38
rs191906183 Chr.19: 13778579-13778579 on Build GRCh38
rs907516974 Chr.19: 13778481-13778481 on Build GRCh38
rs10416625 Chr.19: 13778575-13778575 on Build GRCh38
rs1013210740 Chr.19: 13778457-13778457 on Build GRCh38
rs8107914 Chr.19: 13778631-13778631 on Build GRCh38
rs1031609534 Chr.19: 13778648-13778648 on Build GRCh38
rs150552970 Chr.19: 13778365-13778365 on Build GRCh38
rs898242317 Chr.19: 13778789-13778789 on Build GRCh38
rs1019823504 Chr.19: 13778679-13778679 on Build GRCh38
rs965194317 Chr.19: 13778678-13778678 on Build GRCh38
rs2277947 Chr.19: 13778554-13778554 on Build GRCh38
rs951607826 Chr.19: 13778468-13778468 on Build GRCh38
rs981899164 Chr.19: 13778294-13778294 on Build GRCh38
rs187579039 Chr.19: 13778534-13778534 on Build GRCh38
rs1039290973 Chr.19: 13778347-13778347 on Build GRCh38
rs8108058 Chr.19: 13778775-13778775 on Build GRCh38
rs996410819 Chr.19: 13778336-13778336 on Build GRCh38
rs141745867 Chr.19: 13778311-13778311 on Build GRCh38
rs140825055 Chr.19: 13778509-13778509 on Build GRCh38
rs974635876 Chr.19: 13778438-13778438 on Build GRCh38
rs372889675 Chr.19: 13778333-13778333 on Build GRCh38
rs537387155 Chr.19: 13778546-13778546 on Build GRCh38
rs896385605 Chr.19: 13778749-13778749 on Build GRCh38
rs903757314 Chr.19: 13778341-13778341 on Build GRCh38
rs58655394 Chr.19: 13778550-13778551 on Build GRCh38
rs772037215 Chr.19: 13778523-13778523 on Build GRCh38
rs563415073 Chr.19: 13778342-13778342 on Build GRCh38
rs933589614 Chr.19: 13778533-13778533 on Build GRCh38
rs181505761 Chr.19: 13778388-13778388 on Build GRCh38
rs1016750263 Chr.19: 13778428-13778428 on Build GRCh38
rs546403374 Chr.19: 13778447-13778447 on Build GRCh38
rs1024224913 Chr.19: 13778397-13778397 on Build GRCh38
rs943594126 Chr.19: 13778330-13778330 on Build GRCh38
rs1023754321 Chr.19: 13778618-13778618 on Build GRCh38
rs921671608 Chr.19: 13778540-13778540 on Build GRCh38
rs566227691 Chr.19: 13778474-13778474 on Build GRCh38
rs10416998 Chr.19: 13778697-13778697 on Build GRCh38
rs914703612 Chr.19: 13778518-13778518 on Build GRCh38
rs147468887 Chr.19: 13778778-13778778 on Build GRCh38
rs908171849 Chr.19: 13778758-13778758 on Build GRCh38
rs923542959 Chr.19: 13778526-13778526 on Build GRCh38
rs867958718 Chr.19: 13778659-13778659 on Build GRCh38
rs368325308 Chr.19: 13778713-13778713 on Build GRCh38
rs540936301 Chr.19: 13778672-13778672 on Build GRCh38
rs1048029222 Chr.19: 13778748-13778748 on Build GRCh38
rs965775877 Chr.19: 13778685-13778685 on Build GRCh38
rs114060822 Chr.19: 13778360-13778360 on Build GRCh38
rs931861768 Chr.19: 13778543-13778543 on Build GRCh38
rs1003215287 Chr.19: 13778636-13778636 on Build GRCh38
rs949438594 Chr.19: 13778757-13778757 on Build GRCh38
rs1027081752 Chr.19: 13778638-13778638 on Build GRCh38
rs981464959 Chr.19: 13778493-13778493 on Build GRCh38
rs1054537822 Chr.19: 13778308-13778308 on Build GRCh38
rs943555921 Chr.19: 13778521-13778521 on Build GRCh38
rs542041835 Chr.19: 13778318-13778318 on Build GRCh38
rs935238310 Chr.19: 13778720-13778720 on Build GRCh38
rs555170779 Chr.19: 13778516-13778516 on Build GRCh38
rs921724928 Chr.19: 13778699-13778699 on Build GRCh38
rs974918356 Chr.19: 13778681-13778681 on Build GRCh38
rs1014000284 Chr.19: 13778608-13778608 on Build GRCh38
rs971755675 Chr.19: 13778482-13778482 on Build GRCh38
rs903788183 Chr.19: 13778553-13778553 on Build GRCh38
rs559451058 Chr.19: 13778307-13778307 on Build GRCh38
rs956094725 Chr.19: 13778508-13778508 on Build GRCh38
rs924691501 Chr.19: 13778297-13778297 on Build GRCh38
rs914649462 Chr.19: 13778658-13778658 on Build GRCh38
rs777674294 Chr.19: 13778539-13778539 on Build GRCh38
rs563516168 Chr.19: 13778657-13778657 on Build GRCh38
rs189118512 Chr.19: 13778376-13778376 on Build GRCh38
rs538045351 Chr.19: 13778649-13778649 on Build GRCh38
rs1043696384 Chr.19: 13778338-13778338 on Build GRCh38
rs924643342 Chr.19: 13778501-13778501 on Build GRCh38
rs564045781 Chr.19: 13778712-13778712 on Build GRCh38
rs144669075 Chr.19: 13778627-13778627 on Build GRCh38

View Full Product Details